Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01069291_m1
          See other DMBT1 GE Assays ›
          Gene Symbol:
          DMBT1
          Gene Name:
          deleted in malignant brain tumors 1
          Gene Aliases:
          GP340, SAG, muclin
          Chromosome Location:
          Chr.10: 122560665 - 122643736 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01069291_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: DMBT1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001320644.1 NP_001307573.1 38 - 39 4939 70
          NM_004406.2 NP_004397.2 25 - 26 3055 70
          NM_007329.2 NP_015568.2 38 - 39 4939 70
          NM_017579.2 NP_060049.2 37 - 38 4909 70
          XM_006717660.3 XP_006717723.1 40 - 41 5266 70
          XM_006717665.3 XP_006717728.1 39 - 40 5218 70
          XM_011539388.2 XP_011537690.1 41 - 42 5314 70
          XM_011539389.2 XP_011537691.1 41 - 42 5314 70
          XM_011539390.2 XP_011537692.1 40 - 41 5284 70
          XM_011539391.2 XP_011537693.1 40 - 41 5284 70
          XM_011539392.2 XP_011537694.1 40 - 41 5284 70
          XM_011539393.2 XP_011537695.1 40 - 41 5284 70
          XM_011539394.2 XP_011537696.1 40 - 41 5284 70
          XM_011539395.2 XP_011537697.1 40 - 41 5284 70
          XM_011539396.2 XP_011537698.1 40 - 41 5266 70
          XM_011539398.2 XP_011537700.1 39 - 40 5251 70
          XM_011539399.2 XP_011537701.1 39 - 40 5218 70
          XM_011539400.2 XP_011537702.1 40 - 41 4990 70
          XM_011539401.2 XP_011537703.1 38 - 39 4927 70
          XM_011539402.2 XP_011537704.1 38 - 39 4927 70
          XM_011539405.2 XP_011537707.1 38 - 39 4928 70
          XM_011539407.2 XP_011537709.1 38 - 39 4921 70
          XM_011539408.2 XP_011537710.1 38 - 39 4921 70
          XM_011539409.2 XP_011537711.1 38 - 39 4921 70
          XM_011539411.2 XP_011537713.1 37 - 38 4897 70
          XM_011539413.2 XP_011537715.1 35 - 36 4533 70
          XM_011539414.2 XP_011537716.1 25 - 26 3034 70
          XM_017015798.1 XP_016871287.1 26 - 27 3576 70
          GenBank mRNA AF159456.1 - 38 - 39 4939 70
          AJ000342.1 - 25 - 26 3055 70
          AJ243212.1 - 38 - 39 4939 70
          AJ243224.1 - 37 - 38 4909 70
          AJ297935.1 - 38 - 39 4920 70
          BC153299.1 - 22 - 23 2673 40147087 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1000009155.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC10000890.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene DMBT1

          Panther Classification:

          Molecular Function -

          serine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          pattern recognition receptor signaling pathway
          receptor-mediated endocytosis
          multicellular organism development
          protein transport
          epithelial cell differentiation
          induction of bacterial agglutination
          cellular protein metabolic process
          innate immune response
          defense response to virus
          scavenger receptor activity
          protein binding
          signaling pattern recognition receptor activity
          zymogen binding
          calcium-dependent protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR10010886.hg.1 ENST00000327438.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10010886.hg.1 ENST00000330163.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10010886.hg.1 ENST00000338354.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10010886.hg.1 ENST00000339712.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10010886.hg.1 ENST00000339871.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10010886.hg.1 ENST00000341278.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10010886.hg.1 ENST00000344338.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10010886.hg.1 ENST00000344467.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10010886.hg.1 ENST00000368909.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10010886.hg.1 ENST00000368915.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10010886.hg.1 ENST00000368942.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10010886.hg.1 ENST00000368953.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10010886.hg.1 ENST00000368955.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10010886.hg.1 ENST00000368956.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10010886.hg.1 NM_004406.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10010886.hg.1 NM_007329.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10010886.hg.1 NM_017579.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10010886.hg.1 TC10000890.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1000162770.hg.1 ENST00000330163.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1000162770.hg.1 ENST00000338354.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1000162770.hg.1 ENST00000344338.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1000162770.hg.1 ENST00000368909.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1000162770.hg.1 ENST00000368955.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1000162770.hg.1 ENST00000368956.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1000162770.hg.1 ENST00000619379.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000162770.hg.1 NM_004406.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000162770.hg.1 NM_007329.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000162770.hg.1 NM_017579.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000162770.hg.1 ENST00000330163.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1000162770.hg.1 ENST00000338354.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1000162770.hg.1 ENST00000344338.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1000162770.hg.1 ENST00000368909.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1000162770.hg.1 ENST00000368955.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1000162770.hg.1 ENST00000368956.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1000162770.hg.1 ENST00000619379.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000162770.hg.1 NM_004406.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000162770.hg.1 NM_007329.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000162770.hg.1 NM_017579.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000162770.hg.1 TC1000009155.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline