| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
ENST00000262844.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
ENST00000262844.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
ENST00000262844.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
ENST00000262844.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
ENST00000262844.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
ENST00000262844.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
ENST00000262844.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
ENST00000262844.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
ENST00000262844.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
ENST00000262844.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
ENST00000262844.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
ENST00000262844.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
ENST00000262844.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
ENST00000372057.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
ENST00000372057.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
ENST00000372057.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
ENST00000372057.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
ENST00000372057.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
ENST00000372057.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
ENST00000372057.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
ENST00000372057.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
ENST00000372057.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
ENST00000372057.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
ENST00000372057.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
ENST00000372059.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
ENST00000372059.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
ENST00000372059.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
ENST00000372059.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
ENST00000372059.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
ENST00000372059.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
ENST00000372059.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
ENST00000372059.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
ENST00000372059.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
ENST00000473662.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
ENST00000473662.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
ENST00000473662.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
ENST00000473662.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
ENST00000496695.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
ENST00000496695.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
ENST00000496695.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
NM_001025580.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
NM_001025580.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
NM_001025580.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
NM_001025580.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
NM_001025580.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
NM_001025580.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
NM_001025580.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
NM_001025580.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
NM_001025580.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
NM_001025580.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
NM_001025580.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
NM_001171689.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
NM_001171689.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
NM_001171689.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
NM_001171689.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
NM_001171689.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
NM_001171689.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
NM_001171689.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
NM_001171689.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
NM_001171689.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
NM_001171689.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
NM_001171689.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
NM_001171689.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
NM_015365.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
NM_015365.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
NM_015365.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
NM_015365.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
NM_015365.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
NM_015365.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
NM_015365.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
NM_015365.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
NM_015365.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
NM_015365.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
NM_015365.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010506.hg.1 |
NM_015365.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X017274.hg.1 |
TC0X001273.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X017274.hg.1 |
ENST00000262844.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X017274.hg.1 |
ENST00000372057.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X017274.hg.1 |
ENST00000372059.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X017274.hg.1 |
NM_001025580.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X017274.hg.1 |
NM_001171689.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X017274.hg.1 |
NM_015365.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
ENST00000262844.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
ENST00000262844.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
ENST00000262844.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
ENST00000262844.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
ENST00000262844.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
ENST00000262844.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
ENST00000262844.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
ENST00000262844.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
ENST00000262844.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
ENST00000262844.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
ENST00000262844.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
ENST00000262844.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
ENST00000262844.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
ENST00000372057.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
ENST00000372057.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
ENST00000372057.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
ENST00000372057.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
ENST00000372057.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
ENST00000372057.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
ENST00000372057.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
ENST00000372057.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
ENST00000372057.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
ENST00000372057.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
ENST00000372057.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
ENST00000372059.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
ENST00000372059.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
ENST00000372059.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
ENST00000372059.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
ENST00000372059.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
ENST00000372059.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
ENST00000372059.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
ENST00000372059.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
ENST00000372059.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
ENST00000473662.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
ENST00000473662.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
ENST00000473662.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
ENST00000473662.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
ENST00000496695.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
ENST00000496695.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
ENST00000496695.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
NM_001025580.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
NM_001025580.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
NM_001025580.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
NM_001025580.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
NM_001025580.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
NM_001025580.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
NM_001025580.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
NM_001025580.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
NM_001025580.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
NM_001025580.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
NM_001025580.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
NM_001171689.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
NM_001171689.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
NM_001171689.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
NM_001171689.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
NM_001171689.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
NM_001171689.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
NM_001171689.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
NM_001171689.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
NM_001171689.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
NM_001171689.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
NM_001171689.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
NM_001171689.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
NM_015365.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
NM_015365.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
NM_015365.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
NM_015365.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
NM_015365.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
NM_015365.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
NM_015365.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
NM_015365.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
NM_015365.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
NM_015365.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
NM_015365.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010506.hg.1 |
NM_015365.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00171632.hg.1 |
ENST00000262844.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00171632.hg.1 |
ENST00000372057.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00171632.hg.1 |
ENST00000372059.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00171632.hg.1 |
NM_001025580.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00171632.hg.1 |
NM_001171689.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00171632.hg.1 |
NM_015365.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00171632.hg.1 |
ENST00000262844.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00171632.hg.1 |
ENST00000372057.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00171632.hg.1 |
ENST00000372059.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00171632.hg.1 |
NM_001025580.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00171632.hg.1 |
NM_001171689.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00171632.hg.1 |
NM_015365.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00171632.hg.1 |
TC0X00010506.hg.1 |
Clariom D TC |