| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X017448.hg.1 |
ENST00000243213.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X017448.hg.1 |
ENST00000371936.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X017448.hg.1 |
ENST00000468224.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X017448.hg.1 |
NM_000640.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X017448.hg.1 |
TC0X001290.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010557.hg.1 |
ENST00000243213.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010557.hg.1 |
ENST00000243213.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010557.hg.1 |
ENST00000243213.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010557.hg.1 |
ENST00000243213.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010557.hg.1 |
ENST00000243213.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010557.hg.1 |
ENST00000243213.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010557.hg.1 |
ENST00000243213.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010557.hg.1 |
ENST00000243213.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010557.hg.1 |
ENST00000243213.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010557.hg.1 |
ENST00000243213.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010557.hg.1 |
ENST00000243213.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010557.hg.1 |
ENST00000243213.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010557.hg.1 |
ENST00000243213.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010557.hg.1 |
ENST00000371936.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010557.hg.1 |
ENST00000371936.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010557.hg.1 |
ENST00000371936.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010557.hg.1 |
ENST00000371936.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010557.hg.1 |
ENST00000371936.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010557.hg.1 |
ENST00000371936.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010557.hg.1 |
ENST00000371936.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010557.hg.1 |
ENST00000371936.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010557.hg.1 |
ENST00000371936.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010557.hg.1 |
ENST00000371936.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010557.hg.1 |
ENST00000371936.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010557.hg.1 |
ENST00000371936.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010557.hg.1 |
ENST00000371936.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010557.hg.1 |
ENST00000371936.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010557.hg.1 |
ENST00000371936.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010557.hg.1 |
ENST00000468224.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010557.hg.1 |
ENST00000468224.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010557.hg.1 |
ENST00000468224.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010557.hg.1 |
ENST00000468224.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010557.hg.1 |
ENST00000468224.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010557.hg.1 |
ENST00000468224.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010557.hg.1 |
ENST00000468224.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010557.hg.1 |
NM_000640.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010557.hg.1 |
NM_000640.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010557.hg.1 |
NM_000640.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010557.hg.1 |
NM_000640.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010557.hg.1 |
NM_000640.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010557.hg.1 |
NM_000640.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010557.hg.1 |
NM_000640.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010557.hg.1 |
NM_000640.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010557.hg.1 |
NM_000640.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010557.hg.1 |
NM_000640.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010557.hg.1 |
NM_000640.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010557.hg.1 |
NM_000640.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010557.hg.1 |
NM_000640.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010557.hg.1 |
ENST00000243213.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010557.hg.1 |
ENST00000243213.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010557.hg.1 |
ENST00000243213.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010557.hg.1 |
ENST00000243213.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010557.hg.1 |
ENST00000243213.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010557.hg.1 |
ENST00000243213.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010557.hg.1 |
ENST00000243213.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010557.hg.1 |
ENST00000243213.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010557.hg.1 |
ENST00000243213.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010557.hg.1 |
ENST00000243213.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010557.hg.1 |
ENST00000243213.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010557.hg.1 |
ENST00000243213.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010557.hg.1 |
ENST00000243213.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010557.hg.1 |
ENST00000371936.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010557.hg.1 |
ENST00000371936.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010557.hg.1 |
ENST00000371936.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010557.hg.1 |
ENST00000371936.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010557.hg.1 |
ENST00000371936.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010557.hg.1 |
ENST00000371936.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010557.hg.1 |
ENST00000371936.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010557.hg.1 |
ENST00000371936.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010557.hg.1 |
ENST00000371936.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010557.hg.1 |
ENST00000371936.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010557.hg.1 |
ENST00000371936.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010557.hg.1 |
ENST00000371936.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010557.hg.1 |
ENST00000371936.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010557.hg.1 |
ENST00000371936.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010557.hg.1 |
ENST00000371936.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010557.hg.1 |
ENST00000468224.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010557.hg.1 |
ENST00000468224.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010557.hg.1 |
ENST00000468224.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010557.hg.1 |
ENST00000468224.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010557.hg.1 |
ENST00000468224.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010557.hg.1 |
ENST00000468224.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010557.hg.1 |
ENST00000468224.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010557.hg.1 |
NM_000640.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010557.hg.1 |
NM_000640.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010557.hg.1 |
NM_000640.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010557.hg.1 |
NM_000640.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010557.hg.1 |
NM_000640.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010557.hg.1 |
NM_000640.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010557.hg.1 |
NM_000640.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010557.hg.1 |
NM_000640.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010557.hg.1 |
NM_000640.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010557.hg.1 |
NM_000640.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010557.hg.1 |
NM_000640.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010557.hg.1 |
NM_000640.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010557.hg.1 |
NM_000640.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00171916.hg.1 |
ENST00000243213.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00171916.hg.1 |
ENST00000371936.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00171916.hg.1 |
ENST00000468224.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00171916.hg.1 |
NM_000640.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00171916.hg.1 |
ENST00000243213.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00171916.hg.1 |
ENST00000371936.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00171916.hg.1 |
ENST00000468224.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00171916.hg.1 |
NM_000640.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00171916.hg.1 |
TC0X00010557.hg.1 |
Clariom D TC |