Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01071872_m1
          See other PPP2R5B GE Assays ›
          Gene Symbol:
          PPP2R5B
          Gene Name:
          protein phosphatase 2 regulatory subunit B'beta
          Gene Aliases:
          B56B, PR61B
          Chromosome Location:
          Chr.11: 64917553 - 64934478 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01071872_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PPP2R5B
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_006244.3 NP_006235.1 3 - 4 1017 66
          XM_011545132.2 XP_011543434.1 4 - 5 450 66
          XM_011545133.1 XP_011543435.1 5 - 6 570 66
          XM_011545134.2 XP_011543436.1 3 - 4 1856 66
          GenBank mRNA AK092730.1 - 3 - 4 1001 66
          BC045619.1 - 3 - 4 1011 5294756 66
          L42374.1 - 3 - 4 720 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1100013036.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC11000611.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Protein Phosphatase Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene PPP2R5B

          Panther Classification:

          Molecular Function -

          protein phosphatase

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of receptor activity
          positive regulation of neuron projection development
          regulation of phosphatidylinositol 3-kinase signaling
          positive regulation of protein complex assembly
          regulation of protein autophosphorylation
          regulation of protein phosphatase type 2A activity
          IRE1-mediated unfolded protein response
          positive regulation of transcription from RNA polymerase II promoter
          regulation of peptidyl-tyrosine phosphorylation
          positive regulation of sequence-specific DNA binding transcription factor activity
          positive regulation of neurotrophin TRK receptor signaling pathway
          negative regulation of G0 to G1 transition
          positive regulation of cell cycle arrest
          cellular response to growth factor stimulus
          protein binding
          protein phosphatase type 2A regulator activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1100013036.hg.1 ENST00000164133.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100013036.hg.1 ENST00000164133.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100013036.hg.1 ENST00000164133.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100013036.hg.1 ENST00000164133.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100013036.hg.1 ENST00000164133.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100013036.hg.1 ENST00000164133.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100013036.hg.1 ENST00000164133.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100013036.hg.1 ENST00000164133.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100013036.hg.1 ENST00000164133.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100013036.hg.1 ENST00000164133.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100013036.hg.1 ENST00000164133.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100013036.hg.1 ENST00000164133.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100013036.hg.1 ENST00000164133.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100013036.hg.1 ENST00000164133.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100013036.hg.1 ENST00000164133.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100013036.hg.1 ENST00000164133.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100013036.hg.1 ENST00000164133.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100013036.hg.1 ENST00000164133.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100013036.hg.1 ENST00000528530.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100013036.hg.1 ENST00000528530.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100013036.hg.1 ENST00000528530.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100013036.hg.1 ENST00000528530.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100013036.hg.1 ENST00000532850.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100013036.hg.1 ENST00000532850.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100013036.hg.1 ENST00000532850.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100013036.hg.1 ENST00000532850.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100013036.hg.1 ENST00000532850.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100013036.hg.1 NM_006244.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100013036.hg.1 NM_006244.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100013036.hg.1 NM_006244.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100013036.hg.1 NM_006244.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100013036.hg.1 NM_006244.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100013036.hg.1 NM_006244.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100013036.hg.1 NM_006244.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100013036.hg.1 NM_006244.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100013036.hg.1 NM_006244.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100013036.hg.1 NM_006244.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100013036.hg.1 NM_006244.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100013036.hg.1 NM_006244.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100013036.hg.1 NM_006244.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100013036.hg.1 NM_006244.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100013036.hg.1 NM_006244.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100013036.hg.1 NM_006244.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100013036.hg.1 NM_006244.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100013036.hg.1 NM_006244.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11007508.hg.1 ENST00000164133.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11007508.hg.1 ENST00000359279.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11007508.hg.1 ENST00000527441.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11007508.hg.1 NM_006244.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11007508.hg.1 TC11000611.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1100201137.hg.1 ENST00000164133.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1100201137.hg.1 ENST00000528530.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100201137.hg.1 ENST00000532850.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100201137.hg.1 NM_006244.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100201137.hg.1 ENST00000164133.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1100201137.hg.1 ENST00000528530.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100201137.hg.1 ENST00000532850.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100201137.hg.1 NM_006244.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100201137.hg.1 TC1100013036.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1100013036.hg.1 ENST00000164133.6 Ensembl
          Affymetrix Human Clariom D Assay TC1100013036.hg.1 ENST00000164133.6 Ensembl
          Affymetrix Human Clariom D Assay TC1100013036.hg.1 ENST00000164133.6 Ensembl
          Affymetrix Human Clariom D Assay TC1100013036.hg.1 ENST00000164133.6 Ensembl
          Affymetrix Human Clariom D Assay TC1100013036.hg.1 ENST00000164133.6 Ensembl
          Affymetrix Human Clariom D Assay TC1100013036.hg.1 ENST00000164133.6 Ensembl
          Affymetrix Human Clariom D Assay TC1100013036.hg.1 ENST00000164133.6 Ensembl
          Affymetrix Human Clariom D Assay TC1100013036.hg.1 ENST00000164133.6 Ensembl
          Affymetrix Human Clariom D Assay TC1100013036.hg.1 ENST00000164133.6 Ensembl
          Affymetrix Human Clariom D Assay TC1100013036.hg.1 ENST00000164133.6 Ensembl
          Affymetrix Human Clariom D Assay TC1100013036.hg.1 ENST00000164133.6 Ensembl
          Affymetrix Human Clariom D Assay TC1100013036.hg.1 ENST00000164133.6 Ensembl
          Affymetrix Human Clariom D Assay TC1100013036.hg.1 ENST00000164133.6 Ensembl
          Affymetrix Human Clariom D Assay TC1100013036.hg.1 ENST00000164133.6 Ensembl
          Affymetrix Human Clariom D Assay TC1100013036.hg.1 ENST00000164133.6 Ensembl
          Affymetrix Human Clariom D Assay TC1100013036.hg.1 ENST00000164133.6 Ensembl
          Affymetrix Human Clariom D Assay TC1100013036.hg.1 ENST00000164133.6 Ensembl
          Affymetrix Human Clariom D Assay TC1100013036.hg.1 ENST00000164133.6 Ensembl
          Affymetrix Human Clariom D Assay TC1100013036.hg.1 ENST00000528530.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100013036.hg.1 ENST00000528530.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100013036.hg.1 ENST00000528530.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100013036.hg.1 ENST00000528530.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100013036.hg.1 ENST00000532850.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100013036.hg.1 ENST00000532850.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100013036.hg.1 ENST00000532850.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100013036.hg.1 ENST00000532850.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100013036.hg.1 ENST00000532850.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100013036.hg.1 NM_006244.3 Refseq
          Affymetrix Human Clariom D Assay TC1100013036.hg.1 NM_006244.3 Refseq
          Affymetrix Human Clariom D Assay TC1100013036.hg.1 NM_006244.3 Refseq
          Affymetrix Human Clariom D Assay TC1100013036.hg.1 NM_006244.3 Refseq
          Affymetrix Human Clariom D Assay TC1100013036.hg.1 NM_006244.3 Refseq
          Affymetrix Human Clariom D Assay TC1100013036.hg.1 NM_006244.3 Refseq
          Affymetrix Human Clariom D Assay TC1100013036.hg.1 NM_006244.3 Refseq
          Affymetrix Human Clariom D Assay TC1100013036.hg.1 NM_006244.3 Refseq
          Affymetrix Human Clariom D Assay TC1100013036.hg.1 NM_006244.3 Refseq
          Affymetrix Human Clariom D Assay TC1100013036.hg.1 NM_006244.3 Refseq
          Affymetrix Human Clariom D Assay TC1100013036.hg.1 NM_006244.3 Refseq
          Affymetrix Human Clariom D Assay TC1100013036.hg.1 NM_006244.3 Refseq
          Affymetrix Human Clariom D Assay TC1100013036.hg.1 NM_006244.3 Refseq
          Affymetrix Human Clariom D Assay TC1100013036.hg.1 NM_006244.3 Refseq
          Affymetrix Human Clariom D Assay TC1100013036.hg.1 NM_006244.3 Refseq
          Affymetrix Human Clariom D Assay TC1100013036.hg.1 NM_006244.3 Refseq
          Affymetrix Human Clariom D Assay TC1100013036.hg.1 NM_006244.3 Refseq
          Affymetrix Human Clariom D Assay TC1100013036.hg.1 NM_006244.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline