Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01072273_g1
          See other EPHA2 GE Assays ›
          Gene Symbol:
          EPHA2
          Gene Name:
          EPH receptor A2
          Gene Aliases:
          ARCC2, CTPA, CTPP1, CTRCT6, ECK
          Chromosome Location:
          Chr.1: 16124337 - 16156104 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01072273_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: EPHA2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_017000534.1 XP_016856023.1 6 - 7 1698 93
          XM_017000535.1 XP_016856024.1 6 - 7 1658 93
          XM_017000536.1 XP_016856025.1 5 - 6 1506 93
          XM_017000537.1 XP_016856026.1 6 - 7 1600 93
          GenBank mRNA AK296788.1 - 5 - 6 751 93
          BC037166.2 - 6 - 7 1546 5090654 93
          M59371.1 - 6 - 7 1541 93

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100013028.hg.1

          Set Membership:

          Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          skeletal system development
          angiogenesis
          vasculogenesis
          osteoblast differentiation
          multicellular organism development
          intrinsic apoptotic signaling pathway in response to DNA damage
          regulation of lamellipodium assembly
          notochord formation
          viral process
          cell migration
          peptidyl-tyrosine phosphorylation
          neural tube development
          neuron differentiation
          keratinocyte differentiation
          osteoclast differentiation
          mammary gland epithelial cell proliferation
          regulation of cell adhesion mediated by integrin
          post-anal tail morphogenesis
          protein kinase B signaling
          regulation of blood vessel endothelial cell migration
          regulation of angiogenesis
          bone remodeling
          ephrin receptor signaling pathway
          axial mesoderm formation
          negative regulation of protein kinase B signaling
          notochord cell development
          cell chemotaxis
          branching involved in mammary gland duct morphogenesis
          lens fiber cell morphogenesis
          regulation of ERK1 and ERK2 cascade
          response to growth factor
          positive regulation of establishment of protein localization to plasma membrane
          activation of GTPase activity
          cell-cell adhesion
          transmembrane receptor protein tyrosine kinase activity
          ephrin receptor activity
          protein binding
          ATP binding
          cadherin binding involved in cell-cell adhesion

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0100013028.hg.1 ENST00000358432.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013028.hg.1 ENST00000358432.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013028.hg.1 ENST00000358432.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013028.hg.1 ENST00000358432.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013028.hg.1 ENST00000358432.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013028.hg.1 ENST00000358432.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013028.hg.1 ENST00000358432.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013028.hg.1 ENST00000358432.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013028.hg.1 ENST00000358432.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013028.hg.1 ENST00000358432.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013028.hg.1 ENST00000358432.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013028.hg.1 ENST00000358432.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013028.hg.1 ENST00000358432.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013028.hg.1 ENST00000358432.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013028.hg.1 ENST00000358432.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013028.hg.1 ENST00000358432.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013028.hg.1 ENST00000358432.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013028.hg.1 ENST00000358432.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013028.hg.1 ENST00000358432.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013028.hg.1 ENST00000358432.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013028.hg.1 ENST00000358432.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013028.hg.1 ENST00000461614.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013028.hg.1 ENST00000461614.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013028.hg.1 ENST00000461614.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013028.hg.1 ENST00000462805.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013028.hg.1 ENST00000462805.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013028.hg.1 ENST00000462805.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013028.hg.1 ENST00000480202.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013028.hg.1 ENST00000480202.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013028.hg.1 ENST00000480202.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013028.hg.1 ENST00000480202.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013028.hg.1 ENST00000480202.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013028.hg.1 NM_004431.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013028.hg.1 NM_004431.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013028.hg.1 NM_004431.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013028.hg.1 NM_004431.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013028.hg.1 NM_004431.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013028.hg.1 NM_004431.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013028.hg.1 NM_004431.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013028.hg.1 NM_004431.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013028.hg.1 NM_004431.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013028.hg.1 NM_004431.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013028.hg.1 NM_004431.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013028.hg.1 NM_004431.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013028.hg.1 NM_004431.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013028.hg.1 NM_004431.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013028.hg.1 NM_004431.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013028.hg.1 NM_004431.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013028.hg.1 NM_004431.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013028.hg.1 NM_004431.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013028.hg.1 NM_004431.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013028.hg.1 NM_004431.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013028.hg.1 NM_004431.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100196017.hg.1 ENST00000358432.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100196017.hg.1 ENST00000480202.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100196017.hg.1 NM_004431.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100196017.hg.1 ENST00000358432.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100196017.hg.1 ENST00000480202.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100196017.hg.1 NM_004431.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100196017.hg.1 TC0100013028.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0100013028.hg.1 ENST00000358432.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100013028.hg.1 ENST00000358432.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100013028.hg.1 ENST00000358432.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100013028.hg.1 ENST00000358432.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100013028.hg.1 ENST00000358432.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100013028.hg.1 ENST00000358432.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100013028.hg.1 ENST00000358432.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100013028.hg.1 ENST00000358432.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100013028.hg.1 ENST00000358432.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100013028.hg.1 ENST00000358432.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100013028.hg.1 ENST00000358432.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100013028.hg.1 ENST00000358432.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100013028.hg.1 ENST00000358432.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100013028.hg.1 ENST00000358432.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100013028.hg.1 ENST00000358432.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100013028.hg.1 ENST00000358432.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100013028.hg.1 ENST00000358432.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100013028.hg.1 ENST00000358432.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100013028.hg.1 ENST00000358432.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100013028.hg.1 ENST00000358432.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100013028.hg.1 ENST00000358432.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100013028.hg.1 ENST00000461614.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013028.hg.1 ENST00000461614.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013028.hg.1 ENST00000461614.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013028.hg.1 ENST00000462805.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013028.hg.1 ENST00000462805.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013028.hg.1 ENST00000462805.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013028.hg.1 ENST00000480202.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013028.hg.1 ENST00000480202.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013028.hg.1 ENST00000480202.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013028.hg.1 ENST00000480202.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013028.hg.1 ENST00000480202.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100013028.hg.1 NM_004431.4 Refseq
          Affymetrix Human Clariom D Assay TC0100013028.hg.1 NM_004431.4 Refseq
          Affymetrix Human Clariom D Assay TC0100013028.hg.1 NM_004431.4 Refseq
          Affymetrix Human Clariom D Assay TC0100013028.hg.1 NM_004431.4 Refseq
          Affymetrix Human Clariom D Assay TC0100013028.hg.1 NM_004431.4 Refseq
          Affymetrix Human Clariom D Assay TC0100013028.hg.1 NM_004431.4 Refseq
          Affymetrix Human Clariom D Assay TC0100013028.hg.1 NM_004431.4 Refseq
          Affymetrix Human Clariom D Assay TC0100013028.hg.1 NM_004431.4 Refseq
          Affymetrix Human Clariom D Assay TC0100013028.hg.1 NM_004431.4 Refseq
          Affymetrix Human Clariom D Assay TC0100013028.hg.1 NM_004431.4 Refseq
          Affymetrix Human Clariom D Assay TC0100013028.hg.1 NM_004431.4 Refseq
          Affymetrix Human Clariom D Assay TC0100013028.hg.1 NM_004431.4 Refseq
          Affymetrix Human Clariom D Assay TC0100013028.hg.1 NM_004431.4 Refseq
          Affymetrix Human Clariom D Assay TC0100013028.hg.1 NM_004431.4 Refseq
          Affymetrix Human Clariom D Assay TC0100013028.hg.1 NM_004431.4 Refseq
          Affymetrix Human Clariom D Assay TC0100013028.hg.1 NM_004431.4 Refseq
          Affymetrix Human Clariom D Assay TC0100013028.hg.1 NM_004431.4 Refseq
          Affymetrix Human Clariom D Assay TC0100013028.hg.1 NM_004431.4 Refseq
          Affymetrix Human Clariom D Assay TC0100013028.hg.1 NM_004431.4 Refseq
          Affymetrix Human Clariom D Assay TC0100013028.hg.1 NM_004431.4 Refseq
          Affymetrix Human Clariom D Assay TC0100013028.hg.1 NM_004431.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline