Search
Search

C7orf33| Gene Symbol: C7orf33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145304.3 | NP_660347.1 | 1 - 2 | 566 | 91 | |
| GenBank mRNA | BC021251.2 | - | 1 - 2 | 566 | 3352885 | 91 |
Set Membership: |
|