| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01060482.hg.1 |
ENST00000366597.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01060482.hg.1 |
ENST00000366598.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01060482.hg.1 |
ENST00000391854.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01060482.hg.1 |
ENST00000450593.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01060482.hg.1 |
ENST00000484517.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01060482.hg.1 |
NM_001098721.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01060482.hg.1 |
NM_001098722.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01060482.hg.1 |
NM_004485.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01060482.hg.1 |
TC01003995.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017834.hg.1 |
ENST00000366597.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017834.hg.1 |
ENST00000366597.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017834.hg.1 |
ENST00000366597.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017834.hg.1 |
ENST00000366597.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017834.hg.1 |
ENST00000366597.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017834.hg.1 |
ENST00000366598.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017834.hg.1 |
ENST00000366598.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017834.hg.1 |
ENST00000366598.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017834.hg.1 |
ENST00000366598.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017834.hg.1 |
ENST00000366598.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017834.hg.1 |
ENST00000391854.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017834.hg.1 |
ENST00000391854.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017834.hg.1 |
ENST00000391854.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017834.hg.1 |
ENST00000391854.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017834.hg.1 |
ENST00000391854.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017834.hg.1 |
ENST00000391854.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017834.hg.1 |
ENST00000391854.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017834.hg.1 |
ENST00000391854.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017834.hg.1 |
ENST00000391854.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017834.hg.1 |
ENST00000450593.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017834.hg.1 |
ENST00000450593.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017834.hg.1 |
ENST00000450593.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017834.hg.1 |
ENST00000450593.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017834.hg.1 |
ENST00000450593.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017834.hg.1 |
ENST00000450593.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017834.hg.1 |
ENST00000450593.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017834.hg.1 |
ENST00000450593.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017834.hg.1 |
ENST00000450593.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017834.hg.1 |
ENST00000484517.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017834.hg.1 |
ENST00000484517.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017834.hg.1 |
ENST00000484517.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017834.hg.1 |
ENST00000484517.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017834.hg.1 |
NM_001098721.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017834.hg.1 |
NM_001098721.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017834.hg.1 |
NM_001098721.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017834.hg.1 |
NM_001098721.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017834.hg.1 |
NM_001098721.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017834.hg.1 |
NM_001098721.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017834.hg.1 |
NM_001098721.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017834.hg.1 |
NM_001098721.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017834.hg.1 |
NM_001098721.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017834.hg.1 |
NM_001098722.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017834.hg.1 |
NM_001098722.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017834.hg.1 |
NM_001098722.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017834.hg.1 |
NM_001098722.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017834.hg.1 |
NM_001098722.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017834.hg.1 |
NM_001098722.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017834.hg.1 |
NM_001098722.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017834.hg.1 |
NM_001098722.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017834.hg.1 |
NM_001098722.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017834.hg.1 |
NM_004485.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017834.hg.1 |
NM_004485.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017834.hg.1 |
NM_004485.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017834.hg.1 |
NM_004485.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017834.hg.1 |
NM_004485.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017834.hg.1 |
NM_004485.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017834.hg.1 |
NM_004485.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100234544.hg.1 |
ENST00000366597.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100234544.hg.1 |
ENST00000366598.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100234544.hg.1 |
ENST00000391854.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100234544.hg.1 |
ENST00000450593.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100234544.hg.1 |
ENST00000484517.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100234544.hg.1 |
NM_001098721.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100234544.hg.1 |
NM_001098722.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100234544.hg.1 |
NM_004485.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100234544.hg.1 |
ENST00000366597.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100234544.hg.1 |
ENST00000366598.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100234544.hg.1 |
ENST00000391854.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100234544.hg.1 |
ENST00000450593.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100234544.hg.1 |
ENST00000484517.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100234544.hg.1 |
NM_001098721.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100234544.hg.1 |
NM_001098722.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100234544.hg.1 |
NM_004485.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100234544.hg.1 |
TC0100017834.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100017834.hg.1 |
ENST00000366597.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017834.hg.1 |
ENST00000366597.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017834.hg.1 |
ENST00000366597.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017834.hg.1 |
ENST00000366597.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017834.hg.1 |
ENST00000366597.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017834.hg.1 |
ENST00000366598.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017834.hg.1 |
ENST00000366598.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017834.hg.1 |
ENST00000366598.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017834.hg.1 |
ENST00000366598.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017834.hg.1 |
ENST00000366598.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017834.hg.1 |
ENST00000391854.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017834.hg.1 |
ENST00000391854.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017834.hg.1 |
ENST00000391854.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017834.hg.1 |
ENST00000391854.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017834.hg.1 |
ENST00000391854.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017834.hg.1 |
ENST00000391854.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017834.hg.1 |
ENST00000391854.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017834.hg.1 |
ENST00000391854.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017834.hg.1 |
ENST00000391854.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017834.hg.1 |
ENST00000450593.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017834.hg.1 |
ENST00000450593.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017834.hg.1 |
ENST00000450593.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017834.hg.1 |
ENST00000450593.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017834.hg.1 |
ENST00000450593.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017834.hg.1 |
ENST00000450593.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017834.hg.1 |
ENST00000450593.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017834.hg.1 |
ENST00000450593.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017834.hg.1 |
ENST00000450593.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017834.hg.1 |
ENST00000484517.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017834.hg.1 |
ENST00000484517.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017834.hg.1 |
ENST00000484517.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017834.hg.1 |
ENST00000484517.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017834.hg.1 |
NM_001098721.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017834.hg.1 |
NM_001098721.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017834.hg.1 |
NM_001098721.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017834.hg.1 |
NM_001098721.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017834.hg.1 |
NM_001098721.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017834.hg.1 |
NM_001098721.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017834.hg.1 |
NM_001098721.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017834.hg.1 |
NM_001098721.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017834.hg.1 |
NM_001098721.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017834.hg.1 |
NM_001098722.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017834.hg.1 |
NM_001098722.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017834.hg.1 |
NM_001098722.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017834.hg.1 |
NM_001098722.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017834.hg.1 |
NM_001098722.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017834.hg.1 |
NM_001098722.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017834.hg.1 |
NM_001098722.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017834.hg.1 |
NM_001098722.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017834.hg.1 |
NM_001098722.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017834.hg.1 |
NM_004485.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017834.hg.1 |
NM_004485.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017834.hg.1 |
NM_004485.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017834.hg.1 |
NM_004485.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017834.hg.1 |
NM_004485.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017834.hg.1 |
NM_004485.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100017834.hg.1 |
NM_004485.3 |
Refseq |