Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01076145_g1
          See other DIS3 GE Assays ›
          Gene Symbol:
          DIS3
          Gene Name:
          DIS3 homolog, exosome endoribonuclease and 3'-5' exoribonuclease
          Gene Aliases:
          2810028N01Rik, EXOSC11, KIAA1008, RRP44, dis3p
          Chromosome Location:
          Chr.13: 72755402 - 72782128 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01076145_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: DIS3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001128226.2 NP_001121698.1 12 - 13 1879 98
          NM_001322348.1 NP_001309277.1 11 - 12 1811 98
          NM_001322349.1 NP_001309278.1 13 - 14 2037 98
          NM_014953.4 NP_055768.3 12 - 13 1969 98
          GenBank mRNA AB001743.1 - 12 - 13 1700 98
          AB023225.1 - 12 - 13 1676 98
          AF330044.1 - 12 - 13 1709 98
          AK025634.1 - 10 - 11 1279 98
          AK314715.1 - 12 - 13 1785 98
          AL080158.1 - 6 - 7 696 98
          AL832266.1 - 12 - 13 1762 98
          BC038101.1 - 13 - 14 2008 4827328 98
          BC056143.1 - 12 - 13 1774 5505671 98

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1300009232.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC13000727.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene DIS3

          Panther Classification:

          Molecular Function -

          exoribonuclease

          Gene Ontology Categories:

          Function(s) Process(es)

          rRNA processing
          mRNA catabolic process
          rRNA catabolic process
          regulation of mRNA stability
          positive regulation of GTPase activity
          exonucleolytic nuclear-transcribed mRNA catabolic process involved in deadenylation-dependent decay
          CUT catabolic process
          RNA phosphodiester bond hydrolysis, exonucleolytic
          3'-5'-exoribonuclease activity
          RNA binding
          endonuclease activity
          guanyl-nucleotide exchange factor activity
          protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR13008377.hg.1 ENST00000377767.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR13008377.hg.1 ENST00000377780.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR13008377.hg.1 ENST00000545453.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR13008377.hg.1 NM_001128226.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR13008377.hg.1 NM_014953.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR13008377.hg.1 TC13000727.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1300162053.hg.1 ENST00000377767.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1300162053.hg.1 ENST00000377780.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1300162053.hg.1 ENST00000545453.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1300162053.hg.1 NM_001128226.2 Refseq
          Affymetrix Human Clariom D Assay PSR1300162053.hg.1 NM_014953.4 Refseq
          Affymetrix Human Clariom D Assay PSR1300162053.hg.1 ENST00000377767.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1300162053.hg.1 ENST00000377780.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1300162053.hg.1 ENST00000545453.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1300162053.hg.1 NM_001128226.2 Refseq
          Affymetrix Human Clariom D Assay PSR1300162053.hg.1 NM_014953.4 Refseq
          Affymetrix Human Clariom D Assay PSR1300162053.hg.1 TC1300009232.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline