Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01076259_g1
          See other TIAM2 GE Assays ›
          Gene Symbol:
          TIAM2
          Gene Name:
          T-cell lymphoma invasion and metastasis 2
          Gene Aliases:
          STEF, TIAM-2
          Chromosome Location:
          Chr.6: 155090289 - 155257723 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01076259_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TIAM2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001010927.2 NP_001010927.1 3 - 4 373 72
          NM_012454.3 NP_036586.2 16 - 17 3621 72
          GenBank mRNA AF120323.1 - 12 - 13 1591 72
          AF120324.1 - 3 - 4 349 72
          AK123859.1 - 17 - 18 3716 72
          AK125459.1 - 13 - 14 2080 72
          AK299723.1 - 3 - 4 370 72
          AK302860.1 - 12 - 13 1532 72
          AL122086.1 - 11 - 12 1450 72
          BC066979.1 - 3 - 4 370 72
          BC137275.1 - 15 - 16 3494 9021272 72
          BP199501.1 - 3 - 4 376 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0600014194.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC06004104.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene TIAM2

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of Rho protein signal transduction
          intracellular signal transduction
          positive regulation of apoptotic process
          positive regulation of GTPase activity
          regulation of small GTPase mediated signal transduction
          receptor signaling protein activity
          guanyl-nucleotide exchange factor activity
          Rho guanyl-nucleotide exchange factor activity
          GTPase activator activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR06013269.hg.1 ENST00000275246.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06013269.hg.1 ENST00000318981.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06013269.hg.1 ENST00000360366.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06013269.hg.1 ENST00000367174.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06013269.hg.1 ENST00000456144.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06013269.hg.1 ENST00000456877.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06013269.hg.1 ENST00000528391.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06013269.hg.1 ENST00000529824.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06013269.hg.1 NM_001010927.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06013269.hg.1 NM_012454.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06013269.hg.1 TC06004104.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0600199738.hg.1 ENST00000275246.11 Ensembl
          Affymetrix Human Clariom D Assay PSR0600199738.hg.1 ENST00000318981.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0600199738.hg.1 ENST00000360366.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0600199738.hg.1 ENST00000456877.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0600199738.hg.1 ENST00000462408.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0600199738.hg.1 ENST00000528391.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0600199738.hg.1 ENST00000528535.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0600199738.hg.1 ENST00000529824.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0600199738.hg.1 ENST00000543712.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0600199738.hg.1 ENST00000546145.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600199738.hg.1 NM_001010927.2 Refseq
          Affymetrix Human Clariom D Assay PSR0600199738.hg.1 NM_012454.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600199738.hg.1 ENST00000275246.11 Ensembl
          Affymetrix Human Clariom D Assay PSR0600199738.hg.1 ENST00000318981.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0600199738.hg.1 ENST00000360366.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0600199738.hg.1 ENST00000456877.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0600199738.hg.1 ENST00000462408.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0600199738.hg.1 ENST00000528391.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0600199738.hg.1 ENST00000528535.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0600199738.hg.1 ENST00000529824.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0600199738.hg.1 ENST00000543712.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0600199738.hg.1 ENST00000546145.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600199738.hg.1 NM_001010927.2 Refseq
          Affymetrix Human Clariom D Assay PSR0600199738.hg.1 NM_012454.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600199738.hg.1 TC0600014194.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline