| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
ENST00000262262.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
ENST00000262262.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
ENST00000262262.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
ENST00000262262.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
ENST00000262262.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
ENST00000262262.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
ENST00000262262.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
ENST00000262262.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
ENST00000262262.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
ENST00000262262.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
ENST00000262262.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
ENST00000262262.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
ENST00000391796.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
ENST00000391796.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
ENST00000391796.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
ENST00000391796.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
ENST00000391796.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
ENST00000391796.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
ENST00000391796.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
ENST00000391796.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
ENST00000421133.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
ENST00000421133.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
ENST00000421133.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
ENST00000421133.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
ENST00000421133.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
ENST00000421133.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
ENST00000421133.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
ENST00000421133.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
ENST00000421133.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
ENST00000436584.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
ENST00000436584.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
ENST00000436584.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
ENST00000436584.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
ENST00000436584.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
ENST00000436584.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
ENST00000436584.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
ENST00000436584.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
ENST00000436584.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
ENST00000598473.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
ENST00000598473.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
ENST00000598473.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
ENST00000598473.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
ENST00000598473.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
ENST00000598473.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
ENST00000598473.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
ENST00000598473.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
ENST00000598473.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
ENST00000598473.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
ENST00000598473.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
ENST00000598473.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
ENST00000600557.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
ENST00000600557.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
ENST00000600557.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
ENST00000600557.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
ENST00000600557.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
ENST00000600557.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
ENST00000601785.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
ENST00000601785.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
ENST00000601785.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
ENST00000601785.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
ENST00000601785.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
ENST00000601785.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
ENST00000601785.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
ENST00000601785.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
ENST00000601785.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
ENST00000601785.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
ENST00000601785.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
ENST00000601785.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
ENST00000601785.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
NM_001082618.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
NM_001082618.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
NM_001082618.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
NM_001082618.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
NM_001082618.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
NM_001082618.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
NM_001082618.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
NM_001082618.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
NM_001082618.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
NM_001082618.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
NM_001082618.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
NM_001177608.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
NM_001177608.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
NM_001177608.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
NM_001177608.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
NM_001177608.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
NM_001177608.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
NM_001177608.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
NM_001177608.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
NM_001772.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
NM_001772.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
NM_001772.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
NM_001772.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
NM_001772.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
NM_001772.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
NM_001772.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
NM_001772.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
NM_001772.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
NM_001772.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
NM_001772.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008633.hg.1 |
NM_001772.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
ENST00000262262.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
ENST00000262262.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
ENST00000262262.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
ENST00000262262.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
ENST00000262262.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
ENST00000262262.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
ENST00000262262.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
ENST00000262262.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
ENST00000262262.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
ENST00000262262.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
ENST00000262262.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
ENST00000262262.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
ENST00000391796.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
ENST00000391796.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
ENST00000391796.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
ENST00000391796.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
ENST00000391796.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
ENST00000391796.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
ENST00000391796.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
ENST00000391796.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
ENST00000421133.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
ENST00000421133.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
ENST00000421133.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
ENST00000421133.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
ENST00000421133.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
ENST00000421133.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
ENST00000421133.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
ENST00000421133.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
ENST00000421133.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
ENST00000436584.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
ENST00000436584.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
ENST00000436584.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
ENST00000436584.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
ENST00000436584.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
ENST00000436584.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
ENST00000436584.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
ENST00000436584.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
ENST00000436584.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
ENST00000598473.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
ENST00000598473.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
ENST00000598473.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
ENST00000598473.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
ENST00000598473.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
ENST00000598473.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
ENST00000598473.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
ENST00000598473.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
ENST00000598473.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
ENST00000598473.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
ENST00000598473.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
ENST00000598473.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
ENST00000600557.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
ENST00000600557.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
ENST00000600557.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
ENST00000600557.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
ENST00000600557.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
ENST00000600557.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
ENST00000601785.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
ENST00000601785.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
ENST00000601785.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
ENST00000601785.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
ENST00000601785.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
ENST00000601785.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
ENST00000601785.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
ENST00000601785.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
ENST00000601785.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
ENST00000601785.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
ENST00000601785.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
ENST00000601785.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
ENST00000601785.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
NM_001082618.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
NM_001082618.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
NM_001082618.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
NM_001082618.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
NM_001082618.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
NM_001082618.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
NM_001082618.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
NM_001082618.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
NM_001082618.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
NM_001082618.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
NM_001082618.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
NM_001177608.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
NM_001177608.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
NM_001177608.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
NM_001177608.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
NM_001177608.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
NM_001177608.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
NM_001177608.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
NM_001177608.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
NM_001772.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
NM_001772.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
NM_001772.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
NM_001772.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
NM_001772.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
NM_001772.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
NM_001772.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
NM_001772.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
NM_001772.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
NM_001772.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
NM_001772.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008633.hg.1 |
NM_001772.3 |
Refseq |