| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01046214.hg.1 |
ENST00000369763.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01046214.hg.1 |
ENST00000494675.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01046214.hg.1 |
NM_001006945.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01046214.hg.1 |
NM_018372.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01046214.hg.1 |
TC01002979.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01046217.hg.1 |
ENST00000369763.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01046217.hg.1 |
ENST00000485275.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01046217.hg.1 |
ENST00000494675.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01046217.hg.1 |
NM_001006945.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01046217.hg.1 |
NM_018372.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01046217.hg.1 |
TC01002979.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01046218.hg.1 |
ENST00000369763.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01046218.hg.1 |
ENST00000485275.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01046218.hg.1 |
ENST00000494675.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01046218.hg.1 |
NM_001006945.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01046218.hg.1 |
NM_018372.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01046218.hg.1 |
TC01002979.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015254.hg.1 |
ENST00000369763.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015254.hg.1 |
ENST00000369763.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015254.hg.1 |
ENST00000369763.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015254.hg.1 |
ENST00000369763.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015254.hg.1 |
ENST00000369763.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015254.hg.1 |
ENST00000369763.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015254.hg.1 |
ENST00000369763.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015254.hg.1 |
ENST00000369763.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015254.hg.1 |
ENST00000369763.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015254.hg.1 |
ENST00000369763.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015254.hg.1 |
ENST00000485275.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015254.hg.1 |
ENST00000485275.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015254.hg.1 |
ENST00000485275.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015254.hg.1 |
ENST00000485275.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015254.hg.1 |
ENST00000485275.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015254.hg.1 |
ENST00000494675.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015254.hg.1 |
ENST00000494675.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015254.hg.1 |
ENST00000494675.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015254.hg.1 |
ENST00000494675.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015254.hg.1 |
ENST00000494675.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015254.hg.1 |
ENST00000494675.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015254.hg.1 |
NM_001006945.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015254.hg.1 |
NM_001006945.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015254.hg.1 |
NM_001006945.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015254.hg.1 |
NM_001006945.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015254.hg.1 |
NM_001006945.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015254.hg.1 |
NM_001006945.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015254.hg.1 |
NM_018372.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015254.hg.1 |
NM_018372.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015254.hg.1 |
NM_018372.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015254.hg.1 |
NM_018372.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015254.hg.1 |
NM_018372.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015254.hg.1 |
NM_018372.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015254.hg.1 |
NM_018372.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015254.hg.1 |
NM_018372.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015254.hg.1 |
NM_018372.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01046215.hg.1 |
ENST00000369763.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01046215.hg.1 |
ENST00000485275.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01046215.hg.1 |
ENST00000494675.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01046215.hg.1 |
NM_001006945.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01046215.hg.1 |
NM_018372.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01046215.hg.1 |
TC01002979.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0100015254.hg.1 |
ENST00000369763.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015254.hg.1 |
ENST00000369763.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015254.hg.1 |
ENST00000369763.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015254.hg.1 |
ENST00000369763.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015254.hg.1 |
ENST00000369763.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015254.hg.1 |
ENST00000369763.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015254.hg.1 |
ENST00000369763.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015254.hg.1 |
ENST00000369763.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015254.hg.1 |
ENST00000369763.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015254.hg.1 |
ENST00000369763.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015254.hg.1 |
ENST00000485275.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015254.hg.1 |
ENST00000485275.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015254.hg.1 |
ENST00000485275.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015254.hg.1 |
ENST00000485275.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015254.hg.1 |
ENST00000485275.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015254.hg.1 |
ENST00000494675.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015254.hg.1 |
ENST00000494675.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015254.hg.1 |
ENST00000494675.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015254.hg.1 |
ENST00000494675.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015254.hg.1 |
ENST00000494675.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015254.hg.1 |
ENST00000494675.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015254.hg.1 |
NM_001006945.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015254.hg.1 |
NM_001006945.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015254.hg.1 |
NM_001006945.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015254.hg.1 |
NM_001006945.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015254.hg.1 |
NM_001006945.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015254.hg.1 |
NM_001006945.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015254.hg.1 |
NM_018372.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015254.hg.1 |
NM_018372.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015254.hg.1 |
NM_018372.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015254.hg.1 |
NM_018372.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015254.hg.1 |
NM_018372.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015254.hg.1 |
NM_018372.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015254.hg.1 |
NM_018372.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015254.hg.1 |
NM_018372.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015254.hg.1 |
NM_018372.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100213709.hg.1 |
ENST00000369763.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100213709.hg.1 |
ENST00000485275.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100213709.hg.1 |
ENST00000494675.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100213709.hg.1 |
NM_001006945.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100213709.hg.1 |
NM_018372.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100213709.hg.1 |
ENST00000369763.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100213709.hg.1 |
ENST00000485275.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100213709.hg.1 |
ENST00000494675.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100213709.hg.1 |
NM_001006945.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100213709.hg.1 |
NM_018372.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100213709.hg.1 |
TC0100015254.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100213708.hg.1 |
ENST00000369763.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100213708.hg.1 |
ENST00000485275.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100213708.hg.1 |
ENST00000494675.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100213708.hg.1 |
NM_001006945.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100213708.hg.1 |
NM_018372.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100213708.hg.1 |
ENST00000369763.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100213708.hg.1 |
ENST00000485275.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100213708.hg.1 |
ENST00000494675.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100213708.hg.1 |
NM_001006945.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100213708.hg.1 |
NM_018372.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100213708.hg.1 |
TC0100015254.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100213707.hg.1 |
ENST00000369763.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100213707.hg.1 |
ENST00000494675.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100213707.hg.1 |
NM_001006945.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100213707.hg.1 |
NM_018372.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100213707.hg.1 |
ENST00000369763.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100213707.hg.1 |
ENST00000494675.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100213707.hg.1 |
NM_001006945.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100213707.hg.1 |
NM_018372.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100213707.hg.1 |
TC0100015254.hg.1 |
Clariom D TC |