| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015754.hg.1 |
ENST00000271651.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015754.hg.1 |
ENST00000271651.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015754.hg.1 |
ENST00000271651.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015754.hg.1 |
ENST00000271651.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015754.hg.1 |
ENST00000271651.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015754.hg.1 |
ENST00000271651.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015754.hg.1 |
ENST00000271651.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015754.hg.1 |
ENST00000271651.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015754.hg.1 |
ENST00000271651.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015754.hg.1 |
ENST00000271651.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015754.hg.1 |
ENST00000443913.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015754.hg.1 |
ENST00000443913.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015754.hg.1 |
ENST00000443913.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015754.hg.1 |
ENST00000443913.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015754.hg.1 |
ENST00000443913.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015754.hg.1 |
ENST00000443913.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015754.hg.1 |
ENST00000480670.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015754.hg.1 |
ENST00000480670.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015754.hg.1 |
ENST00000480670.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015754.hg.1 |
NM_000396.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015754.hg.1 |
NM_000396.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015754.hg.1 |
NM_000396.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015754.hg.1 |
NM_000396.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015754.hg.1 |
NM_000396.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015754.hg.1 |
NM_000396.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015754.hg.1 |
NM_000396.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015754.hg.1 |
NM_000396.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015754.hg.1 |
NM_000396.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015754.hg.1 |
NM_000396.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015754.hg.1 |
NM_000396.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01049284.hg.1 |
ENST00000271651.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01049284.hg.1 |
ENST00000480670.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01049284.hg.1 |
NM_000396.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01049284.hg.1 |
TC01003211.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0100015754.hg.1 |
ENST00000271651.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015754.hg.1 |
ENST00000271651.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015754.hg.1 |
ENST00000271651.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015754.hg.1 |
ENST00000271651.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015754.hg.1 |
ENST00000271651.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015754.hg.1 |
ENST00000271651.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015754.hg.1 |
ENST00000271651.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015754.hg.1 |
ENST00000271651.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015754.hg.1 |
ENST00000271651.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015754.hg.1 |
ENST00000271651.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015754.hg.1 |
ENST00000443913.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015754.hg.1 |
ENST00000443913.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015754.hg.1 |
ENST00000443913.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015754.hg.1 |
ENST00000443913.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015754.hg.1 |
ENST00000443913.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015754.hg.1 |
ENST00000443913.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015754.hg.1 |
ENST00000480670.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015754.hg.1 |
ENST00000480670.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015754.hg.1 |
ENST00000480670.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015754.hg.1 |
NM_000396.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015754.hg.1 |
NM_000396.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015754.hg.1 |
NM_000396.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015754.hg.1 |
NM_000396.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015754.hg.1 |
NM_000396.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015754.hg.1 |
NM_000396.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015754.hg.1 |
NM_000396.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015754.hg.1 |
NM_000396.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015754.hg.1 |
NM_000396.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015754.hg.1 |
NM_000396.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015754.hg.1 |
NM_000396.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100218031.hg.1 |
ENST00000271651.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100218031.hg.1 |
ENST00000443913.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100218031.hg.1 |
ENST00000480670.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100218031.hg.1 |
NM_000396.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100218031.hg.1 |
ENST00000271651.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100218031.hg.1 |
ENST00000443913.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100218031.hg.1 |
ENST00000480670.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100218031.hg.1 |
NM_000396.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100218031.hg.1 |
TC0100015754.hg.1 |
Clariom D TC |