Search
Search
| Gene Symbol: UBR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015255.2 | NP_056070.1 | 20 - 21 | 2583 | 130 | |
| XM_005248965.4 | XP_005249022.1 | 20 - 21 | 2700 | 130 | ||
| XM_005248966.3 | XP_005249023.1 | 19 - 20 | 1983 | 130 | ||
| XM_011514438.2 | XP_011512740.2 | 20 - 21 | 2783 | 130 | ||
| XM_011514439.1 | XP_011512741.1 | 11 - 12 | 1150 | 130 | ||
| XM_011514440.1 | XP_011512742.1 | 11 - 12 | 1150 | 130 | ||
| XM_011514441.2 | XP_011512743.1 | 8 - 9 | 763 | 130 | ||
| XM_017010594.1 | XP_016866083.1 | 20 - 21 | 2784 | 130 | ||
| XM_017010595.1 | XP_016866084.1 | 20 - 21 | 2784 | 130 | ||
| XM_017010596.1 | XP_016866085.1 | 20 - 21 | 2618 | 130 | ||
| XM_017010597.1 | XP_016866086.1 | 20 - 21 | 2785 | 130 | ||
| GenBank mRNA | AB002347.1 | - | 8 - 9 | 845 | 130 | |
| AK125795.1 | - | 12 - 13 | 1249 | 130 | ||
| AK304070.1 | - | 12 - 13 | 1236 | 130 | ||
| AY061884.1 | - | 20 - 21 | 2285 | 130 | ||
Set Membership: |
|