| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006896.hg.1 |
ENST00000299084.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006896.hg.1 |
ENST00000299084.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006896.hg.1 |
ENST00000299084.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006896.hg.1 |
ENST00000299084.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006896.hg.1 |
ENST00000299084.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006896.hg.1 |
ENST00000299084.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006896.hg.1 |
ENST00000299084.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006896.hg.1 |
ENST00000299084.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006896.hg.1 |
ENST00000299084.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006896.hg.1 |
ENST00000299084.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006896.hg.1 |
ENST00000299084.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006896.hg.1 |
ENST00000299084.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006896.hg.1 |
ENST00000561205.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006896.hg.1 |
ENST00000561205.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006896.hg.1 |
ENST00000561205.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006896.hg.1 |
ENST00000561205.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006896.hg.1 |
ENST00000561205.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006896.hg.1 |
ENST00000561205.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006896.hg.1 |
ENST00000561317.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006896.hg.1 |
ENST00000561317.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006896.hg.1 |
ENST00000561317.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006896.hg.1 |
ENST00000561317.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006896.hg.1 |
ENST00000561317.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006896.hg.1 |
ENST00000561317.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006896.hg.1 |
NM_152594.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006896.hg.1 |
NM_152594.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006896.hg.1 |
NM_152594.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006896.hg.1 |
NM_152594.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006896.hg.1 |
NM_152594.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006896.hg.1 |
NM_152594.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006896.hg.1 |
NM_152594.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006896.hg.1 |
NM_152594.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006896.hg.1 |
NM_152594.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006896.hg.1 |
NM_152594.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006896.hg.1 |
NM_152594.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15001624.hg.1 |
ENST00000299084.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15001624.hg.1 |
NM_152594.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15001624.hg.1 |
TC15000259.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC1500006896.hg.1 |
ENST00000299084.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500006896.hg.1 |
ENST00000299084.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500006896.hg.1 |
ENST00000299084.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500006896.hg.1 |
ENST00000299084.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500006896.hg.1 |
ENST00000299084.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500006896.hg.1 |
ENST00000299084.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500006896.hg.1 |
ENST00000299084.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500006896.hg.1 |
ENST00000299084.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500006896.hg.1 |
ENST00000299084.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500006896.hg.1 |
ENST00000299084.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500006896.hg.1 |
ENST00000299084.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500006896.hg.1 |
ENST00000299084.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500006896.hg.1 |
ENST00000561205.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500006896.hg.1 |
ENST00000561205.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500006896.hg.1 |
ENST00000561205.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500006896.hg.1 |
ENST00000561205.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500006896.hg.1 |
ENST00000561205.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500006896.hg.1 |
ENST00000561205.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500006896.hg.1 |
ENST00000561317.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500006896.hg.1 |
ENST00000561317.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500006896.hg.1 |
ENST00000561317.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500006896.hg.1 |
ENST00000561317.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500006896.hg.1 |
ENST00000561317.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500006896.hg.1 |
ENST00000561317.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500006896.hg.1 |
NM_152594.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500006896.hg.1 |
NM_152594.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500006896.hg.1 |
NM_152594.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500006896.hg.1 |
NM_152594.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500006896.hg.1 |
NM_152594.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500006896.hg.1 |
NM_152594.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500006896.hg.1 |
NM_152594.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500006896.hg.1 |
NM_152594.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500006896.hg.1 |
NM_152594.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500006896.hg.1 |
NM_152594.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500006896.hg.1 |
NM_152594.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500147794.hg.1 |
ENST00000299084.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500147794.hg.1 |
NM_152594.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500147794.hg.1 |
ENST00000299084.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500147794.hg.1 |
NM_152594.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500147794.hg.1 |
TC1500006896.hg.1 |
Clariom D TC |