Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01084628_m1
          See other LBP GE Assays ›
          Gene Symbol:
          LBP
          Gene Name:
          lipopolysaccharide binding protein
          Gene Aliases:
          BPIFD2
          Chromosome Location:
          Chr.20: 38346411 - 38377011 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01084628_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LBP
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_004139.4 NP_004130.2 8 - 9 1030 94
          GenBank mRNA AF105067.1 - 8 - 9 941 94
          AK313625.1 - 8 - 9 960 94
          BC022256.1 - 8 - 9 959 4767724 94
          M35533.1 - 8 - 9 912 94

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC2000007313.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC20000282.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene LBP

          Gene Ontology Categories:

          Function(s) Process(es)

          liver development
          leukocyte chemotaxis involved in inflammatory response
          macrophage activation involved in immune response
          acute-phase response
          cellular defense response
          opsonization
          lipopolysaccharide transport
          lipopolysaccharide-mediated signaling pathway
          detection of molecule of bacterial origin
          response to lipopolysaccharide
          negative regulation of tumor necrosis factor production
          positive regulation of chemokine production
          positive regulation of interleukin-6 production
          positive regulation of interleukin-8 production
          positive regulation of tumor necrosis factor production
          toll-like receptor 4 signaling pathway
          positive regulation of toll-like receptor 4 signaling pathway
          positive regulation of tumor necrosis factor biosynthetic process
          positive regulation of macrophage activation
          negative regulation of growth of symbiont in host
          innate immune response
          defense response to Gram-negative bacterium
          defense response to Gram-positive bacterium
          positive regulation of phagocytosis, engulfment
          positive regulation of respiratory burst involved in inflammatory response
          cellular response to lipopolysaccharide
          cellular response to lipoteichoic acid
          positive regulation of neutrophil chemotaxis
          regulation of membrane permeability
          lipopolysaccharide binding
          receptor binding
          protein binding
          lipoteichoic acid binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR20004019.hg.1 ENST00000217407.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20004019.hg.1 ENST00000538599.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20004019.hg.1 NM_004139.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR20004019.hg.1 TC20000282.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC2000007313.hg.1 ENST00000217407.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007313.hg.1 ENST00000217407.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007313.hg.1 ENST00000217407.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007313.hg.1 ENST00000217407.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007313.hg.1 ENST00000217407.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007313.hg.1 ENST00000217407.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007313.hg.1 ENST00000217407.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007313.hg.1 ENST00000217407.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007313.hg.1 ENST00000217407.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007313.hg.1 ENST00000217407.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007313.hg.1 ENST00000217407.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007313.hg.1 ENST00000217407.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007313.hg.1 ENST00000217407.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007313.hg.1 ENST00000217407.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007313.hg.1 ENST00000217407.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007313.hg.1 ENST00000217407.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007313.hg.1 ENST00000217407.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007313.hg.1 ENST00000217407.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007313.hg.1 ENST00000217407.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007313.hg.1 ENST00000217407.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000007313.hg.1 NM_004139.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007313.hg.1 NM_004139.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007313.hg.1 NM_004139.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007313.hg.1 NM_004139.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007313.hg.1 NM_004139.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007313.hg.1 NM_004139.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007313.hg.1 NM_004139.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007313.hg.1 NM_004139.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007313.hg.1 NM_004139.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007313.hg.1 NM_004139.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007313.hg.1 NM_004139.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007313.hg.1 NM_004139.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007313.hg.1 NM_004139.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007313.hg.1 NM_004139.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007313.hg.1 NM_004139.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007313.hg.1 NM_004139.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007313.hg.1 NM_004139.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007313.hg.1 NM_004139.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000007313.hg.1 NM_004139.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR20004020.hg.1 ENST00000217407.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20004020.hg.1 ENST00000538599.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20004020.hg.1 NM_004139.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR20004020.hg.1 TC20000282.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC2000007313.hg.1 ENST00000217407.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000007313.hg.1 ENST00000217407.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000007313.hg.1 ENST00000217407.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000007313.hg.1 ENST00000217407.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000007313.hg.1 ENST00000217407.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000007313.hg.1 ENST00000217407.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000007313.hg.1 ENST00000217407.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000007313.hg.1 ENST00000217407.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000007313.hg.1 ENST00000217407.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000007313.hg.1 ENST00000217407.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000007313.hg.1 ENST00000217407.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000007313.hg.1 ENST00000217407.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000007313.hg.1 ENST00000217407.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000007313.hg.1 ENST00000217407.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000007313.hg.1 ENST00000217407.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000007313.hg.1 ENST00000217407.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000007313.hg.1 ENST00000217407.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000007313.hg.1 ENST00000217407.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000007313.hg.1 ENST00000217407.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000007313.hg.1 ENST00000217407.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000007313.hg.1 NM_004139.4 Refseq
          Affymetrix Human Clariom D Assay TC2000007313.hg.1 NM_004139.4 Refseq
          Affymetrix Human Clariom D Assay TC2000007313.hg.1 NM_004139.4 Refseq
          Affymetrix Human Clariom D Assay TC2000007313.hg.1 NM_004139.4 Refseq
          Affymetrix Human Clariom D Assay TC2000007313.hg.1 NM_004139.4 Refseq
          Affymetrix Human Clariom D Assay TC2000007313.hg.1 NM_004139.4 Refseq
          Affymetrix Human Clariom D Assay TC2000007313.hg.1 NM_004139.4 Refseq
          Affymetrix Human Clariom D Assay TC2000007313.hg.1 NM_004139.4 Refseq
          Affymetrix Human Clariom D Assay TC2000007313.hg.1 NM_004139.4 Refseq
          Affymetrix Human Clariom D Assay TC2000007313.hg.1 NM_004139.4 Refseq
          Affymetrix Human Clariom D Assay TC2000007313.hg.1 NM_004139.4 Refseq
          Affymetrix Human Clariom D Assay TC2000007313.hg.1 NM_004139.4 Refseq
          Affymetrix Human Clariom D Assay TC2000007313.hg.1 NM_004139.4 Refseq
          Affymetrix Human Clariom D Assay TC2000007313.hg.1 NM_004139.4 Refseq
          Affymetrix Human Clariom D Assay TC2000007313.hg.1 NM_004139.4 Refseq
          Affymetrix Human Clariom D Assay TC2000007313.hg.1 NM_004139.4 Refseq
          Affymetrix Human Clariom D Assay TC2000007313.hg.1 NM_004139.4 Refseq
          Affymetrix Human Clariom D Assay TC2000007313.hg.1 NM_004139.4 Refseq
          Affymetrix Human Clariom D Assay TC2000007313.hg.1 NM_004139.4 Refseq
          Affymetrix Human Clariom D Assay PSR2000150581.hg.1 ENST00000217407.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2000150581.hg.1 NM_004139.4 Refseq
          Affymetrix Human Clariom D Assay PSR2000150581.hg.1 TC2000007313.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2000150581.hg.1 ENST00000217407.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2000150581.hg.1 NM_004139.4 Refseq
          Affymetrix Human Clariom D Assay PSR2000150582.hg.1 ENST00000217407.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2000150582.hg.1 NM_004139.4 Refseq
          Affymetrix Human Clariom D Assay PSR2000150582.hg.1 TC2000007313.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2000150582.hg.1 ENST00000217407.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2000150582.hg.1 NM_004139.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline