Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01084651_m1
          See other AGO1 GE Assays ›
          Gene Symbol:
          AGO1
          Gene Name:
          argonaute 1, RISC catalytic component
          Gene Aliases:
          EIF2C, EIF2C1, GERP95, Q99, hAgo1
          Chromosome Location:
          Chr.1: 35869808 - 35929610 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01084651_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: AGO1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001317122.1 NP_001304051.1 15 - 16 2252 84
          NM_001317123.1 NP_001304052.1 15 - 16 2096 84
          NM_012199.4 NP_036331.1 15 - 16 2252 84
          XM_011541236.2 XP_011539538.1 15 - 16 2222 84
          XM_017001016.1 XP_016856505.1 15 - 16 2222 84
          XM_017001017.1 XP_016856506.1 15 - 16 2213 84
          XM_017001018.1 XP_016856507.1 12 - 13 1697 84
          XM_017001019.1 XP_016856508.1 15 - 16 2310 84
          GenBank mRNA AK002003.1 - 1 - 2 74 84
          AK314148.1 - 15 - 16 2213 84
          BC063275.1 - 15 - 16 2232 30344513 84
          CR936823.1 - 5 - 6 715 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100007786.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01000463.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene AGO1

          Panther Classification:

          Molecular Function -

          translation initiation factor

          Gene Ontology Categories:

          Function(s) Process(es)

          nuclear-transcribed mRNA catabolic process
          transcription, DNA-templated
          Wnt signaling pathway, calcium modulating pathway
          RNA secondary structure unwinding
          miRNA metabolic process
          gene silencing by RNA
          pre-miRNA processing
          posttranscriptional gene silencing by RNA
          production of miRNAs involved in gene silencing by miRNA
          miRNA mediated inhibition of translation
          mRNA cleavage involved in gene silencing by miRNA
          miRNA loading onto RISC involved in gene silencing by miRNA
          positive regulation of transcription from RNA polymerase II promoter
          phosphatidylinositol-mediated signaling
          mRNA cleavage involved in gene silencing by siRNA
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II core binding
          core promoter binding
          RNA binding
          double-stranded RNA binding
          single-stranded RNA binding
          endoribonuclease activity
          protein binding
          miRNA binding
          poly(A) RNA binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0100007786.hg.1 ENST00000373204.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007786.hg.1 ENST00000373204.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007786.hg.1 ENST00000373204.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007786.hg.1 ENST00000373204.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007786.hg.1 ENST00000373204.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007786.hg.1 ENST00000373204.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007786.hg.1 ENST00000373204.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007786.hg.1 ENST00000373204.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007786.hg.1 ENST00000373204.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007786.hg.1 ENST00000373204.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007786.hg.1 ENST00000373204.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007786.hg.1 ENST00000373204.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007786.hg.1 ENST00000373204.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007786.hg.1 ENST00000373204.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007786.hg.1 ENST00000373204.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007786.hg.1 ENST00000373204.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007786.hg.1 ENST00000373204.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007786.hg.1 ENST00000373204.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007786.hg.1 ENST00000373204.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007786.hg.1 ENST00000373204.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007786.hg.1 ENST00000373204.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007786.hg.1 ENST00000373204.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007786.hg.1 ENST00000373204.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007786.hg.1 ENST00000373204.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007786.hg.1 ENST00000373206.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007786.hg.1 ENST00000373206.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007786.hg.1 ENST00000373206.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007786.hg.1 ENST00000373206.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007786.hg.1 ENST00000373206.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007786.hg.1 ENST00000373206.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007786.hg.1 ENST00000373206.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007786.hg.1 ENST00000373206.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007786.hg.1 ENST00000373206.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007786.hg.1 ENST00000373206.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007786.hg.1 ENST00000373206.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007786.hg.1 ENST00000373206.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007786.hg.1 ENST00000373206.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007786.hg.1 ENST00000373206.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007786.hg.1 ENST00000373206.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007786.hg.1 ENST00000373206.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007786.hg.1 ENST00000373206.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007786.hg.1 ENST00000373206.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007786.hg.1 ENST00000373206.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007786.hg.1 ENST00000373206.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007786.hg.1 ENST00000624357.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007786.hg.1 NM_012199.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007786.hg.1 NM_012199.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007786.hg.1 NM_012199.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007786.hg.1 NM_012199.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007786.hg.1 NM_012199.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007786.hg.1 NM_012199.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007786.hg.1 NM_012199.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007786.hg.1 NM_012199.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007786.hg.1 NM_012199.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007786.hg.1 NM_012199.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007786.hg.1 NM_012199.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007786.hg.1 NM_012199.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007786.hg.1 NM_012199.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007786.hg.1 NM_012199.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007786.hg.1 NM_012199.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007786.hg.1 NM_012199.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007786.hg.1 NM_012199.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007786.hg.1 NM_012199.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007786.hg.1 NM_012199.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007786.hg.1 NM_012199.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007786.hg.1 NM_012199.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007786.hg.1 NM_012199.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01007146.hg.1 ENST00000373204.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01007146.hg.1 ENST00000373206.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01007146.hg.1 NM_012199.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01007146.hg.1 TC01000463.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC0100007786.hg.1 ENST00000373204.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100007786.hg.1 ENST00000373204.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100007786.hg.1 ENST00000373204.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100007786.hg.1 ENST00000373204.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100007786.hg.1 ENST00000373204.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100007786.hg.1 ENST00000373204.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100007786.hg.1 ENST00000373204.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100007786.hg.1 ENST00000373204.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100007786.hg.1 ENST00000373204.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100007786.hg.1 ENST00000373204.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100007786.hg.1 ENST00000373204.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100007786.hg.1 ENST00000373204.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100007786.hg.1 ENST00000373204.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100007786.hg.1 ENST00000373204.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100007786.hg.1 ENST00000373204.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100007786.hg.1 ENST00000373204.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100007786.hg.1 ENST00000373204.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100007786.hg.1 ENST00000373204.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100007786.hg.1 ENST00000373204.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100007786.hg.1 ENST00000373204.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100007786.hg.1 ENST00000373204.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100007786.hg.1 ENST00000373204.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100007786.hg.1 ENST00000373204.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100007786.hg.1 ENST00000373204.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100007786.hg.1 ENST00000373206.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100007786.hg.1 ENST00000373206.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100007786.hg.1 ENST00000373206.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100007786.hg.1 ENST00000373206.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100007786.hg.1 ENST00000373206.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100007786.hg.1 ENST00000373206.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100007786.hg.1 ENST00000373206.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100007786.hg.1 ENST00000373206.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100007786.hg.1 ENST00000373206.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100007786.hg.1 ENST00000373206.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100007786.hg.1 ENST00000373206.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100007786.hg.1 ENST00000373206.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100007786.hg.1 ENST00000373206.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100007786.hg.1 ENST00000373206.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100007786.hg.1 ENST00000373206.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100007786.hg.1 ENST00000373206.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100007786.hg.1 ENST00000373206.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100007786.hg.1 ENST00000373206.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100007786.hg.1 ENST00000373206.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100007786.hg.1 ENST00000373206.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100007786.hg.1 ENST00000624357.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100007786.hg.1 NM_012199.4 Refseq
          Affymetrix Human Clariom D Assay TC0100007786.hg.1 NM_012199.4 Refseq
          Affymetrix Human Clariom D Assay TC0100007786.hg.1 NM_012199.4 Refseq
          Affymetrix Human Clariom D Assay TC0100007786.hg.1 NM_012199.4 Refseq
          Affymetrix Human Clariom D Assay TC0100007786.hg.1 NM_012199.4 Refseq
          Affymetrix Human Clariom D Assay TC0100007786.hg.1 NM_012199.4 Refseq
          Affymetrix Human Clariom D Assay TC0100007786.hg.1 NM_012199.4 Refseq
          Affymetrix Human Clariom D Assay TC0100007786.hg.1 NM_012199.4 Refseq
          Affymetrix Human Clariom D Assay TC0100007786.hg.1 NM_012199.4 Refseq
          Affymetrix Human Clariom D Assay TC0100007786.hg.1 NM_012199.4 Refseq
          Affymetrix Human Clariom D Assay TC0100007786.hg.1 NM_012199.4 Refseq
          Affymetrix Human Clariom D Assay TC0100007786.hg.1 NM_012199.4 Refseq
          Affymetrix Human Clariom D Assay TC0100007786.hg.1 NM_012199.4 Refseq
          Affymetrix Human Clariom D Assay TC0100007786.hg.1 NM_012199.4 Refseq
          Affymetrix Human Clariom D Assay TC0100007786.hg.1 NM_012199.4 Refseq
          Affymetrix Human Clariom D Assay TC0100007786.hg.1 NM_012199.4 Refseq
          Affymetrix Human Clariom D Assay TC0100007786.hg.1 NM_012199.4 Refseq
          Affymetrix Human Clariom D Assay TC0100007786.hg.1 NM_012199.4 Refseq
          Affymetrix Human Clariom D Assay TC0100007786.hg.1 NM_012199.4 Refseq
          Affymetrix Human Clariom D Assay TC0100007786.hg.1 NM_012199.4 Refseq
          Affymetrix Human Clariom D Assay TC0100007786.hg.1 NM_012199.4 Refseq
          Affymetrix Human Clariom D Assay TC0100007786.hg.1 NM_012199.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100154822.hg.1 ENST00000373204.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100154822.hg.1 ENST00000373206.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100154822.hg.1 NM_012199.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100154822.hg.1 ENST00000373204.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100154822.hg.1 ENST00000373206.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100154822.hg.1 NM_012199.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100154822.hg.1 TC0100007786.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline