Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01088414_g1
          See other CA4 GE Assays ›
          Gene Symbol:
          CA4
          Gene Name:
          carbonic anhydrase 4
          Gene Aliases:
          CAIV, Car4, RP17
          Chromosome Location:
          Chr.17: 60149941 - 60178118 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01088414_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CA4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_005257639.2 XP_005257696.1 4 - 5 508 71
          XM_011525183.2 XP_011523485.1 4 - 5 497 71
          XM_017025012.1 XP_016880501.1 4 - 5 497 71
          GenBank mRNA AK289715.1 - 4 - 5 507 71
          BC057792.1 - 4 - 5 513 30331755 71
          BC069649.1 - 4 - 5 460 7262441 71
          BC074768.2 - 4 - 5 437 30915189 71
          CR541766.1 - 4 - 5 414 71
          DA113846.1 - 4 - 5 508 71
          M83670.1 - 4 - 5 461 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1700012289.hg.1

          Set Membership:

          Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          one-carbon metabolic process
          bicarbonate transport
          carbonate dehydratase activity
          protein binding
          zinc ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1700012289.hg.1 ENST00000300900.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012289.hg.1 ENST00000300900.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012289.hg.1 ENST00000300900.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012289.hg.1 ENST00000300900.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012289.hg.1 ENST00000300900.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012289.hg.1 ENST00000300900.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012289.hg.1 ENST00000300900.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012289.hg.1 ENST00000300900.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012289.hg.1 ENST00000300900.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012289.hg.1 ENST00000300900.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012289.hg.1 ENST00000300900.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012289.hg.1 ENST00000300900.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012289.hg.1 ENST00000300900.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012289.hg.1 ENST00000585705.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012289.hg.1 ENST00000585705.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012289.hg.1 ENST00000585705.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012289.hg.1 ENST00000585705.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012289.hg.1 ENST00000585705.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012289.hg.1 ENST00000587265.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012289.hg.1 ENST00000587265.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012289.hg.1 ENST00000590203.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012289.hg.1 ENST00000590203.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012289.hg.1 ENST00000590203.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012289.hg.1 ENST00000590203.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012289.hg.1 ENST00000590203.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012289.hg.1 ENST00000590203.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012289.hg.1 ENST00000590203.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012289.hg.1 ENST00000591725.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012289.hg.1 ENST00000591725.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012289.hg.1 ENST00000591725.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012289.hg.1 ENST00000591725.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012289.hg.1 ENST00000591725.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012289.hg.1 ENST00000591725.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700012289.hg.1 NM_000717.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700012289.hg.1 NM_000717.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700012289.hg.1 NM_000717.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700012289.hg.1 NM_000717.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700012289.hg.1 NM_000717.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700012289.hg.1 NM_000717.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700012289.hg.1 NM_000717.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700012289.hg.1 NM_000717.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700012289.hg.1 NM_000717.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700012289.hg.1 NM_000717.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700012289.hg.1 NM_000717.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700012289.hg.1 NM_000717.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700012289.hg.1 NM_000717.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline