Search
Search

SMARCD1| Gene Symbol: SMARCD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005269107.3 | XP_005269164.2 | 4 - 5 | 694 | 130 | |
| GenBank mRNA | AK292700.1 | - | 5 - 6 | 480 | 131 | |
Set Membership: |
|