Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01092197_g1
          See other DTX1 GE Assays ›
          Gene Symbol:
          DTX1
          Gene Name:
          deltex E3 ubiquitin ligase 1
          Gene Aliases:
          hDx-1
          Chromosome Location:
          Chr.12: 113056711 - 113098028 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01092197_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: DTX1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_004416.2 NP_004407.2 4 - 5 1504 72
          XM_011538009.2 XP_011536311.1 5 - 6 2146 72
          GenBank mRNA AF053700.1 - 4 - 5 1671 72
          BC048216.1 - 4 - 5 1446 5931062 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1200008924.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC12000887.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene DTX1

          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          transcription from RNA polymerase II promoter
          cell surface receptor signaling pathway
          Notch signaling pathway
          regulation of Notch signaling pathway
          glial cell differentiation
          protein ubiquitination
          negative regulation of T cell differentiation
          negative regulation of neuron differentiation
          regulation of nucleic acid-templated transcription
          transcription coactivator activity
          Notch binding
          protein binding
          zinc ion binding
          ligase activity
          SH3 domain binding
          ubiquitin protein ligase binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1200008924.hg.1 ENST00000257600.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008924.hg.1 ENST00000257600.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008924.hg.1 ENST00000257600.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008924.hg.1 ENST00000257600.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008924.hg.1 ENST00000257600.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008924.hg.1 ENST00000257600.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008924.hg.1 ENST00000257600.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008924.hg.1 ENST00000257600.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008924.hg.1 ENST00000257600.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008924.hg.1 ENST00000257600.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008924.hg.1 ENST00000257600.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008924.hg.1 ENST00000257600.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008924.hg.1 ENST00000257600.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008924.hg.1 ENST00000257600.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008924.hg.1 ENST00000257600.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008924.hg.1 ENST00000547730.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008924.hg.1 ENST00000547730.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008924.hg.1 ENST00000547730.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008924.hg.1 ENST00000547974.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008924.hg.1 ENST00000547974.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008924.hg.1 ENST00000547974.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008924.hg.1 ENST00000547974.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008924.hg.1 ENST00000547974.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008924.hg.1 ENST00000547974.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008924.hg.1 ENST00000547974.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008924.hg.1 ENST00000547974.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008924.hg.1 ENST00000547974.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008924.hg.1 ENST00000548759.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008924.hg.1 ENST00000548759.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008924.hg.1 ENST00000548759.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008924.hg.1 ENST00000548759.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008924.hg.1 ENST00000548759.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008924.hg.1 ENST00000553140.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008924.hg.1 ENST00000553140.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008924.hg.1 ENST00000553140.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008924.hg.1 ENST00000553140.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200008924.hg.1 NM_004416.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200008924.hg.1 NM_004416.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200008924.hg.1 NM_004416.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200008924.hg.1 NM_004416.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200008924.hg.1 NM_004416.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200008924.hg.1 NM_004416.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200008924.hg.1 NM_004416.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200008924.hg.1 NM_004416.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200008924.hg.1 NM_004416.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200008924.hg.1 NM_004416.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200008924.hg.1 NM_004416.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200008924.hg.1 NM_004416.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200008924.hg.1 NM_004416.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200008924.hg.1 NM_004416.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12011730.hg.1 ENST00000257600.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12011730.hg.1 ENST00000547974.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12011730.hg.1 NM_004416.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12011730.hg.1 TC12000887.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR12011731.hg.1 ENST00000257600.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12011731.hg.1 ENST00000547974.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12011731.hg.1 NM_004416.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12011731.hg.1 TC12000887.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1200166360.hg.1 ENST00000257600.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1200166360.hg.1 ENST00000547974.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200166360.hg.1 ENST00000553140.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200166360.hg.1 NM_004416.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200166360.hg.1 ENST00000257600.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1200166360.hg.1 ENST00000547974.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200166360.hg.1 ENST00000553140.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200166360.hg.1 NM_004416.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200166360.hg.1 TC1200008924.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200166361.hg.1 ENST00000257600.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1200166361.hg.1 ENST00000547974.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200166361.hg.1 ENST00000553140.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200166361.hg.1 NM_004416.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200166361.hg.1 ENST00000257600.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1200166361.hg.1 ENST00000547974.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200166361.hg.1 ENST00000553140.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200166361.hg.1 NM_004416.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200166361.hg.1 TC1200008924.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1200008924.hg.1 ENST00000257600.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200008924.hg.1 ENST00000257600.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200008924.hg.1 ENST00000257600.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200008924.hg.1 ENST00000257600.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200008924.hg.1 ENST00000257600.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200008924.hg.1 ENST00000257600.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200008924.hg.1 ENST00000257600.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200008924.hg.1 ENST00000257600.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200008924.hg.1 ENST00000257600.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200008924.hg.1 ENST00000257600.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200008924.hg.1 ENST00000257600.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200008924.hg.1 ENST00000257600.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200008924.hg.1 ENST00000257600.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200008924.hg.1 ENST00000257600.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200008924.hg.1 ENST00000257600.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200008924.hg.1 ENST00000547730.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200008924.hg.1 ENST00000547730.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200008924.hg.1 ENST00000547730.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200008924.hg.1 ENST00000547974.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200008924.hg.1 ENST00000547974.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200008924.hg.1 ENST00000547974.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200008924.hg.1 ENST00000547974.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200008924.hg.1 ENST00000547974.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200008924.hg.1 ENST00000547974.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200008924.hg.1 ENST00000547974.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200008924.hg.1 ENST00000547974.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200008924.hg.1 ENST00000547974.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200008924.hg.1 ENST00000548759.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200008924.hg.1 ENST00000548759.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200008924.hg.1 ENST00000548759.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200008924.hg.1 ENST00000548759.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200008924.hg.1 ENST00000548759.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200008924.hg.1 ENST00000553140.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200008924.hg.1 ENST00000553140.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200008924.hg.1 ENST00000553140.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200008924.hg.1 ENST00000553140.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200008924.hg.1 NM_004416.2 Refseq
          Affymetrix Human Clariom D Assay TC1200008924.hg.1 NM_004416.2 Refseq
          Affymetrix Human Clariom D Assay TC1200008924.hg.1 NM_004416.2 Refseq
          Affymetrix Human Clariom D Assay TC1200008924.hg.1 NM_004416.2 Refseq
          Affymetrix Human Clariom D Assay TC1200008924.hg.1 NM_004416.2 Refseq
          Affymetrix Human Clariom D Assay TC1200008924.hg.1 NM_004416.2 Refseq
          Affymetrix Human Clariom D Assay TC1200008924.hg.1 NM_004416.2 Refseq
          Affymetrix Human Clariom D Assay TC1200008924.hg.1 NM_004416.2 Refseq
          Affymetrix Human Clariom D Assay TC1200008924.hg.1 NM_004416.2 Refseq
          Affymetrix Human Clariom D Assay TC1200008924.hg.1 NM_004416.2 Refseq
          Affymetrix Human Clariom D Assay TC1200008924.hg.1 NM_004416.2 Refseq
          Affymetrix Human Clariom D Assay TC1200008924.hg.1 NM_004416.2 Refseq
          Affymetrix Human Clariom D Assay TC1200008924.hg.1 NM_004416.2 Refseq
          Affymetrix Human Clariom D Assay TC1200008924.hg.1 NM_004416.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline