Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01092434_m1
          See other DHCR24 GE Assays ›
          Gene Symbol:
          DHCR24
          Gene Name:
          24-dehydrocholesterol reductase
          Gene Aliases:
          DCE, Nbla03646, SELADIN1, seladin-1
          Chromosome Location:
          Chr.1: 54849627 - 54887248 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01092434_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: DHCR24
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_014762.3 NP_055577.1 2 - 3 516 112
          GenBank mRNA AB073391.1 - 3 - 4 391 112
          AF261758.1 - 2 - 3 486 112
          AK302774.1 - 2 - 3 347 112
          AK304437.1 - 2 - 3 233 112
          AK316419.1 - 2 - 3 278 112
          AK316427.1 - 2 - 3 217 112
          BC004375.1 - 2 - 3 422 3638639 112
          BC011669.2 - 2 - 3 449 4130396 112
          BP200916.1 - 2 - 3 516 112
          D13643.3 - 2 - 3 499 112

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100014276.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01002694.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene DHCR24

          Panther Classification:

          Molecular Function -

          reductase

          Gene Ontology Categories:

          Function(s) Process(es)

          cholesterol biosynthetic process
          apoptotic process
          response to oxidative stress
          cell cycle arrest
          Ras protein signal transduction
          protein localization
          negative regulation of cell proliferation
          response to hormone
          tissue development
          male genitalia development
          plasminogen activation
          cholesterol biosynthetic process via desmosterol
          cholesterol biosynthetic process via lathosterol
          amyloid precursor protein catabolic process
          negative regulation of apoptotic process
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          skin development
          oxidation-reduction process
          membrane organization
          regulation of neuron death
          delta24(24-1) sterol reductase activity
          oxidoreductase activity, acting on CH-OH group of donors
          oxidoreductase activity, acting on the CH-CH group of donors, NAD or NADP as acceptor
          enzyme binding
          peptide antigen binding
          delta24-sterol reductase activity
          flavin adenine dinucleotide binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000436604.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000436604.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000436604.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000436604.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000436604.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01042257.hg.1 ENST00000371269.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01042257.hg.1 ENST00000535035.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01042257.hg.1 NM_014762.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01042257.hg.1 TC01002694.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01042256.hg.1 ENST00000371269.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01042256.hg.1 ENST00000535035.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01042256.hg.1 ENST00000537443.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01042256.hg.1 NM_014762.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01042256.hg.1 TC01002694.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000436604.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000436604.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000436604.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000436604.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000436604.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014276.hg.1 NM_014762.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100206994.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100206994.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100206994.hg.1 NM_014762.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100206994.hg.1 ENST00000371269.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100206994.hg.1 ENST00000535035.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100206994.hg.1 NM_014762.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100206994.hg.1 TC0100014276.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline