Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01092917_g1
          See other PDCD5 GE Assays ›
          Gene Symbol:
          PDCD5
          Gene Name:
          programmed cell death 5
          Gene Aliases:
          TFAR19
          Chromosome Location:
          Chr.19: 32581161 - 32587452 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01092917_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PDCD5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_004708.3 NP_004699.1 2 - 3 177 69
          XM_005259392.4 XP_005259449.1 2 - 3 204 69
          GenBank mRNA AF014955.1 - 2 - 3 127 69
          AK293486.1 - 2 - 3 175 69
          BC015519.2 - 2 - 3 143 3888459 69
          BM696336.1 - 2 - 3 167 69
          BT006694.1 - 2 - 3 103 69
          CR456934.1 - 2 - 3 103 69
          DN915905.1 - 2 - 3 177 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1900007748.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC19000427.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene PDCD5

          Panther Classification:

          Molecular Function -

          protein-binding activity modulator

          Gene Ontology Categories:

          Function(s) Process(es)

          apoptotic process
          negative regulation of cell proliferation
          positive regulation of gene expression
          positive regulation of apoptotic process
          positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
          cellular response to transforming growth factor beta stimulus
          positive regulation of release of cytochrome c from mitochondria
          positive regulation of protein import into mitochondrial outer membrane
          negative regulation of chaperone-mediated protein folding
          DNA binding
          protein binding
          heparin binding
          acetyltransferase activator activity
          beta-tubulin binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR19005561.hg.1 ENST00000221784.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19005561.hg.1 ENST00000419343.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19005561.hg.1 NM_004708.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19005561.hg.1 TC19000427.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR19005562.hg.1 ENST00000221784.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19005562.hg.1 ENST00000419343.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19005562.hg.1 NM_004708.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19005562.hg.1 TC19000427.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1900007748.hg.1 ENST00000221784.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007748.hg.1 ENST00000221784.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007748.hg.1 ENST00000221784.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007748.hg.1 ENST00000221784.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007748.hg.1 ENST00000221784.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007748.hg.1 ENST00000221784.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007748.hg.1 ENST00000221784.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007748.hg.1 ENST00000221784.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007748.hg.1 ENST00000379316.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007748.hg.1 ENST00000379316.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007748.hg.1 ENST00000379316.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007748.hg.1 ENST00000419343.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007748.hg.1 ENST00000419343.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007748.hg.1 ENST00000419343.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007748.hg.1 ENST00000419343.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007748.hg.1 ENST00000419343.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007748.hg.1 ENST00000419343.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007748.hg.1 ENST00000419343.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007748.hg.1 ENST00000419343.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007748.hg.1 ENST00000419343.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007748.hg.1 ENST00000419343.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007748.hg.1 ENST00000419343.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007748.hg.1 ENST00000419343.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007748.hg.1 ENST00000586035.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007748.hg.1 ENST00000586035.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007748.hg.1 ENST00000586035.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007748.hg.1 ENST00000586035.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007748.hg.1 ENST00000586035.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007748.hg.1 ENST00000586035.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007748.hg.1 ENST00000586035.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007748.hg.1 ENST00000586316.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007748.hg.1 ENST00000586316.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007748.hg.1 ENST00000586316.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007748.hg.1 ENST00000586316.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007748.hg.1 ENST00000586316.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007748.hg.1 ENST00000586316.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007748.hg.1 ENST00000588622.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007748.hg.1 ENST00000588622.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007748.hg.1 ENST00000588622.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007748.hg.1 ENST00000588622.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007748.hg.1 ENST00000590247.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007748.hg.1 ENST00000590247.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007748.hg.1 ENST00000590247.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007748.hg.1 ENST00000590247.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007748.hg.1 ENST00000590247.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007748.hg.1 ENST00000590247.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007748.hg.1 ENST00000590247.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007748.hg.1 ENST00000590247.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007748.hg.1 ENST00000590247.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007748.hg.1 ENST00000592786.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007748.hg.1 ENST00000592786.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007748.hg.1 ENST00000592786.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007748.hg.1 ENST00000592786.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007748.hg.1 ENST00000592786.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007748.hg.1 ENST00000592786.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007748.hg.1 ENST00000592786.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007748.hg.1 ENST00000592786.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007748.hg.1 NM_004708.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900007748.hg.1 NM_004708.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900007748.hg.1 NM_004708.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900007748.hg.1 NM_004708.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900007748.hg.1 NM_004708.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900007748.hg.1 NM_004708.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900007748.hg.1 NM_004708.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900007748.hg.1 NM_004708.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900007748.hg.1 NM_004708.3 Refseq
          Affymetrix Human Clariom D Assay PSR1900156907.hg.1 ENST00000221784.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1900156907.hg.1 ENST00000419343.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1900156907.hg.1 ENST00000586035.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900156907.hg.1 ENST00000586316.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900156907.hg.1 ENST00000590247.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1900156907.hg.1 NM_004708.3 Refseq
          Affymetrix Human Clariom D Assay PSR1900156907.hg.1 TC1900007748.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900156907.hg.1 ENST00000221784.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1900156907.hg.1 ENST00000419343.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1900156907.hg.1 ENST00000586035.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900156907.hg.1 ENST00000586316.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900156907.hg.1 ENST00000590247.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1900156907.hg.1 NM_004708.3 Refseq
          Affymetrix Human Clariom D Assay TC1900007748.hg.1 ENST00000221784.8 Ensembl
          Affymetrix Human Clariom D Assay TC1900007748.hg.1 ENST00000221784.8 Ensembl
          Affymetrix Human Clariom D Assay TC1900007748.hg.1 ENST00000221784.8 Ensembl
          Affymetrix Human Clariom D Assay TC1900007748.hg.1 ENST00000221784.8 Ensembl
          Affymetrix Human Clariom D Assay TC1900007748.hg.1 ENST00000221784.8 Ensembl
          Affymetrix Human Clariom D Assay TC1900007748.hg.1 ENST00000221784.8 Ensembl
          Affymetrix Human Clariom D Assay TC1900007748.hg.1 ENST00000221784.8 Ensembl
          Affymetrix Human Clariom D Assay TC1900007748.hg.1 ENST00000221784.8 Ensembl
          Affymetrix Human Clariom D Assay TC1900007748.hg.1 ENST00000379316.3 Ensembl
          Affymetrix Human Clariom D Assay TC1900007748.hg.1 ENST00000379316.3 Ensembl
          Affymetrix Human Clariom D Assay TC1900007748.hg.1 ENST00000379316.3 Ensembl
          Affymetrix Human Clariom D Assay TC1900007748.hg.1 ENST00000419343.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900007748.hg.1 ENST00000419343.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900007748.hg.1 ENST00000419343.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900007748.hg.1 ENST00000419343.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900007748.hg.1 ENST00000419343.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900007748.hg.1 ENST00000419343.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900007748.hg.1 ENST00000419343.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900007748.hg.1 ENST00000419343.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900007748.hg.1 ENST00000419343.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900007748.hg.1 ENST00000419343.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900007748.hg.1 ENST00000419343.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900007748.hg.1 ENST00000419343.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900007748.hg.1 ENST00000586035.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007748.hg.1 ENST00000586035.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007748.hg.1 ENST00000586035.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007748.hg.1 ENST00000586035.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007748.hg.1 ENST00000586035.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007748.hg.1 ENST00000586035.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007748.hg.1 ENST00000586035.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007748.hg.1 ENST00000586316.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900007748.hg.1 ENST00000586316.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900007748.hg.1 ENST00000586316.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900007748.hg.1 ENST00000586316.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900007748.hg.1 ENST00000586316.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900007748.hg.1 ENST00000586316.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900007748.hg.1 ENST00000588622.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007748.hg.1 ENST00000588622.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007748.hg.1 ENST00000588622.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007748.hg.1 ENST00000588622.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007748.hg.1 ENST00000590247.6 Ensembl
          Affymetrix Human Clariom D Assay TC1900007748.hg.1 ENST00000590247.6 Ensembl
          Affymetrix Human Clariom D Assay TC1900007748.hg.1 ENST00000590247.6 Ensembl
          Affymetrix Human Clariom D Assay TC1900007748.hg.1 ENST00000590247.6 Ensembl
          Affymetrix Human Clariom D Assay TC1900007748.hg.1 ENST00000590247.6 Ensembl
          Affymetrix Human Clariom D Assay TC1900007748.hg.1 ENST00000590247.6 Ensembl
          Affymetrix Human Clariom D Assay TC1900007748.hg.1 ENST00000590247.6 Ensembl
          Affymetrix Human Clariom D Assay TC1900007748.hg.1 ENST00000590247.6 Ensembl
          Affymetrix Human Clariom D Assay TC1900007748.hg.1 ENST00000590247.6 Ensembl
          Affymetrix Human Clariom D Assay TC1900007748.hg.1 ENST00000592786.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900007748.hg.1 ENST00000592786.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900007748.hg.1 ENST00000592786.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900007748.hg.1 ENST00000592786.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900007748.hg.1 ENST00000592786.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900007748.hg.1 ENST00000592786.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900007748.hg.1 ENST00000592786.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900007748.hg.1 ENST00000592786.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900007748.hg.1 NM_004708.3 Refseq
          Affymetrix Human Clariom D Assay TC1900007748.hg.1 NM_004708.3 Refseq
          Affymetrix Human Clariom D Assay TC1900007748.hg.1 NM_004708.3 Refseq
          Affymetrix Human Clariom D Assay TC1900007748.hg.1 NM_004708.3 Refseq
          Affymetrix Human Clariom D Assay TC1900007748.hg.1 NM_004708.3 Refseq
          Affymetrix Human Clariom D Assay TC1900007748.hg.1 NM_004708.3 Refseq
          Affymetrix Human Clariom D Assay TC1900007748.hg.1 NM_004708.3 Refseq
          Affymetrix Human Clariom D Assay TC1900007748.hg.1 NM_004708.3 Refseq
          Affymetrix Human Clariom D Assay TC1900007748.hg.1 NM_004708.3 Refseq
          Affymetrix Human Clariom D Assay PSR1900156905.hg.1 ENST00000221784.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1900156905.hg.1 ENST00000419343.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1900156905.hg.1 ENST00000586035.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900156905.hg.1 ENST00000586316.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900156905.hg.1 ENST00000590247.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1900156905.hg.1 ENST00000592786.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900156905.hg.1 NM_004708.3 Refseq
          Affymetrix Human Clariom D Assay PSR1900156905.hg.1 TC1900007748.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900156905.hg.1 ENST00000221784.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1900156905.hg.1 ENST00000419343.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1900156905.hg.1 ENST00000586035.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900156905.hg.1 ENST00000586316.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900156905.hg.1 ENST00000590247.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1900156905.hg.1 ENST00000592786.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900156905.hg.1 NM_004708.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline