Search
Search

CTNNA2| Gene Symbol: CTNNA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282597.2 | NP_001269526.1 | 18 - 19 | 2858 | 60 | |
| NM_001320810.1 | NP_001307739.1 | 12 - 13 | 1742 | 60 | ||
| XM_011532555.2 | XP_011530857.1 | 18 - 19 | 2705 | 60 | ||
| XM_011532556.2 | XP_011530858.1 | 19 - 20 | 2901 | 60 | ||
| XM_011532557.2 | XP_011530859.1 | 10 - 11 | 1447 | 60 | ||
| GenBank mRNA | M94151.2 | - | 17 - 18 | 2609 | 60 | |
Set Membership: |
|