Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01093553_m1
          See other PUM2 GE Assays ›
          Gene Symbol:
          PUM2
          Gene Name:
          pumilio RNA binding family member 2
          Gene Aliases:
          PUMH2, PUML2
          Chromosome Location:
          Chr.2: 20248692 - 20352423 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01093553_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PUM2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001282752.1 NP_001269681.1 12 - 13 2120 84
          NM_015317.2 NP_056132.1 13 - 14 2229 84
          XM_005262607.1 XP_005262664.1 13 - 14 2203 84
          XM_006711973.1 XP_006712036.1 13 - 14 2000 84
          XM_006711974.2 XP_006712037.1 13 - 14 2040 84
          XM_006711975.2 XP_006712038.1 14 - 15 2164 84
          XM_011532719.1 XP_011531021.1 15 - 16 2399 84
          XM_011532720.2 XP_011531022.1 15 - 16 2408 84
          XM_011532721.2 XP_011531023.1 14 - 15 2200 84
          XM_011532723.1 XP_011531025.1 12 - 13 1830 84
          XM_017003701.1 XP_016859190.1 13 - 14 2000 84
          XM_017003702.1 XP_016859191.1 13 - 14 2040 84
          XM_017003703.1 XP_016859192.1 14 - 15 2228 84
          XM_017003704.1 XP_016859193.1 16 - 17 2546 84
          XM_017003705.1 XP_016859194.1 16 - 17 2533 84
          XM_017003706.1 XP_016859195.1 15 - 16 2325 84
          XM_017003714.1 XP_016859203.1 9 - 10 1609 84
          GenBank mRNA AF315591.1 - 12 - 13 1980 84
          AK304198.1 - 12 - 13 1890 84
          BC024218.2 - 1 - 2 226 84
          BC112046.1 - 12 - 13 1983 8327514 84
          BC112048.1 - 12 - 13 1983 8327516 84
          D87078.3 - 12 - 13 1957 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as HTA transcript cluster(s) TC02001621.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene PUM2

          Panther Classification:

          Molecular Function -

          RNA metabolism protein nucleic acid metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          posttranscriptional regulation of gene expression
          stress granule assembly
          regulation of mRNA stability
          regulation of chromosome segregation
          regulation of gene silencing by miRNA
          positive regulation of RIG-I signaling pathway
          positive regulation of gene silencing by miRNA
          RNA binding
          mRNA 3'-UTR binding
          protein binding
          poly(A) RNA binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR02025963.hg.1 ENST00000319801.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02025963.hg.1 ENST00000338086.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02025963.hg.1 ENST00000361078.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02025963.hg.1 ENST00000403432.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02025963.hg.1 ENST00000536417.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02025963.hg.1 NM_015317.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02025963.hg.1 TC02001621.hg.1 HTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline