Search
Search

UBE3B| Gene Symbol: UBE3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130466.3 | NP_569733.2 | 23 - 24 | 3181 | 71 | |
| NM_183415.2 | NP_904324.1 | 23 - 24 | 2836 | 71 | ||
| XM_005253987.2 | XP_005254044.1 | 23 - 24 | 3061 | 71 | ||
| XM_006719681.3 | XP_006719744.1 | 23 - 24 | 3394 | 71 | ||
| XM_006719682.2 | XP_006719745.1 | 23 - 24 | 3394 | 71 | ||
| XM_011538959.2 | XP_011537261.1 | 22 - 23 | 2943 | 71 | ||
| XM_011538961.1 | XP_011537263.1 | 23 - 24 | 3394 | 71 | ||
| XM_017020195.1 | XP_016875684.1 | 23 - 24 | 3604 | 71 | ||
| GenBank mRNA | AF130072.1 | - | 21 - 22 | 2573 | 71 | |
| AF251046.2 | - | 23 - 24 | 3151 | 71 | ||
| AK127227.1 | - | 8 - 9 | 1968 | 71 | ||
| AK307385.1 | - | 22 - 23 | 2715 | 71 | ||
| AL096740.1 | - | 24 - 25 | 3247 | 71 | ||
| BC020348.1 | - | 8 - 9 | 875 | 71 | ||
| BC032301.1 | - | 23 - 24 | 2814 | 71 | ||
| BC141880.1 | - | 23 - 24 | 3059 | 40124513 | 71 | |
| BC144684.1 | - | 23 - 24 | 3157 | 71 | ||
| BC150637.1 | - | 23 - 24 | 2812 | 9057007 | 71 | |
| BX537403.1 | - | 23 - 24 | 2826 | 71 | ||
Set Membership: |
|