Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01099016_g1
          See other HFE2 GE Assays ›
          Gene Symbol:
          HFE2
          Gene Name:
          hemochromatosis type 2 (juvenile)
          Gene Aliases:
          HFE2A, HJV, JH, RGMC
          Chromosome Location:
          Chr.1: 146017468 - 146021822 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01099016_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: HFE2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_145277.4 NP_660320.3 2 - 3 797 147
          NM_213653.3 NP_998818.1 3 - 4 983 147
          XM_005272932.1 XP_005272989.1 3 - 4 914 147
          GenBank mRNA AK092682.1 - 2 - 3 707 147
          AK124273.1 - 3 - 4 867 147
          AK223575.1 - 2 - 3 717 147
          AK290831.1 - 2 - 3 709 147
          AK292742.1 - 3 - 4 893 147
          AK315852.1 - 2 - 3 710 147
          AY372521.1 - 2 - 3 658 147
          BC085604.1 - 2 - 3 650 6198223 147

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100015599.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01001089.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene HFE2

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          cellular iron ion homeostasis
          protein autoprocessing
          BMP signaling pathway
          negative regulation of BMP signaling pathway
          activin receptor signaling pathway
          positive regulation of transcription from RNA polymerase II promoter
          iron ion homeostasis
          cellular response to BMP stimulus
          glycoprotein binding
          receptor binding
          protein binding
          coreceptor activity
          BMP binding
          BMP receptor activity
          transferrin receptor binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR01017156.hg.1 ENST00000336751.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01017156.hg.1 ENST00000357836.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01017156.hg.1 ENST00000475797.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01017156.hg.1 ENST00000497365.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01017156.hg.1 NM_145277.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01017156.hg.1 NM_202004.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01017156.hg.1 NM_213652.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01017156.hg.1 NM_213653.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01017156.hg.1 TC01001089.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0100015599.hg.1 ENST00000336751.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015599.hg.1 ENST00000336751.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015599.hg.1 ENST00000336751.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015599.hg.1 ENST00000336751.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015599.hg.1 ENST00000336751.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015599.hg.1 ENST00000336751.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015599.hg.1 ENST00000336751.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015599.hg.1 ENST00000336751.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015599.hg.1 ENST00000336751.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015599.hg.1 ENST00000336751.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015599.hg.1 ENST00000357836.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015599.hg.1 ENST00000357836.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015599.hg.1 ENST00000357836.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015599.hg.1 ENST00000357836.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015599.hg.1 ENST00000357836.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015599.hg.1 ENST00000357836.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015599.hg.1 ENST00000357836.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015599.hg.1 ENST00000357836.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015599.hg.1 ENST00000357836.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015599.hg.1 ENST00000357836.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015599.hg.1 ENST00000421822.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015599.hg.1 ENST00000421822.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015599.hg.1 ENST00000421822.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015599.hg.1 ENST00000421822.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015599.hg.1 ENST00000421822.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015599.hg.1 ENST00000421822.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015599.hg.1 ENST00000475797.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015599.hg.1 ENST00000475797.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015599.hg.1 ENST00000475797.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015599.hg.1 ENST00000475797.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015599.hg.1 ENST00000497365.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015599.hg.1 ENST00000497365.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015599.hg.1 ENST00000497365.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015599.hg.1 ENST00000497365.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015599.hg.1 ENST00000497365.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015599.hg.1 NM_145277.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015599.hg.1 NM_145277.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015599.hg.1 NM_145277.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015599.hg.1 NM_145277.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015599.hg.1 NM_145277.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015599.hg.1 NM_145277.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015599.hg.1 NM_145277.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015599.hg.1 NM_145277.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015599.hg.1 NM_145277.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015599.hg.1 NM_202004.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015599.hg.1 NM_202004.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015599.hg.1 NM_202004.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015599.hg.1 NM_202004.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015599.hg.1 NM_202004.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015599.hg.1 NM_202004.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015599.hg.1 NM_213652.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015599.hg.1 NM_213652.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015599.hg.1 NM_213652.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015599.hg.1 NM_213652.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015599.hg.1 NM_213652.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015599.hg.1 NM_213653.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015599.hg.1 NM_213653.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015599.hg.1 NM_213653.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015599.hg.1 NM_213653.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015599.hg.1 NM_213653.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015599.hg.1 NM_213653.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015599.hg.1 NM_213653.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015599.hg.1 NM_213653.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015599.hg.1 NM_213653.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015599.hg.1 NM_213653.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015599.hg.1 NM_213653.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100216699.hg.1 ENST00000336751.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0100216699.hg.1 ENST00000357836.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100216699.hg.1 ENST00000475797.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100216699.hg.1 ENST00000497365.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100216699.hg.1 NM_145277.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100216699.hg.1 NM_202004.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100216699.hg.1 NM_213652.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100216699.hg.1 NM_213653.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100216699.hg.1 ENST00000336751.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0100216699.hg.1 ENST00000357836.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100216699.hg.1 ENST00000475797.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100216699.hg.1 ENST00000497365.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100216699.hg.1 NM_145277.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100216699.hg.1 NM_202004.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100216699.hg.1 NM_213652.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100216699.hg.1 NM_213653.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100216699.hg.1 TC0100015599.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0100015599.hg.1 ENST00000336751.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100015599.hg.1 ENST00000336751.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100015599.hg.1 ENST00000336751.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100015599.hg.1 ENST00000336751.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100015599.hg.1 ENST00000336751.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100015599.hg.1 ENST00000336751.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100015599.hg.1 ENST00000336751.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100015599.hg.1 ENST00000336751.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100015599.hg.1 ENST00000336751.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100015599.hg.1 ENST00000336751.9 Ensembl
          Affymetrix Human Clariom D Assay TC0100015599.hg.1 ENST00000357836.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100015599.hg.1 ENST00000357836.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100015599.hg.1 ENST00000357836.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100015599.hg.1 ENST00000357836.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100015599.hg.1 ENST00000357836.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100015599.hg.1 ENST00000357836.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100015599.hg.1 ENST00000357836.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100015599.hg.1 ENST00000357836.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100015599.hg.1 ENST00000357836.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100015599.hg.1 ENST00000357836.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100015599.hg.1 ENST00000421822.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100015599.hg.1 ENST00000421822.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100015599.hg.1 ENST00000421822.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100015599.hg.1 ENST00000421822.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100015599.hg.1 ENST00000421822.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100015599.hg.1 ENST00000421822.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100015599.hg.1 ENST00000475797.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100015599.hg.1 ENST00000475797.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100015599.hg.1 ENST00000475797.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100015599.hg.1 ENST00000475797.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100015599.hg.1 ENST00000497365.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100015599.hg.1 ENST00000497365.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100015599.hg.1 ENST00000497365.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100015599.hg.1 ENST00000497365.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100015599.hg.1 ENST00000497365.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100015599.hg.1 NM_145277.4 Refseq
          Affymetrix Human Clariom D Assay TC0100015599.hg.1 NM_145277.4 Refseq
          Affymetrix Human Clariom D Assay TC0100015599.hg.1 NM_145277.4 Refseq
          Affymetrix Human Clariom D Assay TC0100015599.hg.1 NM_145277.4 Refseq
          Affymetrix Human Clariom D Assay TC0100015599.hg.1 NM_145277.4 Refseq
          Affymetrix Human Clariom D Assay TC0100015599.hg.1 NM_145277.4 Refseq
          Affymetrix Human Clariom D Assay TC0100015599.hg.1 NM_145277.4 Refseq
          Affymetrix Human Clariom D Assay TC0100015599.hg.1 NM_145277.4 Refseq
          Affymetrix Human Clariom D Assay TC0100015599.hg.1 NM_145277.4 Refseq
          Affymetrix Human Clariom D Assay TC0100015599.hg.1 NM_202004.3 Refseq
          Affymetrix Human Clariom D Assay TC0100015599.hg.1 NM_202004.3 Refseq
          Affymetrix Human Clariom D Assay TC0100015599.hg.1 NM_202004.3 Refseq
          Affymetrix Human Clariom D Assay TC0100015599.hg.1 NM_202004.3 Refseq
          Affymetrix Human Clariom D Assay TC0100015599.hg.1 NM_202004.3 Refseq
          Affymetrix Human Clariom D Assay TC0100015599.hg.1 NM_202004.3 Refseq
          Affymetrix Human Clariom D Assay TC0100015599.hg.1 NM_213652.3 Refseq
          Affymetrix Human Clariom D Assay TC0100015599.hg.1 NM_213652.3 Refseq
          Affymetrix Human Clariom D Assay TC0100015599.hg.1 NM_213652.3 Refseq
          Affymetrix Human Clariom D Assay TC0100015599.hg.1 NM_213652.3 Refseq
          Affymetrix Human Clariom D Assay TC0100015599.hg.1 NM_213652.3 Refseq
          Affymetrix Human Clariom D Assay TC0100015599.hg.1 NM_213653.3 Refseq
          Affymetrix Human Clariom D Assay TC0100015599.hg.1 NM_213653.3 Refseq
          Affymetrix Human Clariom D Assay TC0100015599.hg.1 NM_213653.3 Refseq
          Affymetrix Human Clariom D Assay TC0100015599.hg.1 NM_213653.3 Refseq
          Affymetrix Human Clariom D Assay TC0100015599.hg.1 NM_213653.3 Refseq
          Affymetrix Human Clariom D Assay TC0100015599.hg.1 NM_213653.3 Refseq
          Affymetrix Human Clariom D Assay TC0100015599.hg.1 NM_213653.3 Refseq
          Affymetrix Human Clariom D Assay TC0100015599.hg.1 NM_213653.3 Refseq
          Affymetrix Human Clariom D Assay TC0100015599.hg.1 NM_213653.3 Refseq
          Affymetrix Human Clariom D Assay TC0100015599.hg.1 NM_213653.3 Refseq
          Affymetrix Human Clariom D Assay TC0100015599.hg.1 NM_213653.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline