Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01099854_m1
          See other GHITM GE Assays ›
          Gene Symbol:
          GHITM
          Gene Name:
          growth hormone inducible transmembrane protein
          Gene Aliases:
          DERP2, HSPC282, MICS1, My021, PTD010, TMBIM5
          Chromosome Location:
          Chr.10: 84139429 - 84153555 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01099854_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: GHITM
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_014394.2 NP_055209.2 2 - 3 334 71
          GenBank mRNA AB009685.1 - 1 - 2 130 71
          AF060923.1 - 2 - 3 208 71
          AF078863.1 - 2 - 3 259 71
          AF131820.1 - 2 - 3 261 71
          AF161400.1 - 2 - 3 243 71
          AK098770.1 - 2 - 3 207 71
          AK130094.1 - 2 - 3 237 71
          AK130097.1 - 2 - 3 202 71
          AK225318.1 - 2 - 3 211 71
          AK292864.1 - 2 - 3 202 71
          AK297960.1 - 2 - 3 201 71
          AK299937.1 - 240 71
          AL136713.1 - 2 - 3 244 71
          AY358824.1 - 2 - 3 209 71
          BC010354.1 - 2 - 3 205 4093037 71
          BC022872.1 - 2 - 3 216 71
          CR533519.1 - 1 - 2 130 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1000008269.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC10000591.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the AK297960.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene GHITM

          Gene Ontology Categories:

          Function(s) Process(es)

          apoptotic process
          biological_process
          molecular_function
          protein binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR10006426.hg.1 ENST00000339736.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10006426.hg.1 ENST00000372134.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10006426.hg.1 ENST00000436406.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10006426.hg.1 ENST00000538477.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10006426.hg.1 NM_014394.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10006426.hg.1 TC10000591.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR10006424.hg.1 ENST00000372134.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10006424.hg.1 ENST00000436406.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10006424.hg.1 ENST00000538477.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10006424.hg.1 NM_014394.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10006424.hg.1 TC10000591.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR10006427.hg.1 ENST00000339736.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10006427.hg.1 ENST00000372134.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10006427.hg.1 ENST00000436406.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10006427.hg.1 ENST00000538477.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10006427.hg.1 NM_014394.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10006427.hg.1 TC10000591.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR10006429.hg.1 ENST00000339736.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10006429.hg.1 ENST00000372134.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10006429.hg.1 ENST00000436406.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10006429.hg.1 ENST00000538477.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10006429.hg.1 NM_014394.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10006429.hg.1 TC10000591.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR10006422.hg.1 ENST00000372134.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10006422.hg.1 NM_014394.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10006422.hg.1 TC10000591.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR10006428.hg.1 ENST00000339736.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10006428.hg.1 ENST00000372134.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10006428.hg.1 ENST00000436406.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10006428.hg.1 NM_014394.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10006428.hg.1 TC10000591.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1000008269.hg.1 ENST00000372134.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008269.hg.1 ENST00000372134.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008269.hg.1 ENST00000372134.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008269.hg.1 ENST00000372134.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008269.hg.1 ENST00000372134.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008269.hg.1 ENST00000372134.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008269.hg.1 ENST00000372134.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008269.hg.1 ENST00000372134.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008269.hg.1 ENST00000372134.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008269.hg.1 ENST00000372134.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008269.hg.1 ENST00000372134.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008269.hg.1 ENST00000372134.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008269.hg.1 ENST00000372134.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008269.hg.1 ENST00000372134.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008269.hg.1 ENST00000618907.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008269.hg.1 ENST00000618907.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008269.hg.1 ENST00000618907.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008269.hg.1 ENST00000618907.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008269.hg.1 ENST00000618907.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008269.hg.1 ENST00000618907.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008269.hg.1 ENST00000618907.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008269.hg.1 ENST00000618907.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008269.hg.1 ENST00000618907.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008269.hg.1 ENST00000618907.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008269.hg.1 ENST00000618907.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008269.hg.1 ENST00000618907.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008269.hg.1 ENST00000618907.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008269.hg.1 ENST00000618907.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008269.hg.1 NM_014394.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008269.hg.1 NM_014394.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008269.hg.1 NM_014394.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008269.hg.1 NM_014394.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008269.hg.1 NM_014394.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008269.hg.1 NM_014394.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008269.hg.1 NM_014394.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008269.hg.1 NM_014394.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008269.hg.1 NM_014394.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008269.hg.1 NM_014394.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008269.hg.1 NM_014394.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008269.hg.1 NM_014394.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008269.hg.1 NM_014394.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008269.hg.1 NM_014394.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008269.hg.1 NM_014394.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10006431.hg.1 ENST00000339736.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10006431.hg.1 ENST00000372134.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10006431.hg.1 ENST00000436406.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10006431.hg.1 ENST00000538477.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10006431.hg.1 NM_014394.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10006431.hg.1 TC10000591.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1000155987.hg.1 ENST00000372134.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1000155987.hg.1 ENST00000618907.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1000155987.hg.1 NM_014394.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000155987.hg.1 ENST00000372134.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1000155987.hg.1 ENST00000618907.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1000155987.hg.1 NM_014394.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000155987.hg.1 TC1000008269.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000155985.hg.1 ENST00000372134.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1000155985.hg.1 NM_014394.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000155985.hg.1 ENST00000372134.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1000155985.hg.1 NM_014394.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000155985.hg.1 TC1000008269.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1000008269.hg.1 ENST00000372134.5 Ensembl
          Affymetrix Human Clariom D Assay TC1000008269.hg.1 ENST00000372134.5 Ensembl
          Affymetrix Human Clariom D Assay TC1000008269.hg.1 ENST00000372134.5 Ensembl
          Affymetrix Human Clariom D Assay TC1000008269.hg.1 ENST00000372134.5 Ensembl
          Affymetrix Human Clariom D Assay TC1000008269.hg.1 ENST00000372134.5 Ensembl
          Affymetrix Human Clariom D Assay TC1000008269.hg.1 ENST00000372134.5 Ensembl
          Affymetrix Human Clariom D Assay TC1000008269.hg.1 ENST00000372134.5 Ensembl
          Affymetrix Human Clariom D Assay TC1000008269.hg.1 ENST00000372134.5 Ensembl
          Affymetrix Human Clariom D Assay TC1000008269.hg.1 ENST00000372134.5 Ensembl
          Affymetrix Human Clariom D Assay TC1000008269.hg.1 ENST00000372134.5 Ensembl
          Affymetrix Human Clariom D Assay TC1000008269.hg.1 ENST00000372134.5 Ensembl
          Affymetrix Human Clariom D Assay TC1000008269.hg.1 ENST00000372134.5 Ensembl
          Affymetrix Human Clariom D Assay TC1000008269.hg.1 ENST00000372134.5 Ensembl
          Affymetrix Human Clariom D Assay TC1000008269.hg.1 ENST00000372134.5 Ensembl
          Affymetrix Human Clariom D Assay TC1000008269.hg.1 ENST00000618907.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000008269.hg.1 ENST00000618907.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000008269.hg.1 ENST00000618907.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000008269.hg.1 ENST00000618907.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000008269.hg.1 ENST00000618907.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000008269.hg.1 ENST00000618907.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000008269.hg.1 ENST00000618907.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000008269.hg.1 ENST00000618907.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000008269.hg.1 ENST00000618907.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000008269.hg.1 ENST00000618907.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000008269.hg.1 ENST00000618907.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000008269.hg.1 ENST00000618907.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000008269.hg.1 ENST00000618907.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000008269.hg.1 ENST00000618907.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000008269.hg.1 NM_014394.2 Refseq
          Affymetrix Human Clariom D Assay TC1000008269.hg.1 NM_014394.2 Refseq
          Affymetrix Human Clariom D Assay TC1000008269.hg.1 NM_014394.2 Refseq
          Affymetrix Human Clariom D Assay TC1000008269.hg.1 NM_014394.2 Refseq
          Affymetrix Human Clariom D Assay TC1000008269.hg.1 NM_014394.2 Refseq
          Affymetrix Human Clariom D Assay TC1000008269.hg.1 NM_014394.2 Refseq
          Affymetrix Human Clariom D Assay TC1000008269.hg.1 NM_014394.2 Refseq
          Affymetrix Human Clariom D Assay TC1000008269.hg.1 NM_014394.2 Refseq
          Affymetrix Human Clariom D Assay TC1000008269.hg.1 NM_014394.2 Refseq
          Affymetrix Human Clariom D Assay TC1000008269.hg.1 NM_014394.2 Refseq
          Affymetrix Human Clariom D Assay TC1000008269.hg.1 NM_014394.2 Refseq
          Affymetrix Human Clariom D Assay TC1000008269.hg.1 NM_014394.2 Refseq
          Affymetrix Human Clariom D Assay TC1000008269.hg.1 NM_014394.2 Refseq
          Affymetrix Human Clariom D Assay TC1000008269.hg.1 NM_014394.2 Refseq
          Affymetrix Human Clariom D Assay TC1000008269.hg.1 NM_014394.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000155986.hg.1 ENST00000372134.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1000155986.hg.1 ENST00000618907.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1000155986.hg.1 NM_014394.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000155986.hg.1 ENST00000372134.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1000155986.hg.1 ENST00000618907.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1000155986.hg.1 NM_014394.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000155986.hg.1 TC1000008269.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000155988.hg.1 ENST00000372134.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1000155988.hg.1 ENST00000618907.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1000155988.hg.1 NM_014394.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000155988.hg.1 ENST00000372134.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1000155988.hg.1 ENST00000618907.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1000155988.hg.1 NM_014394.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000155988.hg.1 TC1000008269.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000155989.hg.1 ENST00000372134.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1000155989.hg.1 ENST00000618907.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1000155989.hg.1 NM_014394.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000155989.hg.1 ENST00000372134.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1000155989.hg.1 ENST00000618907.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1000155989.hg.1 NM_014394.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000155989.hg.1 TC1000008269.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000155990.hg.1 ENST00000372134.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1000155990.hg.1 ENST00000618907.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1000155990.hg.1 NM_014394.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000155990.hg.1 ENST00000372134.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1000155990.hg.1 ENST00000618907.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1000155990.hg.1 NM_014394.2 Refseq
          Affymetrix Human Clariom D Assay PSR1000155990.hg.1 TC1000008269.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 180427 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline