Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01100158_m1
          See other SPOPL GE Assays ›
          Gene Symbol:
          SPOPL
          Gene Name:
          speckle type BTB/POZ protein like
          Gene Aliases:
          BTBD33
          Chromosome Location:
          Chr.2: 138501780 - 138573235 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01100158_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SPOPL
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001001664.2 NP_001001664.1 1 - 2 338 85
          XM_005263655.4 XP_005263712.1 1 - 2 204 85
          XM_005263656.4 XP_005263713.1 1 - 2 204 85
          XM_005263657.4 XP_005263714.1 1 - 2 214 85
          GenBank mRNA BC071613.1 - 1 - 2 317 5275316 85
          CN315400.1 - 1 - 2 341 85

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0200009487.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC02000894.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the BC071613.1,NM_001001664.2,XM_005263655.4,XM_005263656.4,XM_005263657.4 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene SPOPL

          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of proteolysis
          negative regulation of protein ubiquitination
          protein ubiquitination involved in ubiquitin-dependent protein catabolic process
          proteasome-mediated ubiquitin-dependent protein catabolic process
          protein binding
          ubiquitin protein ligase binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0400007752.hg.1 ENST00000506559.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009487.hg.1 ENST00000280098.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009487.hg.1 ENST00000280098.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009487.hg.1 ENST00000280098.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009487.hg.1 ENST00000280098.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009487.hg.1 ENST00000280098.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009487.hg.1 ENST00000280098.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009487.hg.1 ENST00000280098.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009487.hg.1 ENST00000280098.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009487.hg.1 ENST00000280098.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009487.hg.1 ENST00000280098.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009487.hg.1 ENST00000280098.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009487.hg.1 ENST00000280098.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009487.hg.1 ENST00000280098.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009487.hg.1 ENST00000280098.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009487.hg.1 ENST00000280098.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009487.hg.1 ENST00000280098.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009487.hg.1 ENST00000280098.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009487.hg.1 ENST00000280098.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009487.hg.1 ENST00000280098.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009487.hg.1 ENST00000280098.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009487.hg.1 ENST00000280098.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009487.hg.1 ENST00000420679.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009487.hg.1 ENST00000420679.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009487.hg.1 ENST00000420679.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009487.hg.1 ENST00000420679.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009487.hg.1 ENST00000420679.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009487.hg.1 ENST00000420679.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009487.hg.1 ENST00000420679.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009487.hg.1 ENST00000420679.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009487.hg.1 ENST00000420679.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009487.hg.1 ENST00000420679.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009487.hg.1 ENST00000420679.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009487.hg.1 ENST00000420679.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009487.hg.1 ENST00000420679.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009487.hg.1 ENST00000420679.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009487.hg.1 ENST00000420679.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009487.hg.1 ENST00000430968.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009487.hg.1 ENST00000430968.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009487.hg.1 ENST00000430968.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009487.hg.1 ENST00000430968.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009487.hg.1 ENST00000430968.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009487.hg.1 ENST00000430968.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009487.hg.1 ENST00000449869.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009487.hg.1 ENST00000449869.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009487.hg.1 ENST00000449869.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009487.hg.1 ENST00000458007.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009487.hg.1 ENST00000458007.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009487.hg.1 ENST00000458007.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009487.hg.1 ENST00000458007.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009487.hg.1 ENST00000467775.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009487.hg.1 ENST00000467775.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009487.hg.1 ENST00000467775.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009487.hg.1 ENST00000467775.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009487.hg.1 NM_001001664.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009487.hg.1 NM_001001664.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009487.hg.1 NM_001001664.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009487.hg.1 NM_001001664.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009487.hg.1 NM_001001664.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009487.hg.1 NM_001001664.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009487.hg.1 NM_001001664.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009487.hg.1 NM_001001664.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009487.hg.1 NM_001001664.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009487.hg.1 NM_001001664.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009487.hg.1 NM_001001664.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009487.hg.1 NM_001001664.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009487.hg.1 NM_001001664.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009487.hg.1 NM_001001664.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009487.hg.1 NM_001001664.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009487.hg.1 NM_001001664.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009487.hg.1 NM_001001664.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009487.hg.1 NM_001001664.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009487.hg.1 NM_001001664.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009487.hg.1 NM_001001664.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02013240.hg.1 ENST00000280098.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02013240.hg.1 NM_001001664.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02013240.hg.1 TC02000894.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR02013241.hg.1 ENST00000280098.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02013241.hg.1 NM_001001664.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02013241.hg.1 TC02000894.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR02013242.hg.1 ENST00000280098.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02013242.hg.1 NM_001001664.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02013242.hg.1 TC02000894.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC0400007752.hg.1 ENST00000506559.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200166384.hg.1 ENST00000280098.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0200166384.hg.1 ENST00000430968.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200166384.hg.1 NM_001001664.2 Refseq
          Affymetrix Human Clariom D Assay PSR0200166384.hg.1 ENST00000280098.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0200166384.hg.1 ENST00000430968.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200166384.hg.1 NM_001001664.2 Refseq
          Affymetrix Human Clariom D Assay PSR0200166384.hg.1 TC0200009487.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0200009487.hg.1 ENST00000280098.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200009487.hg.1 ENST00000280098.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200009487.hg.1 ENST00000280098.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200009487.hg.1 ENST00000280098.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200009487.hg.1 ENST00000280098.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200009487.hg.1 ENST00000280098.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200009487.hg.1 ENST00000280098.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200009487.hg.1 ENST00000280098.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200009487.hg.1 ENST00000280098.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200009487.hg.1 ENST00000280098.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200009487.hg.1 ENST00000280098.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200009487.hg.1 ENST00000280098.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200009487.hg.1 ENST00000280098.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200009487.hg.1 ENST00000280098.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200009487.hg.1 ENST00000280098.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200009487.hg.1 ENST00000280098.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200009487.hg.1 ENST00000280098.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200009487.hg.1 ENST00000280098.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200009487.hg.1 ENST00000280098.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200009487.hg.1 ENST00000280098.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200009487.hg.1 ENST00000280098.8 Ensembl
          Affymetrix Human Clariom D Assay TC0200009487.hg.1 ENST00000420679.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200009487.hg.1 ENST00000420679.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200009487.hg.1 ENST00000420679.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200009487.hg.1 ENST00000420679.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200009487.hg.1 ENST00000420679.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200009487.hg.1 ENST00000420679.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200009487.hg.1 ENST00000420679.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200009487.hg.1 ENST00000420679.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200009487.hg.1 ENST00000420679.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200009487.hg.1 ENST00000420679.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200009487.hg.1 ENST00000420679.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200009487.hg.1 ENST00000420679.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200009487.hg.1 ENST00000420679.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200009487.hg.1 ENST00000420679.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200009487.hg.1 ENST00000420679.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200009487.hg.1 ENST00000430968.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200009487.hg.1 ENST00000430968.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200009487.hg.1 ENST00000430968.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200009487.hg.1 ENST00000430968.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200009487.hg.1 ENST00000430968.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200009487.hg.1 ENST00000430968.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200009487.hg.1 ENST00000449869.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200009487.hg.1 ENST00000449869.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200009487.hg.1 ENST00000449869.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200009487.hg.1 ENST00000458007.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200009487.hg.1 ENST00000458007.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200009487.hg.1 ENST00000458007.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200009487.hg.1 ENST00000458007.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200009487.hg.1 ENST00000467775.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200009487.hg.1 ENST00000467775.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200009487.hg.1 ENST00000467775.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200009487.hg.1 ENST00000467775.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200009487.hg.1 NM_001001664.2 Refseq
          Affymetrix Human Clariom D Assay TC0200009487.hg.1 NM_001001664.2 Refseq
          Affymetrix Human Clariom D Assay TC0200009487.hg.1 NM_001001664.2 Refseq
          Affymetrix Human Clariom D Assay TC0200009487.hg.1 NM_001001664.2 Refseq
          Affymetrix Human Clariom D Assay TC0200009487.hg.1 NM_001001664.2 Refseq
          Affymetrix Human Clariom D Assay TC0200009487.hg.1 NM_001001664.2 Refseq
          Affymetrix Human Clariom D Assay TC0200009487.hg.1 NM_001001664.2 Refseq
          Affymetrix Human Clariom D Assay TC0200009487.hg.1 NM_001001664.2 Refseq
          Affymetrix Human Clariom D Assay TC0200009487.hg.1 NM_001001664.2 Refseq
          Affymetrix Human Clariom D Assay TC0200009487.hg.1 NM_001001664.2 Refseq
          Affymetrix Human Clariom D Assay TC0200009487.hg.1 NM_001001664.2 Refseq
          Affymetrix Human Clariom D Assay TC0200009487.hg.1 NM_001001664.2 Refseq
          Affymetrix Human Clariom D Assay TC0200009487.hg.1 NM_001001664.2 Refseq
          Affymetrix Human Clariom D Assay TC0200009487.hg.1 NM_001001664.2 Refseq
          Affymetrix Human Clariom D Assay TC0200009487.hg.1 NM_001001664.2 Refseq
          Affymetrix Human Clariom D Assay TC0200009487.hg.1 NM_001001664.2 Refseq
          Affymetrix Human Clariom D Assay TC0200009487.hg.1 NM_001001664.2 Refseq
          Affymetrix Human Clariom D Assay TC0200009487.hg.1 NM_001001664.2 Refseq
          Affymetrix Human Clariom D Assay TC0200009487.hg.1 NM_001001664.2 Refseq
          Affymetrix Human Clariom D Assay TC0200009487.hg.1 NM_001001664.2 Refseq
          Affymetrix Human Clariom D Assay PSR0200166383.hg.1 ENST00000280098.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0200166383.hg.1 NM_001001664.2 Refseq
          Affymetrix Human Clariom D Assay PSR0200166383.hg.1 ENST00000280098.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0200166383.hg.1 NM_001001664.2 Refseq
          Affymetrix Human Clariom D Assay PSR0200166383.hg.1 TC0200009487.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200166386.hg.1 ENST00000280098.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0200166386.hg.1 ENST00000420679.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200166386.hg.1 ENST00000430968.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200166386.hg.1 NM_001001664.2 Refseq
          Affymetrix Human Clariom D Assay PSR0200166386.hg.1 ENST00000280098.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0200166386.hg.1 ENST00000420679.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200166386.hg.1 ENST00000430968.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200166386.hg.1 NM_001001664.2 Refseq
          Affymetrix Human Clariom D Assay PSR0200166386.hg.1 TC0200009487.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200166385.hg.1 ENST00000280098.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0200166385.hg.1 ENST00000420679.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200166385.hg.1 ENST00000430968.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200166385.hg.1 NM_001001664.2 Refseq
          Affymetrix Human Clariom D Assay PSR0200166385.hg.1 ENST00000280098.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0200166385.hg.1 ENST00000420679.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200166385.hg.1 ENST00000430968.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200166385.hg.1 NM_001001664.2 Refseq
          Affymetrix Human Clariom D Assay PSR0200166385.hg.1 TC0200009487.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline