| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000357508.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000357508.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000357508.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000357508.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000357508.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000357508.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000357508.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000357508.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000357508.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000357508.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000357508.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000357508.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000357508.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000357508.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000357508.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000357508.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000357508.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000357508.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000357508.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000357508.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000357508.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000357508.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000357508.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000357508.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000357508.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000418644.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000418644.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000418644.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000418644.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000418644.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000418644.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000418644.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000418644.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000418644.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000418644.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000418644.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000418779.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000418779.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000418779.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000418779.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000418779.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000418779.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000418779.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000418779.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000418779.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000421398.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000421398.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000421398.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000421398.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000421398.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000424484.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000424484.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000424484.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000424484.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000424484.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000424484.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000424484.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000424484.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000424484.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000424484.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000427321.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000427321.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000427321.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000427321.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000427336.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000427336.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000427336.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000427336.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000434068.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000434068.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000434068.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000434068.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000434068.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000434520.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000434520.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000440383.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000440383.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000440383.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000440383.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000441519.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000441519.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000441519.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000441519.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000441519.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000441519.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000441519.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000441519.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000441519.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000441519.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000441519.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000441519.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000444525.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000444525.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000444525.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000444525.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000444751.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000444751.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000444751.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000444751.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000444751.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000444751.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000444751.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000445071.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000445071.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000445071.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000445071.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000445071.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000445071.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000445071.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000445071.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000445071.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000445071.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000445071.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000445071.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000445071.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000446792.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000446792.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000446792.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000446792.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000453711.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000453711.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000453711.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000453711.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000453711.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000453711.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000453711.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000455805.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000455805.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000455805.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000455805.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000455805.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000455805.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000455805.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000455805.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000455805.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000458657.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000458657.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000458657.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000458657.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000458657.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000458657.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000458657.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000458657.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000458657.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000458657.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000458657.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000458657.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000458657.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000458657.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000458657.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000458657.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000458657.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000458657.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000458657.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000458657.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000535358.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000535358.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000535358.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000535358.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000535358.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000535358.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000535358.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000535358.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000535358.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000535358.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000535358.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000535358.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000535358.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000535358.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000535358.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000535358.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000535358.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000535358.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000535358.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000568406.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000568406.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000568406.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000568406.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000568406.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000568406.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
ENST00000568406.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
NM_001145636.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
NM_001145636.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
NM_001145636.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
NM_001145636.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
NM_001145636.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
NM_001145636.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
NM_001145636.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
NM_001145636.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
NM_001145636.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
NM_001145636.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
NM_001145636.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
NM_001145636.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
NM_001145636.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
NM_001145636.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
NM_001145636.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
NM_001145636.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
NM_001145636.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
NM_001145636.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008100.hg.1 |
NM_001145636.1 |
Refseq |