Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01104474_m1
          See other SLC6A1 GE Assays ›
          Gene Symbol:
          SLC6A1
          Gene Name:
          solute carrier family 6 member 1
          Gene Aliases:
          GABATHG, GABATR, GAT1, MAE
          Chromosome Location:
          Chr.3: 10992718 - 11039249 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01104474_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SLC6A1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_003042.3 NP_003033.3 4 - 5 780 84
          XM_005265410.4 XP_005265467.1 4 - 5 2137 84
          XM_005265411.4 XP_005265468.1 4 - 5 2076 84
          XM_006713306.3 XP_006713369.1 4 - 5 735 84
          XM_011534025.2 XP_011532327.1 4 - 5 2827 84
          XM_011534027.2 XP_011532329.1 4 - 5 2766 84
          XM_017007071.1 XP_016862560.1 5 - 6 2559 84
          XM_017007072.1 XP_016862561.1 5 - 6 1869 84
          GenBank mRNA BC033904.1 - 4 - 5 748 5264672 84
          BC057291.1 - 4 - 5 760 84
          X54673.1 - 3 - 4 603 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0300006599.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC03000063.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene SLC6A1

          Panther Classification:

          Molecular Function -

          primary active transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          neurotransmitter transport
          chemical synaptic transmission
          learning
          response to toxic substance
          response to sucrose
          response to lead ion
          positive regulation of gamma-aminobutyric acid secretion
          response to purine-containing compound
          gamma-aminobutyric acid transport
          negative regulation of synaptic transmission, GABAergic
          response to estradiol
          response to cocaine
          protein homooligomerization
          response to calcium ion
          gamma-aminobutyric acid import
          transmembrane transport
          mitophagy in response to mitochondrial depolarization
          neurotransmitter:sodium symporter activity
          gamma-aminobutyric acid:sodium symporter activity
          metal ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR03001378.hg.1 ENST00000287766.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03001378.hg.1 ENST00000536032.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03001378.hg.1 NM_003042.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03001378.hg.1 TC03000063.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000425938.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000425938.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000425938.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000425938.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000425938.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000425938.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000460480.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000460480.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000460480.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000460480.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000460480.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000462473.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000462473.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000462473.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000462473.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000462473.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000462473.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000462473.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000495636.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000495636.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000495636.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000495636.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 ENST00000495636.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Clariom D Assay PSR0300145799.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0300145799.hg.1 ENST00000460480.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300145799.hg.1 NM_003042.3 Refseq
          Affymetrix Human Clariom D Assay PSR0300145799.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0300145799.hg.1 ENST00000460480.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300145799.hg.1 NM_003042.3 Refseq
          Affymetrix Human Clariom D Assay PSR0300145799.hg.1 TC0300006599.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000287766.8 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000425938.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000425938.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000425938.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000425938.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000425938.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000425938.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000460480.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000460480.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000460480.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000460480.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000460480.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000462473.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000462473.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000462473.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000462473.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000462473.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000462473.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000462473.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000495636.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000495636.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000495636.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000495636.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 ENST00000495636.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 NM_003042.3 Refseq
          Affymetrix Human Clariom D Assay TC0300006599.hg.1 NM_003042.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline