| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000311638.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000311638.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000311638.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000311638.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000311638.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000311638.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000311638.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000311638.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000380837.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000380837.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000380837.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000380837.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000380837.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000380837.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000380837.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000502620.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000502620.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000502620.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000502620.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000502620.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000502620.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000503831.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000503831.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000503831.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000503831.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000503831.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000503831.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000504190.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000504190.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000504190.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000504190.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000504190.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000504190.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000504190.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000504190.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000504218.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000504218.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000504218.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000504218.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000504218.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000504384.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000504384.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000504384.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000504384.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000504384.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000504384.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000506261.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000506261.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000506261.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000507556.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000507556.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000507556.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000507556.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000507556.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000507556.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000507556.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000507556.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000507556.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000507557.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000507557.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000507557.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000507557.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000507557.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000507557.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000507557.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000507885.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000507885.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000507885.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000507885.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000507885.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000507885.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000507885.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000507885.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000508105.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000508105.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000508105.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000508105.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000508105.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000508105.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000514480.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000514480.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000514480.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000514480.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000514480.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000514480.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
ENST00000514480.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
NM_144721.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
NM_144721.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
NM_144721.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
NM_144721.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
NM_144721.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
NM_144721.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
NM_144721.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
NM_144721.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007876.hg.1 |
NM_144721.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000311638.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000311638.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000311638.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000311638.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000311638.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000311638.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000311638.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000311638.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000380837.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000380837.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000380837.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000380837.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000380837.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000380837.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000380837.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000502620.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000502620.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000502620.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000502620.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000502620.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000502620.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000503831.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000503831.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000503831.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000503831.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000503831.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000503831.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000504190.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000504190.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000504190.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000504190.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000504190.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000504190.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000504190.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000504190.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000504218.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000504218.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000504218.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000504218.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000504218.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000504384.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000504384.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000504384.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000504384.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000504384.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000504384.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000506261.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000506261.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000506261.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000507556.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000507556.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000507556.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000507556.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000507556.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000507556.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000507556.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000507556.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000507556.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000507557.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000507557.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000507557.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000507557.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000507557.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000507557.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000507557.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000507885.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000507885.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000507885.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000507885.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000507885.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000507885.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000507885.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000507885.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000508105.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000508105.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000508105.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000508105.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000508105.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000508105.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000514480.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000514480.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000514480.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000514480.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000514480.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000514480.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
ENST00000514480.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
NM_144721.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
NM_144721.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
NM_144721.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
NM_144721.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
NM_144721.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
NM_144721.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
NM_144721.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
NM_144721.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400007876.hg.1 |
NM_144721.5 |
Refseq |