Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01110096_g1
          See other LILRA4 GE Assays ›
          Gene Symbol:
          LILRA4
          Gene Name:
          leukocyte immunoglobulin like receptor A4
          Gene Aliases:
          CD85g, ILT7
          Chromosome Location:
          Chr.19: 314530 - 320509 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01110096_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LILRA4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012276.5 NP_036408.4 1 - 2 105 97
          GenBank mRNA BC109199.1 - 1 - 2 58 97
          BI834924.1 - 1 - 2 93 97
          DA983933.1 - 1 - 2 112 97

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1900012043.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC19002720.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene LILRA4

          Gene Ontology Categories:

          Function(s) Process(es)

          immune system process
          immune response-regulating signaling pathway
          cell surface receptor signaling pathway
          cytokine-mediated signaling pathway
          negative regulation of interferon-alpha production
          negative regulation of tumor necrosis factor production
          negative regulation of toll-like receptor 7 signaling pathway
          negative regulation of toll-like receptor 9 signaling pathway
          Fc-epsilon receptor signaling pathway
          innate immune response
          receptor binding
          protein binding
          coreceptor activity
          inhibitory MHC class I receptor activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1900012043.hg.1 ENST00000291759.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900012043.hg.1 ENST00000291759.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900012043.hg.1 ENST00000291759.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900012043.hg.1 ENST00000291759.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900012043.hg.1 ENST00000291759.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900012043.hg.1 ENST00000291759.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900012043.hg.1 ENST00000291759.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900012043.hg.1 ENST00000291759.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900012043.hg.1 ENST00000291759.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900012043.hg.1 ENST00000291759.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900012043.hg.1 ENST00000291759.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900012043.hg.1 ENST00000291759.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900012043.hg.1 ENST00000421056.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900012043.hg.1 ENST00000421056.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900012043.hg.1 ENST00000421056.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900012043.hg.1 ENST00000421056.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900012043.hg.1 ENST00000595581.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900012043.hg.1 ENST00000595581.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900012043.hg.1 ENST00000595581.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900012043.hg.1 ENST00000291759.4 Ensembl
          Affymetrix Human Clariom D Assay TC1900012043.hg.1 ENST00000291759.4 Ensembl
          Affymetrix Human Clariom D Assay TC1900012043.hg.1 ENST00000291759.4 Ensembl
          Affymetrix Human Clariom D Assay TC1900012043.hg.1 ENST00000291759.4 Ensembl
          Affymetrix Human Clariom D Assay TC1900012043.hg.1 ENST00000291759.4 Ensembl
          Affymetrix Human Clariom D Assay TC1900012043.hg.1 ENST00000291759.4 Ensembl
          Affymetrix Human Clariom D Assay TC1900012043.hg.1 ENST00000291759.4 Ensembl
          Affymetrix Human Clariom D Assay TC1900012043.hg.1 ENST00000291759.4 Ensembl
          Affymetrix Human Clariom D Assay TC1900012043.hg.1 ENST00000291759.4 Ensembl
          Affymetrix Human Clariom D Assay TC1900012043.hg.1 ENST00000291759.4 Ensembl
          Affymetrix Human Clariom D Assay TC1900012043.hg.1 ENST00000291759.4 Ensembl
          Affymetrix Human Clariom D Assay TC1900012043.hg.1 ENST00000291759.4 Ensembl
          Affymetrix Human Clariom D Assay TC1900012043.hg.1 ENST00000421056.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900012043.hg.1 ENST00000421056.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900012043.hg.1 ENST00000421056.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900012043.hg.1 ENST00000421056.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900012043.hg.1 ENST00000595581.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900012043.hg.1 ENST00000595581.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900012043.hg.1 ENST00000595581.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900208089.hg.1 ENST00000291759.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1900208089.hg.1 ENST00000421056.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1900208089.hg.1 ENST00000291759.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1900208089.hg.1 ENST00000421056.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1900208089.hg.1 TC1900012043.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900208091.hg.1 ENST00000291759.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1900208091.hg.1 ENST00000421056.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1900208091.hg.1 ENST00000291759.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1900208091.hg.1 ENST00000421056.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1900208091.hg.1 TC1900012043.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline