Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01110716_m1
          See other TBX3 GE Assays ›
          Gene Symbol:
          TBX3
          Gene Name:
          T-box 3
          Gene Aliases:
          TBX3-ISO, UMS, XHL
          Chromosome Location:
          Chr.12: 114670254 - 114684164 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01110716_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TBX3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_005996.3 NP_005987.3 2 - 3 1623 143
          GenBank mRNA AF002228.1 - 2 - 3 659 143
          AF170708.2 - 2 - 3 919 143

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene TBX3

          Panther Classification:

          Molecular Function -

          Rel homology transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          skeletal system development
          blood vessel development
          in utero embryonic development
          heart looping
          outflow tract morphogenesis
          atrioventricular bundle cell differentiation
          transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          cell aging
          positive regulation of cell proliferation
          anterior/posterior axis specification, embryo
          organ morphogenesis
          specification of organ position
          stem cell population maintenance
          limbic system development
          male genitalia development
          female genitalia development
          negative regulation of epithelial cell differentiation
          mammary gland development
          luteinizing hormone secretion
          embryonic forelimb morphogenesis
          embryonic hindlimb morphogenesis
          forelimb morphogenesis
          embryonic digit morphogenesis
          negative regulation of apoptotic process
          negative regulation of myoblast differentiation
          positive regulation of cell cycle
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          follicle-stimulating hormone secretion
          mesoderm morphogenesis
          palate development
          ventricular septum morphogenesis
          branching involved in mammary gland duct morphogenesis
          mammary placode formation
          cardiac muscle cell fate commitment
          sinoatrial node cell development
          cellular senescence
          positive regulation of stem cell proliferation
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
          RNA polymerase II transcription factor binding
          RNA polymerase II activating transcription factor binding
          protein binding
          sequence-specific DNA binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline