| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04007163.hg.1 |
ENST00000264790.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04007163.hg.1 |
ENST00000394980.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04007163.hg.1 |
ENST00000394981.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04007163.hg.1 |
ENST00000508372.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04007163.hg.1 |
NM_007351.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04007163.hg.1 |
TC04000493.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008134.hg.1 |
ENST00000264790.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008134.hg.1 |
ENST00000264790.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008134.hg.1 |
ENST00000264790.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008134.hg.1 |
ENST00000264790.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008134.hg.1 |
ENST00000264790.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008134.hg.1 |
ENST00000264790.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008134.hg.1 |
ENST00000264790.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008134.hg.1 |
ENST00000264790.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008134.hg.1 |
ENST00000264790.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008134.hg.1 |
ENST00000264790.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008134.hg.1 |
ENST00000264790.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008134.hg.1 |
ENST00000264790.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008134.hg.1 |
ENST00000394980.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008134.hg.1 |
ENST00000394980.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008134.hg.1 |
ENST00000394980.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008134.hg.1 |
ENST00000394980.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008134.hg.1 |
ENST00000394980.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008134.hg.1 |
ENST00000394980.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008134.hg.1 |
ENST00000394980.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008134.hg.1 |
ENST00000394980.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008134.hg.1 |
ENST00000394980.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008134.hg.1 |
ENST00000394980.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008134.hg.1 |
ENST00000394980.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008134.hg.1 |
ENST00000394980.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008134.hg.1 |
ENST00000394980.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008134.hg.1 |
ENST00000394980.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008134.hg.1 |
ENST00000506328.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008134.hg.1 |
ENST00000506328.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008134.hg.1 |
ENST00000506328.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008134.hg.1 |
ENST00000506328.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008134.hg.1 |
ENST00000508372.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008134.hg.1 |
ENST00000508372.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008134.hg.1 |
ENST00000508372.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008134.hg.1 |
ENST00000508372.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008134.hg.1 |
ENST00000508372.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008134.hg.1 |
ENST00000508372.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008134.hg.1 |
ENST00000508372.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008134.hg.1 |
ENST00000508372.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008134.hg.1 |
ENST00000508372.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008134.hg.1 |
NM_007351.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008134.hg.1 |
NM_007351.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008134.hg.1 |
NM_007351.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008134.hg.1 |
NM_007351.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008134.hg.1 |
NM_007351.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008134.hg.1 |
NM_007351.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008134.hg.1 |
NM_007351.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008134.hg.1 |
NM_007351.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008134.hg.1 |
NM_007351.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008134.hg.1 |
NM_007351.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008134.hg.1 |
NM_007351.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008134.hg.1 |
NM_007351.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008134.hg.1 |
ENST00000264790.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008134.hg.1 |
ENST00000264790.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008134.hg.1 |
ENST00000264790.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008134.hg.1 |
ENST00000264790.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008134.hg.1 |
ENST00000264790.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008134.hg.1 |
ENST00000264790.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008134.hg.1 |
ENST00000264790.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008134.hg.1 |
ENST00000264790.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008134.hg.1 |
ENST00000264790.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008134.hg.1 |
ENST00000264790.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008134.hg.1 |
ENST00000264790.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008134.hg.1 |
ENST00000264790.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008134.hg.1 |
ENST00000394980.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008134.hg.1 |
ENST00000394980.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008134.hg.1 |
ENST00000394980.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008134.hg.1 |
ENST00000394980.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008134.hg.1 |
ENST00000394980.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008134.hg.1 |
ENST00000394980.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008134.hg.1 |
ENST00000394980.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008134.hg.1 |
ENST00000394980.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008134.hg.1 |
ENST00000394980.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008134.hg.1 |
ENST00000394980.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008134.hg.1 |
ENST00000394980.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008134.hg.1 |
ENST00000394980.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008134.hg.1 |
ENST00000394980.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008134.hg.1 |
ENST00000394980.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008134.hg.1 |
ENST00000506328.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008134.hg.1 |
ENST00000506328.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008134.hg.1 |
ENST00000506328.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008134.hg.1 |
ENST00000506328.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008134.hg.1 |
ENST00000508372.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008134.hg.1 |
ENST00000508372.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008134.hg.1 |
ENST00000508372.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008134.hg.1 |
ENST00000508372.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008134.hg.1 |
ENST00000508372.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008134.hg.1 |
ENST00000508372.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008134.hg.1 |
ENST00000508372.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008134.hg.1 |
ENST00000508372.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008134.hg.1 |
ENST00000508372.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008134.hg.1 |
NM_007351.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008134.hg.1 |
NM_007351.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008134.hg.1 |
NM_007351.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008134.hg.1 |
NM_007351.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008134.hg.1 |
NM_007351.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008134.hg.1 |
NM_007351.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008134.hg.1 |
NM_007351.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008134.hg.1 |
NM_007351.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008134.hg.1 |
NM_007351.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008134.hg.1 |
NM_007351.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008134.hg.1 |
NM_007351.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008134.hg.1 |
NM_007351.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400156016.hg.1 |
ENST00000264790.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400156016.hg.1 |
ENST00000394980.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400156016.hg.1 |
ENST00000508372.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400156016.hg.1 |
NM_007351.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400156016.hg.1 |
ENST00000264790.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400156016.hg.1 |
ENST00000394980.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400156016.hg.1 |
ENST00000508372.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400156016.hg.1 |
NM_007351.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400156016.hg.1 |
TC0400008134.hg.1 |
Clariom D TC |