Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01113342_g1
          See other ZW10 GE Assays ›
          Gene Symbol:
          ZW10
          Gene Name:
          zw10 kinetochore protein
          Gene Aliases:
          HZW10, KNTC1AP
          Chromosome Location:
          Chr.11: 113733183 - 113773763 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01113342_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ZW10
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_004724.3 NP_004715.1 8 - 9 1191 132
          XM_017018558.1 XP_016874047.1 7 - 8 939 132
          XM_017018559.1 XP_016874048.1 8 - 9 1147 132
          GenBank mRNA AK303871.1 - 7 - 8 870 132
          AK312925.1 - 8 - 9 1169 132
          BC128264.1 - 8 - 9 1119 40118818 132
          U54996.1 - 8 - 9 1117 132

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1100012353.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC11002305.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene ZW10

          Panther Classification:

          Molecular Function -

          microtubule or microtubule-binding cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          mitotic sister chromatid segregation
          establishment of mitotic spindle orientation
          protein complex assembly
          ER to Golgi vesicle-mediated transport
          retrograde vesicle-mediated transport, Golgi to ER
          Golgi organization
          sister chromatid cohesion
          mitotic metaphase plate congression
          mitotic cell cycle checkpoint
          mitotic spindle assembly checkpoint
          regulation of exit from mitosis
          meiotic nuclear division
          protein transport
          protein localization to kinetochore
          cell division
          protein binding
          centromeric DNA binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1100012353.hg.1 ENST00000200135.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012353.hg.1 ENST00000200135.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012353.hg.1 ENST00000200135.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012353.hg.1 ENST00000200135.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012353.hg.1 ENST00000200135.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012353.hg.1 ENST00000200135.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012353.hg.1 ENST00000200135.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012353.hg.1 ENST00000200135.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012353.hg.1 ENST00000200135.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012353.hg.1 ENST00000200135.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012353.hg.1 ENST00000200135.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012353.hg.1 ENST00000200135.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012353.hg.1 ENST00000200135.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012353.hg.1 ENST00000200135.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012353.hg.1 ENST00000200135.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012353.hg.1 ENST00000200135.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012353.hg.1 ENST00000200135.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012353.hg.1 ENST00000200135.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012353.hg.1 ENST00000200135.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012353.hg.1 ENST00000200135.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012353.hg.1 ENST00000200135.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012353.hg.1 ENST00000200135.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012353.hg.1 ENST00000200135.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012353.hg.1 ENST00000200135.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012353.hg.1 ENST00000535142.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012353.hg.1 ENST00000535142.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012353.hg.1 ENST00000535142.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012353.hg.1 ENST00000535142.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012353.hg.1 ENST00000535142.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012353.hg.1 ENST00000535142.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012353.hg.1 ENST00000535142.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012353.hg.1 ENST00000535142.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012353.hg.1 ENST00000535142.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012353.hg.1 ENST00000535142.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012353.hg.1 ENST00000535142.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012353.hg.1 ENST00000535142.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012353.hg.1 ENST00000535142.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012353.hg.1 ENST00000535142.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012353.hg.1 ENST00000535142.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012353.hg.1 ENST00000535142.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012353.hg.1 ENST00000535142.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012353.hg.1 ENST00000535142.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012353.hg.1 ENST00000535142.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012353.hg.1 ENST00000538209.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012353.hg.1 ENST00000538209.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012353.hg.1 ENST00000538209.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012353.hg.1 ENST00000538209.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012353.hg.1 ENST00000538209.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012353.hg.1 ENST00000538209.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012353.hg.1 ENST00000538209.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012353.hg.1 ENST00000538209.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012353.hg.1 NM_004724.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100012353.hg.1 NM_004724.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100012353.hg.1 NM_004724.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100012353.hg.1 NM_004724.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100012353.hg.1 NM_004724.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100012353.hg.1 NM_004724.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100012353.hg.1 NM_004724.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100012353.hg.1 NM_004724.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100012353.hg.1 NM_004724.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100012353.hg.1 NM_004724.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100012353.hg.1 NM_004724.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100012353.hg.1 NM_004724.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100012353.hg.1 NM_004724.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100012353.hg.1 NM_004724.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100012353.hg.1 NM_004724.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100012353.hg.1 NM_004724.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100012353.hg.1 NM_004724.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100012353.hg.1 NM_004724.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100012353.hg.1 NM_004724.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100012353.hg.1 NM_004724.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100012353.hg.1 NM_004724.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100012353.hg.1 NM_004724.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100012353.hg.1 NM_004724.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11027591.hg.1 ENST00000200135.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11027591.hg.1 NM_004724.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11027591.hg.1 TC11002305.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1100194929.hg.1 ENST00000200135.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1100194929.hg.1 ENST00000535142.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1100194929.hg.1 NM_004724.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100194929.hg.1 ENST00000200135.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1100194929.hg.1 ENST00000535142.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1100194929.hg.1 NM_004724.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100194929.hg.1 TC1100012353.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1100012353.hg.1 ENST00000200135.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100012353.hg.1 ENST00000200135.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100012353.hg.1 ENST00000200135.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100012353.hg.1 ENST00000200135.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100012353.hg.1 ENST00000200135.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100012353.hg.1 ENST00000200135.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100012353.hg.1 ENST00000200135.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100012353.hg.1 ENST00000200135.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100012353.hg.1 ENST00000200135.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100012353.hg.1 ENST00000200135.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100012353.hg.1 ENST00000200135.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100012353.hg.1 ENST00000200135.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100012353.hg.1 ENST00000200135.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100012353.hg.1 ENST00000200135.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100012353.hg.1 ENST00000200135.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100012353.hg.1 ENST00000200135.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100012353.hg.1 ENST00000200135.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100012353.hg.1 ENST00000200135.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100012353.hg.1 ENST00000200135.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100012353.hg.1 ENST00000200135.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100012353.hg.1 ENST00000200135.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100012353.hg.1 ENST00000200135.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100012353.hg.1 ENST00000200135.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100012353.hg.1 ENST00000200135.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100012353.hg.1 ENST00000535142.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100012353.hg.1 ENST00000535142.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100012353.hg.1 ENST00000535142.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100012353.hg.1 ENST00000535142.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100012353.hg.1 ENST00000535142.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100012353.hg.1 ENST00000535142.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100012353.hg.1 ENST00000535142.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100012353.hg.1 ENST00000535142.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100012353.hg.1 ENST00000535142.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100012353.hg.1 ENST00000535142.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100012353.hg.1 ENST00000535142.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100012353.hg.1 ENST00000535142.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100012353.hg.1 ENST00000535142.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100012353.hg.1 ENST00000535142.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100012353.hg.1 ENST00000535142.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100012353.hg.1 ENST00000535142.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100012353.hg.1 ENST00000535142.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100012353.hg.1 ENST00000535142.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100012353.hg.1 ENST00000535142.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100012353.hg.1 ENST00000538209.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100012353.hg.1 ENST00000538209.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100012353.hg.1 ENST00000538209.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100012353.hg.1 ENST00000538209.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100012353.hg.1 ENST00000538209.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100012353.hg.1 ENST00000538209.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100012353.hg.1 ENST00000538209.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100012353.hg.1 ENST00000538209.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100012353.hg.1 NM_004724.3 Refseq
          Affymetrix Human Clariom D Assay TC1100012353.hg.1 NM_004724.3 Refseq
          Affymetrix Human Clariom D Assay TC1100012353.hg.1 NM_004724.3 Refseq
          Affymetrix Human Clariom D Assay TC1100012353.hg.1 NM_004724.3 Refseq
          Affymetrix Human Clariom D Assay TC1100012353.hg.1 NM_004724.3 Refseq
          Affymetrix Human Clariom D Assay TC1100012353.hg.1 NM_004724.3 Refseq
          Affymetrix Human Clariom D Assay TC1100012353.hg.1 NM_004724.3 Refseq
          Affymetrix Human Clariom D Assay TC1100012353.hg.1 NM_004724.3 Refseq
          Affymetrix Human Clariom D Assay TC1100012353.hg.1 NM_004724.3 Refseq
          Affymetrix Human Clariom D Assay TC1100012353.hg.1 NM_004724.3 Refseq
          Affymetrix Human Clariom D Assay TC1100012353.hg.1 NM_004724.3 Refseq
          Affymetrix Human Clariom D Assay TC1100012353.hg.1 NM_004724.3 Refseq
          Affymetrix Human Clariom D Assay TC1100012353.hg.1 NM_004724.3 Refseq
          Affymetrix Human Clariom D Assay TC1100012353.hg.1 NM_004724.3 Refseq
          Affymetrix Human Clariom D Assay TC1100012353.hg.1 NM_004724.3 Refseq
          Affymetrix Human Clariom D Assay TC1100012353.hg.1 NM_004724.3 Refseq
          Affymetrix Human Clariom D Assay TC1100012353.hg.1 NM_004724.3 Refseq
          Affymetrix Human Clariom D Assay TC1100012353.hg.1 NM_004724.3 Refseq
          Affymetrix Human Clariom D Assay TC1100012353.hg.1 NM_004724.3 Refseq
          Affymetrix Human Clariom D Assay TC1100012353.hg.1 NM_004724.3 Refseq
          Affymetrix Human Clariom D Assay TC1100012353.hg.1 NM_004724.3 Refseq
          Affymetrix Human Clariom D Assay TC1100012353.hg.1 NM_004724.3 Refseq
          Affymetrix Human Clariom D Assay TC1100012353.hg.1 NM_004724.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100194930.hg.1 ENST00000200135.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1100194930.hg.1 ENST00000535142.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1100194930.hg.1 ENST00000538209.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100194930.hg.1 NM_004724.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100194930.hg.1 ENST00000200135.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1100194930.hg.1 ENST00000535142.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1100194930.hg.1 ENST00000538209.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100194930.hg.1 NM_004724.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100194930.hg.1 TC1100012353.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline