| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06023095.hg.1 |
ENST00000369816.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06023095.hg.1 |
NM_022726.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06023095.hg.1 |
TC06001902.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012379.hg.1 |
ENST00000369816.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012379.hg.1 |
ENST00000369816.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012379.hg.1 |
ENST00000369816.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012379.hg.1 |
ENST00000369816.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012379.hg.1 |
ENST00000369816.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012379.hg.1 |
ENST00000369816.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012379.hg.1 |
ENST00000369816.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012379.hg.1 |
ENST00000369816.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012379.hg.1 |
ENST00000369816.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012379.hg.1 |
ENST00000369816.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012379.hg.1 |
NM_022726.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012379.hg.1 |
NM_022726.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012379.hg.1 |
NM_022726.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012379.hg.1 |
NM_022726.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012379.hg.1 |
NM_022726.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012379.hg.1 |
NM_022726.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012379.hg.1 |
NM_022726.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012379.hg.1 |
NM_022726.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012379.hg.1 |
NM_022726.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012379.hg.1 |
NM_022726.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012379.hg.1 |
ENST00000369816.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012379.hg.1 |
ENST00000369816.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012379.hg.1 |
ENST00000369816.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012379.hg.1 |
ENST00000369816.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012379.hg.1 |
ENST00000369816.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012379.hg.1 |
ENST00000369816.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012379.hg.1 |
ENST00000369816.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012379.hg.1 |
ENST00000369816.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012379.hg.1 |
ENST00000369816.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012379.hg.1 |
ENST00000369816.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012379.hg.1 |
NM_022726.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012379.hg.1 |
NM_022726.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012379.hg.1 |
NM_022726.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012379.hg.1 |
NM_022726.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012379.hg.1 |
NM_022726.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012379.hg.1 |
NM_022726.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012379.hg.1 |
NM_022726.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012379.hg.1 |
NM_022726.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012379.hg.1 |
NM_022726.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012379.hg.1 |
NM_022726.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600185313.hg.1 |
ENST00000369816.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600185313.hg.1 |
NM_022726.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600185313.hg.1 |
ENST00000369816.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600185313.hg.1 |
NM_022726.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600185313.hg.1 |
TC0600012379.hg.1 |
Clariom D TC |