| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600010864.hg.1 |
ENST00000011619.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600010864.hg.1 |
ENST00000011619.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600010864.hg.1 |
ENST00000011619.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600010864.hg.1 |
ENST00000011619.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600010864.hg.1 |
ENST00000011619.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600010864.hg.1 |
ENST00000011619.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600010864.hg.1 |
ENST00000011619.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600010864.hg.1 |
ENST00000011619.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600010864.hg.1 |
ENST00000011619.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600010864.hg.1 |
ENST00000011619.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600010864.hg.1 |
ENST00000011619.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600010864.hg.1 |
ENST00000011619.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600010864.hg.1 |
ENST00000011619.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600010864.hg.1 |
ENST00000011619.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600010864.hg.1 |
ENST00000011619.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600010864.hg.1 |
ENST00000011619.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600010864.hg.1 |
ENST00000469916.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600010864.hg.1 |
ENST00000469916.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600010864.hg.1 |
ENST00000469916.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600010864.hg.1 |
ENST00000469916.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600010864.hg.1 |
ENST00000469916.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600010864.hg.1 |
NM_005493.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600010864.hg.1 |
NM_005493.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600010864.hg.1 |
NM_005493.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600010864.hg.1 |
NM_005493.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600010864.hg.1 |
NM_005493.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600010864.hg.1 |
NM_005493.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600010864.hg.1 |
NM_005493.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600010864.hg.1 |
NM_005493.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600010864.hg.1 |
NM_005493.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600010864.hg.1 |
NM_005493.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600010864.hg.1 |
NM_005493.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600010864.hg.1 |
NM_005493.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600010864.hg.1 |
NM_005493.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600010864.hg.1 |
NM_005493.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600010864.hg.1 |
NM_005493.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600010864.hg.1 |
NM_005493.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06015381.hg.1 |
ENST00000011619.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06015381.hg.1 |
ENST00000469916.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06015381.hg.1 |
ENST00000539980.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06015381.hg.1 |
NM_005493.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06015381.hg.1 |
TC06001281.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0600010864.hg.1 |
ENST00000011619.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600010864.hg.1 |
ENST00000011619.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600010864.hg.1 |
ENST00000011619.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600010864.hg.1 |
ENST00000011619.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600010864.hg.1 |
ENST00000011619.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600010864.hg.1 |
ENST00000011619.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600010864.hg.1 |
ENST00000011619.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600010864.hg.1 |
ENST00000011619.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600010864.hg.1 |
ENST00000011619.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600010864.hg.1 |
ENST00000011619.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600010864.hg.1 |
ENST00000011619.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600010864.hg.1 |
ENST00000011619.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600010864.hg.1 |
ENST00000011619.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600010864.hg.1 |
ENST00000011619.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600010864.hg.1 |
ENST00000011619.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600010864.hg.1 |
ENST00000011619.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600010864.hg.1 |
ENST00000469916.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600010864.hg.1 |
ENST00000469916.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600010864.hg.1 |
ENST00000469916.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600010864.hg.1 |
ENST00000469916.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600010864.hg.1 |
ENST00000469916.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600010864.hg.1 |
NM_005493.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600010864.hg.1 |
NM_005493.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600010864.hg.1 |
NM_005493.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600010864.hg.1 |
NM_005493.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600010864.hg.1 |
NM_005493.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600010864.hg.1 |
NM_005493.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600010864.hg.1 |
NM_005493.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600010864.hg.1 |
NM_005493.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600010864.hg.1 |
NM_005493.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600010864.hg.1 |
NM_005493.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600010864.hg.1 |
NM_005493.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600010864.hg.1 |
NM_005493.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600010864.hg.1 |
NM_005493.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600010864.hg.1 |
NM_005493.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600010864.hg.1 |
NM_005493.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600010864.hg.1 |
NM_005493.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600173827.hg.1 |
ENST00000011619.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600173827.hg.1 |
ENST00000469916.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600173827.hg.1 |
NM_005493.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600173827.hg.1 |
ENST00000011619.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600173827.hg.1 |
ENST00000469916.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600173827.hg.1 |
NM_005493.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600173827.hg.1 |
TC0600010864.hg.1 |
Clariom D TC |