Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01118635_g1
          See other ANKAR GE Assays ›
          Gene Symbol:
          ANKAR
          Gene Name:
          ankyrin and armadillo repeat containing
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 189674292 - 189763059 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01118635_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ANKAR
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_144708.3 NP_653309.3 14 - 15 3120 109
          XM_011510673.2 XP_011508975.1 14 - 15 3639 109
          XM_011510674.1 XP_011508976.1 14 - 15 3606 109
          XM_011510675.2 XP_011508977.1 13 - 14 3534 109
          XM_011510676.2 XP_011508978.1 14 - 15 3639 109
          XM_011510677.2 XP_011508979.1 14 - 15 3639 109
          XM_011510680.1 XP_011508982.1 14 - 15 3639 109
          XM_011510681.1 XP_011508983.1 14 - 15 3639 109
          XM_011510682.2 XP_011508984.1 14 - 15 3639 109
          XM_011510685.2 XP_011508987.1 14 - 15 3195 109
          XM_011510686.2 XP_011508988.1 14 - 15 3639 109
          XM_011510687.2 XP_011508989.1 13 - 14 3003 109
          XM_011510688.1 XP_011508990.1 14 - 15 3639 109
          XM_011510689.2 XP_011508991.1 14 - 15 3639 109
          XM_011510691.2 XP_011508993.1 10 - 11 1825 109
          XM_011510692.2 XP_011508994.1 12 - 13 2839 109
          XM_011510693.2 XP_011508995.1 11 - 12 2735 109
          XM_011510694.2 XP_011508996.1 10 - 11 2554 109
          XM_017003413.1 XP_016858902.1 14 - 15 3639 109
          XM_017003414.1 XP_016858903.1 14 - 15 3639 109
          XM_017003415.1 XP_016858904.1 9 - 10 2334 109
          XM_017003416.1 XP_016858905.1 11 - 12 2746 109
          XM_017003418.1 XP_016858907.1 9 - 10 2449 109
          XM_017003419.1 XP_016858908.1 9 - 10 2416 109
          XM_017003420.1 XP_016858909.1 9 - 10 2352 109
          GenBank mRNA AJ549812.1 - 13 - 14 2819 109
          AJ605761.1 - 2 - 3 288 109
          AK058144.1 - 4 - 5 702 109
          AK126073.1 - 14 - 15 3120 109
          BC047413.1 - 16 - 17 2545 109

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0200010253.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC02001111.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene ANKAR

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000313581.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000313581.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000313581.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000313581.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000313581.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000313581.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000313581.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000313581.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000313581.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000313581.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000313581.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000313581.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000313581.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000313581.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000313581.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000313581.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000313581.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000313581.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000313581.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000313581.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000313581.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000313581.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000313581.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000313581.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000313581.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000313581.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000313581.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000313581.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000313581.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000411317.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000433782.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000433782.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000433782.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000433782.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000433782.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000433782.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000433782.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000433782.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000433782.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000433782.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000433782.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000433782.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000433782.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000433782.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000433782.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000433782.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000433782.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000433782.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000433782.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000433782.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000433782.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000433782.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000433782.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000433782.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000433782.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000433782.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000441800.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000441800.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000441800.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000441800.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000441800.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000441800.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000441800.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000441800.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000441800.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000441800.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000441800.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000441800.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000441800.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000441800.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000441800.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000441800.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000441800.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000441800.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000441800.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000441800.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000441800.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000441800.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000441800.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000441800.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000441800.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000461516.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000461516.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000461516.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000464687.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000464687.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000464687.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000464687.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000467927.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000467927.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000467927.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000467927.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000474303.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000474303.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000474303.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000474303.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000474303.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000476208.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000476208.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000476208.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000476208.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000520309.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000520309.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000520309.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000520309.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000520309.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000520309.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000520309.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000520309.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000520309.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000520309.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000520309.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000520309.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000520309.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000520309.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000520309.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000520309.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000520309.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000520309.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000520309.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000520309.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000520309.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000520309.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000520309.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000520309.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000520309.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000520309.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000520309.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000520309.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 ENST00000520309.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 NM_144708.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 NM_144708.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 NM_144708.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 NM_144708.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 NM_144708.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 NM_144708.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 NM_144708.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 NM_144708.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 NM_144708.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 NM_144708.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 NM_144708.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 NM_144708.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 NM_144708.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 NM_144708.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 NM_144708.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 NM_144708.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 NM_144708.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 NM_144708.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 NM_144708.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 NM_144708.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 NM_144708.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 NM_144708.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 NM_144708.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 NM_144708.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 NM_144708.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 NM_144708.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 NM_144708.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 NM_144708.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200010253.hg.1 NM_144708.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-pgj68:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline