Search
Search

SRSF3| Gene Symbol: SRSF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003017.4 | NP_003008.1 | 4 - 5 | 549 | 81 | |
| GenBank mRNA | AB451229.1 | - | 3 - 4 | 378 | 81 | |
| AB451352.1 | - | 3 - 4 | 378 | 81 | ||
| AF107405.1 | - | 4 - 5 | 473 | 81 | ||
| AK312553.1 | - | 4 - 5 | 500 | 81 | ||
| BC000914.1 | - | 4 - 5 | 483 | 3049167 | 81 | |
| BC069018.1 | - | 4 - 5 | 479 | 3507371 | 81 | |
| BT007017.1 | - | 3 - 4 | 378 | 81 | ||
| L10838.1 | - | 3 - 4 | 378 | 81 | ||
Set Membership: |
|