Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01124271_g1
          See other IL13 GE Assays ›
          Gene Symbol:
          IL13
          Gene Name:
          interleukin 13
          Gene Aliases:
          IL-13, P600
          Chromosome Location:
          Chr.5: 132658173 - 132661109 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01124271_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: IL13
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_002188.2 NP_002179.2 2 - 3 238 70
          GenBank mRNA BC096138.3 - 1 - 2 1295 40002676 70
          BC096139.1 - 2 - 3 238 40002680 70
          BC096140.3 - 1 - 2 1293 40002684 70
          BC096141.1 - 2 - 3 238 40002686 70
          L06801.1 - 2 - 3 226 70
          X69079.1 - 2 - 3 238 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0500008642.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC05000640.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the BC096138.3,BC096140.3 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Mammalian Gene Collection Detect Genomic DNA Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene IL13

          Gene Ontology Categories:

          Function(s) Process(es)

          microglial cell activation
          positive regulation of immunoglobulin production
          movement of cell or subcellular component
          inflammatory response
          immune response
          signal transduction
          cell-cell signaling
          regulation of proton transport
          positive regulation of B cell proliferation
          response to lipopolysaccharide
          positive regulation of connective tissue growth factor production
          negative regulation of NAD(P)H oxidase activity
          response to nicotine
          positive regulation of tyrosine phosphorylation of Stat6 protein
          positive regulation of macrophage activation
          positive regulation of mast cell degranulation
          response to ethanol
          positive regulation of smooth muscle cell proliferation
          positive regulation of protein secretion
          positive regulation of release of sequestered calcium ion into cytosol
          cellular response to mechanical stimulus
          cellular response to cytokine stimulus
          negative regulation of transforming growth factor beta production
          negative regulation of neuron death
          negative regulation of lung ciliated cell differentiation
          positive regulation of lung goblet cell differentiation
          negative regulation of complement-dependent cytotoxicity
          positive regulation of pancreatic stellate cell proliferation
          negative regulation of endothelial cell apoptotic process
          cytokine activity
          interleukin-13 receptor binding
          protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0500008642.hg.1 ENST00000304506.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008642.hg.1 ENST00000304506.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008642.hg.1 ENST00000304506.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008642.hg.1 ENST00000304506.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008642.hg.1 ENST00000304506.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008642.hg.1 ENST00000304506.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008642.hg.1 ENST00000304506.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008642.hg.1 ENST00000304506.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008642.hg.1 ENST00000304506.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008642.hg.1 ENST00000304506.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008642.hg.1 ENST00000459878.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008642.hg.1 ENST00000459878.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008642.hg.1 ENST00000459878.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008642.hg.1 ENST00000459878.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008642.hg.1 ENST00000459878.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008642.hg.1 ENST00000459878.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008642.hg.1 ENST00000459878.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008642.hg.1 ENST00000459878.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008642.hg.1 ENST00000462480.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008642.hg.1 ENST00000462480.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008642.hg.1 ENST00000462480.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008642.hg.1 ENST00000462480.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008642.hg.1 ENST00000462480.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008642.hg.1 ENST00000462480.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008642.hg.1 ENST00000462480.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008642.hg.1 ENST00000468334.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008642.hg.1 ENST00000468334.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008642.hg.1 ENST00000468334.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008642.hg.1 ENST00000468334.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008642.hg.1 ENST00000468334.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008642.hg.1 ENST00000468334.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008642.hg.1 ENST00000468334.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008642.hg.1 ENST00000468334.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008642.hg.1 ENST00000487267.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008642.hg.1 ENST00000487267.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008642.hg.1 ENST00000487267.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008642.hg.1 ENST00000487267.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008642.hg.1 ENST00000487267.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008642.hg.1 ENST00000487267.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008642.hg.1 ENST00000487267.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008642.hg.1 ENST00000487267.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008642.hg.1 ENST00000487267.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008642.hg.1 ENST00000487267.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008642.hg.1 ENST00000617259.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008642.hg.1 ENST00000617259.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008642.hg.1 ENST00000617259.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008642.hg.1 ENST00000617259.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008642.hg.1 ENST00000617259.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008642.hg.1 ENST00000617259.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008642.hg.1 NM_002188.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500008642.hg.1 NM_002188.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500008642.hg.1 NM_002188.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500008642.hg.1 NM_002188.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500008642.hg.1 NM_002188.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500008642.hg.1 NM_002188.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500008642.hg.1 NM_002188.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500008642.hg.1 NM_002188.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500008642.hg.1 NM_002188.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500008642.hg.1 NM_002188.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05008877.hg.1 ENST00000304506.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05008877.hg.1 ENST00000468334.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05008877.hg.1 NM_002188.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05008877.hg.1 TC05000640.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0500159296.hg.1 ENST00000304506.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0500159296.hg.1 ENST00000459878.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500159296.hg.1 ENST00000462480.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500159296.hg.1 ENST00000468334.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500159296.hg.1 ENST00000487267.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500159296.hg.1 ENST00000617259.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0500159296.hg.1 NM_002188.2 Refseq
          Affymetrix Human Clariom D Assay PSR0500159296.hg.1 ENST00000304506.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0500159296.hg.1 ENST00000459878.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500159296.hg.1 ENST00000462480.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500159296.hg.1 ENST00000468334.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500159296.hg.1 ENST00000487267.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500159296.hg.1 ENST00000617259.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0500159296.hg.1 NM_002188.2 Refseq
          Affymetrix Human Clariom D Assay PSR0500159296.hg.1 TC0500008642.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0500159294.hg.1 ENST00000462480.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500159294.hg.1 ENST00000462480.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500159294.hg.1 TC0500008642.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0500008642.hg.1 ENST00000304506.7 Ensembl
          Affymetrix Human Clariom D Assay TC0500008642.hg.1 ENST00000304506.7 Ensembl
          Affymetrix Human Clariom D Assay TC0500008642.hg.1 ENST00000304506.7 Ensembl
          Affymetrix Human Clariom D Assay TC0500008642.hg.1 ENST00000304506.7 Ensembl
          Affymetrix Human Clariom D Assay TC0500008642.hg.1 ENST00000304506.7 Ensembl
          Affymetrix Human Clariom D Assay TC0500008642.hg.1 ENST00000304506.7 Ensembl
          Affymetrix Human Clariom D Assay TC0500008642.hg.1 ENST00000304506.7 Ensembl
          Affymetrix Human Clariom D Assay TC0500008642.hg.1 ENST00000304506.7 Ensembl
          Affymetrix Human Clariom D Assay TC0500008642.hg.1 ENST00000304506.7 Ensembl
          Affymetrix Human Clariom D Assay TC0500008642.hg.1 ENST00000304506.7 Ensembl
          Affymetrix Human Clariom D Assay TC0500008642.hg.1 ENST00000459878.5 Ensembl
          Affymetrix Human Clariom D Assay TC0500008642.hg.1 ENST00000459878.5 Ensembl
          Affymetrix Human Clariom D Assay TC0500008642.hg.1 ENST00000459878.5 Ensembl
          Affymetrix Human Clariom D Assay TC0500008642.hg.1 ENST00000459878.5 Ensembl
          Affymetrix Human Clariom D Assay TC0500008642.hg.1 ENST00000459878.5 Ensembl
          Affymetrix Human Clariom D Assay TC0500008642.hg.1 ENST00000459878.5 Ensembl
          Affymetrix Human Clariom D Assay TC0500008642.hg.1 ENST00000459878.5 Ensembl
          Affymetrix Human Clariom D Assay TC0500008642.hg.1 ENST00000459878.5 Ensembl
          Affymetrix Human Clariom D Assay TC0500008642.hg.1 ENST00000462480.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500008642.hg.1 ENST00000462480.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500008642.hg.1 ENST00000462480.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500008642.hg.1 ENST00000462480.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500008642.hg.1 ENST00000462480.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500008642.hg.1 ENST00000462480.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500008642.hg.1 ENST00000462480.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500008642.hg.1 ENST00000468334.5 Ensembl
          Affymetrix Human Clariom D Assay TC0500008642.hg.1 ENST00000468334.5 Ensembl
          Affymetrix Human Clariom D Assay TC0500008642.hg.1 ENST00000468334.5 Ensembl
          Affymetrix Human Clariom D Assay TC0500008642.hg.1 ENST00000468334.5 Ensembl
          Affymetrix Human Clariom D Assay TC0500008642.hg.1 ENST00000468334.5 Ensembl
          Affymetrix Human Clariom D Assay TC0500008642.hg.1 ENST00000468334.5 Ensembl
          Affymetrix Human Clariom D Assay TC0500008642.hg.1 ENST00000468334.5 Ensembl
          Affymetrix Human Clariom D Assay TC0500008642.hg.1 ENST00000468334.5 Ensembl
          Affymetrix Human Clariom D Assay TC0500008642.hg.1 ENST00000487267.5 Ensembl
          Affymetrix Human Clariom D Assay TC0500008642.hg.1 ENST00000487267.5 Ensembl
          Affymetrix Human Clariom D Assay TC0500008642.hg.1 ENST00000487267.5 Ensembl
          Affymetrix Human Clariom D Assay TC0500008642.hg.1 ENST00000487267.5 Ensembl
          Affymetrix Human Clariom D Assay TC0500008642.hg.1 ENST00000487267.5 Ensembl
          Affymetrix Human Clariom D Assay TC0500008642.hg.1 ENST00000487267.5 Ensembl
          Affymetrix Human Clariom D Assay TC0500008642.hg.1 ENST00000487267.5 Ensembl
          Affymetrix Human Clariom D Assay TC0500008642.hg.1 ENST00000487267.5 Ensembl
          Affymetrix Human Clariom D Assay TC0500008642.hg.1 ENST00000487267.5 Ensembl
          Affymetrix Human Clariom D Assay TC0500008642.hg.1 ENST00000487267.5 Ensembl
          Affymetrix Human Clariom D Assay TC0500008642.hg.1 ENST00000617259.2 Ensembl
          Affymetrix Human Clariom D Assay TC0500008642.hg.1 ENST00000617259.2 Ensembl
          Affymetrix Human Clariom D Assay TC0500008642.hg.1 ENST00000617259.2 Ensembl
          Affymetrix Human Clariom D Assay TC0500008642.hg.1 ENST00000617259.2 Ensembl
          Affymetrix Human Clariom D Assay TC0500008642.hg.1 ENST00000617259.2 Ensembl
          Affymetrix Human Clariom D Assay TC0500008642.hg.1 ENST00000617259.2 Ensembl
          Affymetrix Human Clariom D Assay TC0500008642.hg.1 NM_002188.2 Refseq
          Affymetrix Human Clariom D Assay TC0500008642.hg.1 NM_002188.2 Refseq
          Affymetrix Human Clariom D Assay TC0500008642.hg.1 NM_002188.2 Refseq
          Affymetrix Human Clariom D Assay TC0500008642.hg.1 NM_002188.2 Refseq
          Affymetrix Human Clariom D Assay TC0500008642.hg.1 NM_002188.2 Refseq
          Affymetrix Human Clariom D Assay TC0500008642.hg.1 NM_002188.2 Refseq
          Affymetrix Human Clariom D Assay TC0500008642.hg.1 NM_002188.2 Refseq
          Affymetrix Human Clariom D Assay TC0500008642.hg.1 NM_002188.2 Refseq
          Affymetrix Human Clariom D Assay TC0500008642.hg.1 NM_002188.2 Refseq
          Affymetrix Human Clariom D Assay TC0500008642.hg.1 NM_002188.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline