Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01127080_m1
          See other LEO1 GE Assays ›
          Gene Symbol:
          LEO1
          Gene Name:
          LEO1 homolog, Paf1/RNA polymerase II complex component
          Gene Aliases:
          RDL
          Chromosome Location:
          Chr.15: 51938024 - 51971801 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01127080_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LEO1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001286430.1 NP_001273359.1 9 - 10 1774 151
          NM_138792.3 NP_620147.1 11 - 12 1954 151
          GenBank mRNA AK055762.1 - 9 - 10 1750 151
          AK297972.1 - 12 - 13 1864 151
          AY302186.1 - 11 - 12 1938 151
          BC018147.2 - 11 - 12 1914 3847809 151

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1500009446.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC15001449.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Homo sapiens 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene LEO1

          Panther Classification:

          Molecular Function -

          DNA-directed RNA polymerase RNA metabolism protein nucleic acid metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          endodermal cell fate commitment
          transcription elongation from RNA polymerase II promoter
          mRNA polyadenylation
          histone monoubiquitination
          Wnt signaling pathway
          stem cell population maintenance
          positive regulation of mRNA 3'-end processing
          positive regulation of transcription elongation from RNA polymerase II promoter
          histone H2B ubiquitination
          negative regulation of myeloid cell differentiation
          positive regulation of transcription from RNA polymerase II promoter
          beta-catenin-TCF complex assembly
          protein binding
          RNA polymerase II C-terminal domain phosphoserine binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR15013368.hg.1 ENST00000299601.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15013368.hg.1 ENST00000315141.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15013368.hg.1 ENST00000538386.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15013368.hg.1 NM_138792.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR15013368.hg.1 TC15001449.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1500009446.hg.1 ENST00000299601.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009446.hg.1 ENST00000299601.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009446.hg.1 ENST00000299601.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009446.hg.1 ENST00000299601.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009446.hg.1 ENST00000299601.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009446.hg.1 ENST00000299601.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009446.hg.1 ENST00000299601.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009446.hg.1 ENST00000299601.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009446.hg.1 ENST00000299601.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009446.hg.1 ENST00000299601.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009446.hg.1 ENST00000299601.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009446.hg.1 ENST00000299601.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009446.hg.1 ENST00000299601.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009446.hg.1 ENST00000299601.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009446.hg.1 ENST00000299601.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009446.hg.1 ENST00000315141.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009446.hg.1 ENST00000315141.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009446.hg.1 ENST00000315141.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009446.hg.1 ENST00000315141.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009446.hg.1 ENST00000315141.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009446.hg.1 ENST00000315141.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009446.hg.1 ENST00000315141.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009446.hg.1 ENST00000315141.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009446.hg.1 ENST00000315141.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009446.hg.1 ENST00000315141.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009446.hg.1 ENST00000315141.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009446.hg.1 ENST00000315141.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009446.hg.1 ENST00000558949.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009446.hg.1 ENST00000558949.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009446.hg.1 ENST00000558949.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009446.hg.1 NM_001286430.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009446.hg.1 NM_001286430.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009446.hg.1 NM_001286430.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009446.hg.1 NM_001286430.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009446.hg.1 NM_001286430.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009446.hg.1 NM_001286430.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009446.hg.1 NM_001286430.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009446.hg.1 NM_001286430.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009446.hg.1 NM_001286430.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009446.hg.1 NM_001286430.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009446.hg.1 NM_001286430.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009446.hg.1 NM_001286430.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009446.hg.1 NM_001286430.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009446.hg.1 NM_138792.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009446.hg.1 NM_138792.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009446.hg.1 NM_138792.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009446.hg.1 NM_138792.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009446.hg.1 NM_138792.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009446.hg.1 NM_138792.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009446.hg.1 NM_138792.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009446.hg.1 NM_138792.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009446.hg.1 NM_138792.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009446.hg.1 NM_138792.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009446.hg.1 NM_138792.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009446.hg.1 NM_138792.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009446.hg.1 NM_138792.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009446.hg.1 NM_138792.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009446.hg.1 NM_138792.3 Refseq
          Affymetrix Human Clariom D Assay TC1500009446.hg.1 ENST00000299601.9 Ensembl
          Affymetrix Human Clariom D Assay TC1500009446.hg.1 ENST00000299601.9 Ensembl
          Affymetrix Human Clariom D Assay TC1500009446.hg.1 ENST00000299601.9 Ensembl
          Affymetrix Human Clariom D Assay TC1500009446.hg.1 ENST00000299601.9 Ensembl
          Affymetrix Human Clariom D Assay TC1500009446.hg.1 ENST00000299601.9 Ensembl
          Affymetrix Human Clariom D Assay TC1500009446.hg.1 ENST00000299601.9 Ensembl
          Affymetrix Human Clariom D Assay TC1500009446.hg.1 ENST00000299601.9 Ensembl
          Affymetrix Human Clariom D Assay TC1500009446.hg.1 ENST00000299601.9 Ensembl
          Affymetrix Human Clariom D Assay TC1500009446.hg.1 ENST00000299601.9 Ensembl
          Affymetrix Human Clariom D Assay TC1500009446.hg.1 ENST00000299601.9 Ensembl
          Affymetrix Human Clariom D Assay TC1500009446.hg.1 ENST00000299601.9 Ensembl
          Affymetrix Human Clariom D Assay TC1500009446.hg.1 ENST00000299601.9 Ensembl
          Affymetrix Human Clariom D Assay TC1500009446.hg.1 ENST00000299601.9 Ensembl
          Affymetrix Human Clariom D Assay TC1500009446.hg.1 ENST00000299601.9 Ensembl
          Affymetrix Human Clariom D Assay TC1500009446.hg.1 ENST00000299601.9 Ensembl
          Affymetrix Human Clariom D Assay TC1500009446.hg.1 ENST00000315141.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500009446.hg.1 ENST00000315141.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500009446.hg.1 ENST00000315141.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500009446.hg.1 ENST00000315141.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500009446.hg.1 ENST00000315141.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500009446.hg.1 ENST00000315141.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500009446.hg.1 ENST00000315141.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500009446.hg.1 ENST00000315141.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500009446.hg.1 ENST00000315141.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500009446.hg.1 ENST00000315141.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500009446.hg.1 ENST00000315141.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500009446.hg.1 ENST00000315141.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500009446.hg.1 ENST00000558949.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500009446.hg.1 ENST00000558949.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500009446.hg.1 ENST00000558949.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500009446.hg.1 NM_001286430.1 Refseq
          Affymetrix Human Clariom D Assay TC1500009446.hg.1 NM_001286430.1 Refseq
          Affymetrix Human Clariom D Assay TC1500009446.hg.1 NM_001286430.1 Refseq
          Affymetrix Human Clariom D Assay TC1500009446.hg.1 NM_001286430.1 Refseq
          Affymetrix Human Clariom D Assay TC1500009446.hg.1 NM_001286430.1 Refseq
          Affymetrix Human Clariom D Assay TC1500009446.hg.1 NM_001286430.1 Refseq
          Affymetrix Human Clariom D Assay TC1500009446.hg.1 NM_001286430.1 Refseq
          Affymetrix Human Clariom D Assay TC1500009446.hg.1 NM_001286430.1 Refseq
          Affymetrix Human Clariom D Assay TC1500009446.hg.1 NM_001286430.1 Refseq
          Affymetrix Human Clariom D Assay TC1500009446.hg.1 NM_001286430.1 Refseq
          Affymetrix Human Clariom D Assay TC1500009446.hg.1 NM_001286430.1 Refseq
          Affymetrix Human Clariom D Assay TC1500009446.hg.1 NM_001286430.1 Refseq
          Affymetrix Human Clariom D Assay TC1500009446.hg.1 NM_001286430.1 Refseq
          Affymetrix Human Clariom D Assay TC1500009446.hg.1 NM_138792.3 Refseq
          Affymetrix Human Clariom D Assay TC1500009446.hg.1 NM_138792.3 Refseq
          Affymetrix Human Clariom D Assay TC1500009446.hg.1 NM_138792.3 Refseq
          Affymetrix Human Clariom D Assay TC1500009446.hg.1 NM_138792.3 Refseq
          Affymetrix Human Clariom D Assay TC1500009446.hg.1 NM_138792.3 Refseq
          Affymetrix Human Clariom D Assay TC1500009446.hg.1 NM_138792.3 Refseq
          Affymetrix Human Clariom D Assay TC1500009446.hg.1 NM_138792.3 Refseq
          Affymetrix Human Clariom D Assay TC1500009446.hg.1 NM_138792.3 Refseq
          Affymetrix Human Clariom D Assay TC1500009446.hg.1 NM_138792.3 Refseq
          Affymetrix Human Clariom D Assay TC1500009446.hg.1 NM_138792.3 Refseq
          Affymetrix Human Clariom D Assay TC1500009446.hg.1 NM_138792.3 Refseq
          Affymetrix Human Clariom D Assay TC1500009446.hg.1 NM_138792.3 Refseq
          Affymetrix Human Clariom D Assay TC1500009446.hg.1 NM_138792.3 Refseq
          Affymetrix Human Clariom D Assay TC1500009446.hg.1 NM_138792.3 Refseq
          Affymetrix Human Clariom D Assay TC1500009446.hg.1 NM_138792.3 Refseq
          Affymetrix Human Clariom D Assay PSR1500169394.hg.1 ENST00000299601.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1500169394.hg.1 ENST00000315141.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500169394.hg.1 NM_001286430.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500169394.hg.1 NM_138792.3 Refseq
          Affymetrix Human Clariom D Assay PSR1500169394.hg.1 TC1500009446.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1500169394.hg.1 ENST00000299601.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1500169394.hg.1 ENST00000315141.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500169394.hg.1 NM_001286430.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500169394.hg.1 NM_138792.3 Refseq
          Affymetrix Human Clariom D Assay PSR1500169395.hg.1 ENST00000299601.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1500169395.hg.1 ENST00000315141.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500169395.hg.1 NM_001286430.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500169395.hg.1 NM_138792.3 Refseq
          Affymetrix Human Clariom D Assay PSR1500169395.hg.1 TC1500009446.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1500169395.hg.1 ENST00000299601.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1500169395.hg.1 ENST00000315141.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500169395.hg.1 NM_001286430.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500169395.hg.1 NM_138792.3 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 212199 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline