Search
Search

GRIN2B| Gene Symbol: GRIN2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005253351.3 | XP_005253408.1 | 4 | 10225 | 92 | |
| XM_011520628.2 | XP_011518930.1 | 14 | 13291 | 92 | ||
| XM_011520629.2 | XP_011518931.1 | 15 | 13098 | 92 | ||
| XM_017019219.1 | XP_016874708.1 | 14 | 12846 | 92 | ||
Set Membership: |
|