| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19002168.hg.1 |
ENST00000512667.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19002168.hg.1 |
NR_002931.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19002168.hg.1 |
TC19000137.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011659.hg.1 |
ENST00000596555.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011659.hg.1 |
ENST00000596555.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011659.hg.1 |
ENST00000596555.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011659.hg.1 |
ENST00000596555.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011659.hg.1 |
ENST00000596555.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011659.hg.1 |
ENST00000596555.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011659.hg.1 |
ENST00000596555.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011659.hg.1 |
ENST00000596555.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011659.hg.1 |
ENST00000596555.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011659.hg.1 |
ENST00000596555.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011659.hg.1 |
NR_002931.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011659.hg.1 |
NR_002931.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011659.hg.1 |
NR_002931.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011659.hg.1 |
NR_002931.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011659.hg.1 |
NR_002931.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011659.hg.1 |
NR_002931.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011659.hg.1 |
NR_002931.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011659.hg.1 |
NR_002931.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011659.hg.1 |
NR_002931.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011659.hg.1 |
NR_002931.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011659.hg.1 |
NR_002931.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011659.hg.1 |
NR_002931.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011659.hg.1 |
NR_002931.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19002166.hg.1 |
ENST00000512667.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19002166.hg.1 |
NR_002931.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19002166.hg.1 |
TC19000137.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC1900011659.hg.1 |
ENST00000596555.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011659.hg.1 |
ENST00000596555.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011659.hg.1 |
ENST00000596555.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011659.hg.1 |
ENST00000596555.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011659.hg.1 |
ENST00000596555.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011659.hg.1 |
ENST00000596555.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011659.hg.1 |
ENST00000596555.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011659.hg.1 |
ENST00000596555.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011659.hg.1 |
ENST00000596555.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011659.hg.1 |
ENST00000596555.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011659.hg.1 |
NR_002931.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011659.hg.1 |
NR_002931.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011659.hg.1 |
NR_002931.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011659.hg.1 |
NR_002931.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011659.hg.1 |
NR_002931.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011659.hg.1 |
NR_002931.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011659.hg.1 |
NR_002931.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011659.hg.1 |
NR_002931.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011659.hg.1 |
NR_002931.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011659.hg.1 |
NR_002931.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011659.hg.1 |
NR_002931.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011659.hg.1 |
NR_002931.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011659.hg.1 |
NR_002931.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900199324.hg.1 |
ENST00000596555.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900199324.hg.1 |
NR_002931.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900199324.hg.1 |
TC1900011659.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900199324.hg.1 |
ENST00000596555.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900199324.hg.1 |
NR_002931.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900199321.hg.1 |
ENST00000596555.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900199321.hg.1 |
NR_002931.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900199321.hg.1 |
TC1900011659.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900199321.hg.1 |
ENST00000596555.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900199321.hg.1 |
NR_002931.2 |
Refseq |