Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01370906_m1
          See other CECR5-AS1 GE Assays ›
          Gene Symbol:
          CECR5-AS1
          Gene Name:
          CECR5 antisense RNA 1
          Gene Aliases:
          CECR4, NCRNA00017
          Chromosome Location:
          Chr.22: 17159389 - 17165445 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01370906_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CECR5-AS1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_024483.1 1 - 2 450 68
          GenBank mRNA AF307448.1 - 1 - 2 444 68
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA W35386.1 - Chr. - - [-] on Build 68
          GenBankmRNA AA505576.1 - 1 - 2 Chr. 22 - 17159599 - 17165447 [-] on Build GRCh38 68

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream uncharacterized LOC100996342 LOC100996342 -27.3kb Sense Chr. 22 - 17121595 - 17132104 on NCBI Build GRCh38
          Downstream cat eye syndrome chromosome region,
          candidate 1
          CECR1 ADA2,
          ADGF,
          IDGFL,
          PAN,
          SNEDS
          13.3kb Anti-sense Chr. 22 - 17178790 - 17221854 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC2200006507.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC22000021.hg.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Homo sapiens 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_32_P131143 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_32_P131143 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 33697 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 23863 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_32_P131143 - -
          SurePrint G3 Human GE Microarray G4851A A_32_P131143 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11423190 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32091386 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11423198 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11423202 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11423201 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32091392 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11423160 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32091377 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11423205 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11423186 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11423164 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11423194 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32091396 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32091389 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11423162 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11423206 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32091359 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32091397 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11423193 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32091353 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11423195 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11423204 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11423185 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11423184 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11423203 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32091384 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11423197 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11423188 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11423182 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11423166 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32091353 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11423188 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11423195 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11423204 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11423182 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32091396 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11423201 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11423193 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32091389 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32091359 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11423162 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11423198 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32091377 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11423184 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11423203 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11423186 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11423206 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11423194 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11423166 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32091392 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11423202 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32091384 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11423185 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32091386 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32091397 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11423160 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11423190 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11423205 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11423197 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11423164 - -
          Whole Human Genome Oligo Microarray G4112A A_32_P131143 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_32_P131143 - -
          Agilent Human 1B Microarray G4111A A_32_P131139 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17251350 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11423206 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11423191 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17251345 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11423168 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11423203 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11423167 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11423162 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11423190 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11423188 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11423185 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11423172 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11423169 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11423171 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11423166 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11423199 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17251347 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11423197 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11423184 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11423193 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11423204 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11423194 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11423173 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11423201 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17251335 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11423164 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11423182 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11423180 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11423177 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8071085 - -
          Affymetrix Human Gene 1.1 ST Array 8071084 - -
          Affymetrix HT HG-U133+ PM Array Plate 1559920_PM_a_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR22000100.hg.1 ENST00000431923.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22000100.hg.1 NR_024483.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR22000100.hg.1 TC22000021.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC2200006507.hg.1 ENST00000329743.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200006507.hg.1 ENST00000329743.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200006507.hg.1 ENST00000329743.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200006507.hg.1 ENST00000329743.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200006507.hg.1 ENST00000431923.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200006507.hg.1 ENST00000431923.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200006507.hg.1 ENST00000431923.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200006507.hg.1 ENST00000431923.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200006507.hg.1 ENST00000431923.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200006507.hg.1 ENST00000431923.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200006507.hg.1 NR_024482.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200006507.hg.1 NR_024482.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200006507.hg.1 NR_024482.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200006507.hg.1 NR_024482.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200006507.hg.1 NR_024483.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200006507.hg.1 NR_024483.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200006507.hg.1 NR_024483.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200006507.hg.1 NR_024483.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200006507.hg.1 NR_024483.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200006507.hg.1 NR_024483.1 Refseq
          Affymetrix Human Clariom D Assay PSR2200144471.hg.1 ENST00000329743.3 Ensembl
          Affymetrix Human Clariom D Assay PSR2200144471.hg.1 ENST00000431923.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200144471.hg.1 NR_024482.1 Refseq
          Affymetrix Human Clariom D Assay PSR2200144471.hg.1 NR_024483.1 Refseq
          Affymetrix Human Clariom D Assay PSR2200144471.hg.1 TC2200006507.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2200144471.hg.1 ENST00000329743.3 Ensembl
          Affymetrix Human Clariom D Assay PSR2200144471.hg.1 ENST00000431923.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200144471.hg.1 NR_024482.1 Refseq
          Affymetrix Human Clariom D Assay PSR2200144471.hg.1 NR_024483.1 Refseq
          Affymetrix Human Clariom D Assay TC2200006507.hg.1 ENST00000329743.3 Ensembl
          Affymetrix Human Clariom D Assay TC2200006507.hg.1 ENST00000329743.3 Ensembl
          Affymetrix Human Clariom D Assay TC2200006507.hg.1 ENST00000329743.3 Ensembl
          Affymetrix Human Clariom D Assay TC2200006507.hg.1 ENST00000329743.3 Ensembl
          Affymetrix Human Clariom D Assay TC2200006507.hg.1 ENST00000431923.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200006507.hg.1 ENST00000431923.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200006507.hg.1 ENST00000431923.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200006507.hg.1 ENST00000431923.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200006507.hg.1 ENST00000431923.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200006507.hg.1 ENST00000431923.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200006507.hg.1 NR_024482.1 Refseq
          Affymetrix Human Clariom D Assay TC2200006507.hg.1 NR_024482.1 Refseq
          Affymetrix Human Clariom D Assay TC2200006507.hg.1 NR_024482.1 Refseq
          Affymetrix Human Clariom D Assay TC2200006507.hg.1 NR_024482.1 Refseq
          Affymetrix Human Clariom D Assay TC2200006507.hg.1 NR_024483.1 Refseq
          Affymetrix Human Clariom D Assay TC2200006507.hg.1 NR_024483.1 Refseq
          Affymetrix Human Clariom D Assay TC2200006507.hg.1 NR_024483.1 Refseq
          Affymetrix Human Clariom D Assay TC2200006507.hg.1 NR_024483.1 Refseq
          Affymetrix Human Clariom D Assay TC2200006507.hg.1 NR_024483.1 Refseq
          Affymetrix Human Clariom D Assay TC2200006507.hg.1 NR_024483.1 Refseq
          Affymetrix Human Clariom D Assay PSR2200144468.hg.1 ENST00000431923.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200144468.hg.1 NR_024483.1 Refseq
          Affymetrix Human Clariom D Assay PSR2200144468.hg.1 TC2200006507.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2200144468.hg.1 ENST00000431923.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200144468.hg.1 NR_024483.1 Refseq
          Affymetrix Human Clariom D Assay PSR2200144466.hg.1 ENST00000431923.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200144466.hg.1 NR_024483.1 Refseq
          Affymetrix Human Clariom D Assay PSR2200144466.hg.1 TC2200006507.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2200144466.hg.1 ENST00000431923.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200144466.hg.1 NR_024483.1 Refseq
          Human Genome U133 Plus 2.0 Array 1559920_a_at - -
          Human Genome U133 Plus 2.0 Array 1559920_PM_a_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559920_a_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559920_a_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559920_a_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559920_a_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559920_a_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559920_a_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559920_a_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559920_a_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559920_a_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559920_a_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559920_a_at7 - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A020043 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A007313 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A007313 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A020043 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 135364 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3177532 NR_024482.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3177532 NR_024483.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3177532 AF307448.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3177532 BI962745.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3234570 NR_024483.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3177532 NR_024482.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3177532 NR_024483.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3177532 AF307448.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3177532 BI962745.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3234570 NR_024483.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3177532 NR_024482.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3177532 NR_024483.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3177532 AF307448.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3177532 BI962745.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3234570 NR_024483.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3234570 NR_024483.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3177532 NR_024482.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3177532 NR_024483.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3177532 AF307448.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3177532 BI962745.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1870365 - -
          Illumina Sentrix Human-6 v2 Expression BeadChip 7200390 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_122668 - -
          Illumina HumanWG-6_V2_0_R2 ILMN_1870365 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline