Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01377067_m1
          See other PATE3 GE Assays ›
          Gene Symbol:
          PATE3
          Gene Name:
          prostate and testis expressed 3
          Gene Aliases:
          HEL-127, PATE-DJ
          Chromosome Location:
          Chr.11: 125785996 - 125791600 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01377067_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PATE3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001129883.3 NP_001123355.3 2 - 3 215 106
          XM_017017054.1 XP_016872543.1 1 - 2 3515 106
          GenBank mRNA EU703625.1 - 2 - 3 174 106

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1100009427.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC11001162.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Recommended Assay Homo sapiens 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human CGH Microarray 4x180K 74332 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 700370 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 58063 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3420904 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_1799 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_1800 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 12131 - -
          Agilent Human Genome CGH + SNP Microarray A_18_P19067369 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P02540834 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P02540830 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3420904 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3420904 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08097619 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28525357 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28525356 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08097589 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28525365 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28525352 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28525370 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28525353 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28525338 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08097595 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16513398 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08097606 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28525341 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28525348 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08097616 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08097594 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08097613 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28525346 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08097617 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08097596 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28525342 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08097620 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28525363 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28525354 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28525340 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28525351 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08097611 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28525347 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28525352 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08097617 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28525370 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08097616 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16513398 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08097606 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08097589 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28525341 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08097611 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28525357 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28525365 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08097620 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08097596 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28525348 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28525363 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28525351 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28525346 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28525340 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28525338 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28525347 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08097619 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28525354 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08097594 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08097595 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28525342 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P08097613 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28525353 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28525356 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08097624 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08097576 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08097577 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08097580 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08097615 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08097579 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08097602 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08097604 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08097635 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08097596 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08097594 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08097587 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08097612 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08097616 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08097621 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08097610 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08097628 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08097633 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08097631 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16513403 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08097614 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08097606 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08097600 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16513396 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16513400 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08097592 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08097590 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16513399 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08097637 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P08097583 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P39531607 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Genome U219 Array 11733693_PM_at - -
          Affymetrix Human Genome U219 Array 11733693_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR11014695.hg.1 ENST00000445202.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11014695.hg.1 NM_001129883.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11014695.hg.1 TC11001162.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1100009427.hg.1 ENST00000445202.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009427.hg.1 ENST00000445202.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009427.hg.1 ENST00000445202.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009427.hg.1 ENST00000445202.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009427.hg.1 ENST00000445202.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009427.hg.1 NM_001129883.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009427.hg.1 NM_001129883.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009427.hg.1 NM_001129883.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009427.hg.1 NM_001129883.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009427.hg.1 NM_001129883.3 Refseq
          Affymetrix Human Clariom D Assay TC1100009427.hg.1 ENST00000445202.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009427.hg.1 ENST00000445202.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009427.hg.1 ENST00000445202.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009427.hg.1 ENST00000445202.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009427.hg.1 ENST00000445202.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009427.hg.1 NM_001129883.3 Refseq
          Affymetrix Human Clariom D Assay TC1100009427.hg.1 NM_001129883.3 Refseq
          Affymetrix Human Clariom D Assay TC1100009427.hg.1 NM_001129883.3 Refseq
          Affymetrix Human Clariom D Assay TC1100009427.hg.1 NM_001129883.3 Refseq
          Affymetrix Human Clariom D Assay TC1100009427.hg.1 NM_001129883.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100170855.hg.1 ENST00000445202.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100170855.hg.1 NM_001129883.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100170855.hg.1 ENST00000445202.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100170855.hg.1 NM_001129883.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100170855.hg.1 TC1100009427.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1100170856.hg.1 ENST00000445202.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100170856.hg.1 NM_001129883.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100170856.hg.1 ENST00000445202.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100170856.hg.1 NM_001129883.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100170856.hg.1 TC1100009427.hg.1 Clariom D TC
          Human Genome U95 Set 77969_PM_at - -
          Human Genome U95 Set 77969_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A047110 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A047110 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 221228 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3238982 NM_001129883.3 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3238982 EF426753.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3238982 NM_001129883.3 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3238982 EF426753.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3238982 NM_001129883.3 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3238982 EF426753.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3238982 NM_001129883.3 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3238982 EF426753.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline