| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012804.hg.1 |
ENST00000631195.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012804.hg.1 |
ENST00000631195.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012804.hg.1 |
ENST00000631195.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012804.hg.1 |
ENST00000631195.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012804.hg.1 |
NR_120582.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012804.hg.1 |
NR_120582.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012804.hg.1 |
NR_120582.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012804.hg.1 |
NR_120582.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100198303.hg.1 |
ENST00000631195.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100198303.hg.1 |
NR_120582.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100198303.hg.1 |
ENST00000631195.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100198303.hg.1 |
NR_120582.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100198303.hg.1 |
TC1100012804.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1100012804.hg.1 |
ENST00000631195.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012804.hg.1 |
ENST00000631195.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012804.hg.1 |
ENST00000631195.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012804.hg.1 |
ENST00000631195.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012804.hg.1 |
NR_120582.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012804.hg.1 |
NR_120582.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012804.hg.1 |
NR_120582.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012804.hg.1 |
NR_120582.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100198302.hg.1 |
ENST00000631195.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100198302.hg.1 |
NR_120582.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100198302.hg.1 |
ENST00000631195.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100198302.hg.1 |
NR_120582.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100198302.hg.1 |
TC1100012804.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1100198304.hg.1 |
ENST00000631195.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100198304.hg.1 |
NR_120582.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100198304.hg.1 |
ENST00000631195.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100198304.hg.1 |
NR_120582.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100198304.hg.1 |
TC1100012804.hg.1 |
Clariom D TC |