Search
Search

LINC02145| Gene Symbol: LINC02145 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_028351.1 | 1 - 2 | 135 | 189 | ||
| GenBank mRNA | AK090679.1 | - | 1 - 2 | 135 | 189 | |
| BC132707.1 | - | 1 - 2 | 123 | 189 | ||
| BC132709.1 | - | 1 - 2 | 123 | 189 | ||
Set Membership: |
|
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P307724 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 3371 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 51627 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P01172386 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P01172423 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P17017578 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P307724 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_24_P307724 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23906744 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23906742 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23906754 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03740944 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03740971 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23906740 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23906764 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23906758 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23906751 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03740969 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23906781 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03740950 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23906725 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03740956 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03741001 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03740995 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23906723 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03740988 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15660433 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23906778 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23906752 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03740954 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23906765 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23906785 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23906757 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03740959 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03741008 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23906786 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23906761 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03740982 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03740948 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15660427 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23906753 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03740976 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03740946 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03740989 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23906732 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23906733 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03740992 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03741006 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03740994 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03740997 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03740949 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03741010 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23906760 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23906756 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03741010 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23906740 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23906765 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15660433 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03740992 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23906725 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03740997 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03740946 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23906785 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03740944 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03740969 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03740949 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15660427 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23906733 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23906754 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23906761 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23906742 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23906764 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23906732 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23906756 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03740959 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23906778 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23906786 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23906760 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23906758 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03740954 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23906753 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03741008 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23906757 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23906723 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03740976 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23906744 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23906752 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03740988 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23906781 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03740989 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03740982 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03741006 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03741001 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03740971 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03740950 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03740956 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23906751 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03740995 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03740948 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03740994 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_24_P307724 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 8110884 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8110885 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8110882 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 1563969_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11738463_at | - | - |
| Affymetrix Human Genome U219 Array | 11738462_at | - | - |
| Affymetrix Human Genome U219 Array | 11738463_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11738462_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR05016350.hg.1 | ENST00000507444.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05016350.hg.1 | NR_028351.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05016350.hg.1 | TC05001136.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009984.hg.1 | ENST00000502644.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009984.hg.1 | ENST00000502644.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009984.hg.1 | ENST00000502644.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009984.hg.1 | ENST00000507444.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009984.hg.1 | ENST00000507444.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009984.hg.1 | ENST00000507444.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009984.hg.1 | ENST00000507444.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009984.hg.1 | NR_028351.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009984.hg.1 | NR_028351.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009984.hg.1 | NR_028351.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009984.hg.1 | NR_028351.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05016351.hg.1 | ENST00000502644.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05016351.hg.1 | ENST00000507444.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05016351.hg.1 | NR_028351.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05016351.hg.1 | TC05001136.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05016348.hg.1 | ENST00000507444.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05016348.hg.1 | NR_028351.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05016348.hg.1 | TC05001136.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05016349.hg.1 | ENST00000507444.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05016349.hg.1 | NR_028351.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05016349.hg.1 | TC05001136.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0500009984.hg.1 | ENST00000502644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009984.hg.1 | ENST00000502644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009984.hg.1 | ENST00000502644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009984.hg.1 | ENST00000507444.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009984.hg.1 | ENST00000507444.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009984.hg.1 | ENST00000507444.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009984.hg.1 | ENST00000507444.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009984.hg.1 | NR_028351.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009984.hg.1 | NR_028351.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009984.hg.1 | NR_028351.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009984.hg.1 | NR_028351.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500170277.hg.1 | ENST00000502644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500170277.hg.1 | ENST00000507444.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500170277.hg.1 | NR_028351.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500170277.hg.1 | ENST00000502644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500170277.hg.1 | ENST00000507444.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500170277.hg.1 | NR_028351.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500170277.hg.1 | TC0500009984.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500170275.hg.1 | ENST00000507444.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500170275.hg.1 | NR_028351.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500170275.hg.1 | ENST00000507444.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500170275.hg.1 | NR_028351.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500170275.hg.1 | TC0500009984.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500170274.hg.1 | ENST00000507444.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500170274.hg.1 | NR_028351.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500170274.hg.1 | ENST00000507444.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500170274.hg.1 | NR_028351.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500170274.hg.1 | TC0500009984.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500170276.hg.1 | ENST00000507444.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500170276.hg.1 | NR_028351.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500170276.hg.1 | ENST00000507444.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500170276.hg.1 | NR_028351.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500170276.hg.1 | TC0500009984.hg.1 | Clariom D TC |
| Human Genome U133 Plus 2.0 Array | 1563969_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1563969_PM_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563969_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563969_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563969_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563969_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563969_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563969_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563969_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563969_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563969_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563969_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1563969_at8 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A026492 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A026492 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 496276 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1664563 | NR_028351.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1664563 | AK090679.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1664563 | NR_028351.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1664563 | AK090679.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1664563 | NR_028351.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1664563 | AK090679.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1664563 | NR_028351.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1664563 | AK090679.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1664563 | NR_028351.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1664563 | AK090679.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1664563 | NR_028351.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1664563 | AK090679.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1664563 | NR_028351.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1664563 | AK090679.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1664563 | NR_028351.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1664563 | AK090679.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1664563 | NR_028351.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1664563 | AK090679.1 | human_transcriptome |