Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01388168_m1
          See other LINC02405 GE Assays ›
          Gene Symbol:
          LINC02405
          Gene Name:
          long intergenic non-protein coding RNA 2405
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 6602 - 6921 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01388168_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LINC02405
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_104646.1 3 - 4 608 167
          GenBank mRNA BC032874.2 - 3 - 4 608 167

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1200012374.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Homo sapiens Probe spans exons

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1200012374.hg.1 ENST00000512624.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012374.hg.1 ENST00000512624.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012374.hg.1 ENST00000512624.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012374.hg.1 ENST00000512624.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012374.hg.1 ENST00000512624.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012374.hg.1 ENST00000512624.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012374.hg.1 ENST00000512624.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012374.hg.1 ENST00000512624.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012374.hg.1 ENST00000512624.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012374.hg.1 ENST00000512624.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012374.hg.1 ENST00000512624.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012374.hg.1 ENST00000512624.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012374.hg.1 ENST00000512624.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012374.hg.1 ENST00000512624.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012374.hg.1 ENST00000512624.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012374.hg.1 ENST00000512624.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012374.hg.1 ENST00000540244.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012374.hg.1 ENST00000540244.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012374.hg.1 ENST00000540244.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012374.hg.1 ENST00000540244.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012374.hg.1 ENST00000540244.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012374.hg.1 ENST00000540244.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012374.hg.1 ENST00000540244.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012374.hg.1 ENST00000540244.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012374.hg.1 ENST00000540244.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012374.hg.1 ENST00000540244.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012374.hg.1 ENST00000546062.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012374.hg.1 ENST00000546062.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012374.hg.1 ENST00000546062.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012374.hg.1 NR_104646.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012374.hg.1 NR_104646.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012374.hg.1 NR_104646.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012374.hg.1 NR_104646.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012374.hg.1 NR_104646.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012374.hg.1 NR_104646.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012374.hg.1 NR_104646.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200012374.hg.1 NR_104646.1 Refseq
          Affymetrix Human Clariom D Assay TC1200012374.hg.1 ENST00000512624.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200012374.hg.1 ENST00000512624.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200012374.hg.1 ENST00000512624.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200012374.hg.1 ENST00000512624.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200012374.hg.1 ENST00000512624.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200012374.hg.1 ENST00000512624.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200012374.hg.1 ENST00000512624.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200012374.hg.1 ENST00000512624.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200012374.hg.1 ENST00000512624.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200012374.hg.1 ENST00000512624.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200012374.hg.1 ENST00000512624.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200012374.hg.1 ENST00000512624.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200012374.hg.1 ENST00000512624.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200012374.hg.1 ENST00000512624.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200012374.hg.1 ENST00000512624.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200012374.hg.1 ENST00000512624.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200012374.hg.1 ENST00000540244.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200012374.hg.1 ENST00000540244.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200012374.hg.1 ENST00000540244.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200012374.hg.1 ENST00000540244.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200012374.hg.1 ENST00000540244.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200012374.hg.1 ENST00000540244.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200012374.hg.1 ENST00000540244.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200012374.hg.1 ENST00000540244.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200012374.hg.1 ENST00000540244.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200012374.hg.1 ENST00000540244.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200012374.hg.1 ENST00000546062.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200012374.hg.1 ENST00000546062.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200012374.hg.1 ENST00000546062.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200012374.hg.1 NR_104646.1 Refseq
          Affymetrix Human Clariom D Assay TC1200012374.hg.1 NR_104646.1 Refseq
          Affymetrix Human Clariom D Assay TC1200012374.hg.1 NR_104646.1 Refseq
          Affymetrix Human Clariom D Assay TC1200012374.hg.1 NR_104646.1 Refseq
          Affymetrix Human Clariom D Assay TC1200012374.hg.1 NR_104646.1 Refseq
          Affymetrix Human Clariom D Assay TC1200012374.hg.1 NR_104646.1 Refseq
          Affymetrix Human Clariom D Assay TC1200012374.hg.1 NR_104646.1 Refseq
          Affymetrix Human Clariom D Assay TC1200012374.hg.1 NR_104646.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200196899.hg.1 ENST00000512624.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1200196899.hg.1 ENST00000512624.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1200196899.hg.1 NR_104646.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200196899.hg.1 ENST00000512624.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1200196899.hg.1 ENST00000512624.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1200196899.hg.1 NR_104646.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200196899.hg.1 TC1200012374.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200196900.hg.1 ENST00000512624.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1200196900.hg.1 ENST00000512624.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1200196900.hg.1 NR_104646.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200196900.hg.1 ENST00000512624.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1200196900.hg.1 ENST00000512624.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1200196900.hg.1 NR_104646.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200196900.hg.1 TC1200012374.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200196901.hg.1 ENST00000512624.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1200196901.hg.1 ENST00000512624.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1200196901.hg.1 NR_104646.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200196901.hg.1 ENST00000512624.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1200196901.hg.1 ENST00000512624.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1200196901.hg.1 NR_104646.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200196901.hg.1 TC1200012374.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200196902.hg.1 ENST00000512624.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1200196902.hg.1 ENST00000512624.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1200196902.hg.1 ENST00000540244.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200196902.hg.1 ENST00000540244.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200196902.hg.1 NR_104646.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200196902.hg.1 ENST00000512624.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1200196902.hg.1 ENST00000512624.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1200196902.hg.1 ENST00000540244.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200196902.hg.1 ENST00000540244.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200196902.hg.1 NR_104646.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200196902.hg.1 TC1200012374.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline