Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01393973_m1
          See other KIAA1024L GE Assays ›
          Gene Symbol:
          KIAA1024L
          Gene Name:
          KIAA1024 like
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 129748191 - 129765063 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01393973_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: KIAA1024L
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001257308.1 NP_001244237.1 2 - 3 391 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0500008588.hg.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Homo sapiens 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Oligo Microarray 4x44K 39956 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 39254 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P600377 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array XM_001721301_at - -
          Affymetrix Human Transcriptome Array 2.0 TC0500008588.hg.1 ENST00000564719.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008588.hg.1 ENST00000564719.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008588.hg.1 ENST00000564719.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008588.hg.1 ENST00000564719.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008588.hg.1 ENST00000564719.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008588.hg.1 ENST00000564719.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008588.hg.1 ENST00000564719.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008588.hg.1 ENST00000564719.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008588.hg.1 NM_001257308.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500008588.hg.1 NM_001257308.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500008588.hg.1 NM_001257308.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500008588.hg.1 NM_001257308.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500008588.hg.1 NM_001257308.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500008588.hg.1 NM_001257308.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500158951.hg.1 ENST00000564719.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500158951.hg.1 NM_001257308.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500158951.hg.1 ENST00000564719.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500158951.hg.1 NM_001257308.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500158951.hg.1 TC0500008588.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0500158952.hg.1 ENST00000564719.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500158952.hg.1 NM_001257308.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500158952.hg.1 ENST00000564719.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500158952.hg.1 NM_001257308.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500158952.hg.1 TC0500008588.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0500008588.hg.1 ENST00000564719.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500008588.hg.1 ENST00000564719.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500008588.hg.1 ENST00000564719.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500008588.hg.1 ENST00000564719.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500008588.hg.1 ENST00000564719.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500008588.hg.1 ENST00000564719.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500008588.hg.1 ENST00000564719.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500008588.hg.1 ENST00000564719.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500008588.hg.1 NM_001257308.1 Refseq
          Affymetrix Human Clariom D Assay TC0500008588.hg.1 NM_001257308.1 Refseq
          Affymetrix Human Clariom D Assay TC0500008588.hg.1 NM_001257308.1 Refseq
          Affymetrix Human Clariom D Assay TC0500008588.hg.1 NM_001257308.1 Refseq
          Affymetrix Human Clariom D Assay TC0500008588.hg.1 NM_001257308.1 Refseq
          Affymetrix Human Clariom D Assay TC0500008588.hg.1 NM_001257308.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 225852 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1865663 DR731264.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3270834 DR731264.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1865663 DR731264.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1865663 DR731264.1 human_transcriptome
          Illumina Sentrix Human-6 v2 Expression BeadChip 5890056 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_134715 - -
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1865663 DR731264.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1865663 DR731264.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1865663 DR731264.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline