Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01394273_m1
          See other INSL3 GE Assays ›
          Gene Symbol:
          INSL3
          Gene Name:
          insulin like 3
          Gene Aliases:
          RLF, RLNL, ley-I-L
          Chromosome Location:
          Chr.19: 17816513 - 17821574 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01394273_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: INSL3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_005543.3 NP_005534.2 1 - 2 260 116
          GenBank mRNA AK302780.1 - 1 - 2 202 116
          BC032810.1 - 1 - 2 190 116
          BC040435.1 - 1 - 2 190 116
          BC053345.1 - 1 - 2 190 116
          BC071706.1 - 1 - 2 193 5167223 116
          BC106721.1 - 1 - 2 192 40014433 116
          BC106722.1 - 1 - 2 197 40014435 116
          S72482.1 - 1 - 2 199 116

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1900009989.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC19001279.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene INSL3

          Gene Ontology Categories:

          Function(s) Process(es)

          oocyte maturation
          adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway
          cell-cell signaling
          spermatogenesis
          positive regulation of cell proliferation
          negative regulation of cell proliferation
          male gonad development
          positive regulation of epithelial cell migration
          positive regulation of cAMP biosynthetic process
          negative regulation of apoptotic process
          positive regulation of wound healing
          G-protein coupled receptor binding
          protease binding
          receptor binding
          insulin receptor binding
          hormone activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR19018135.hg.1 ENST00000317306.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19018135.hg.1 ENST00000379695.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19018135.hg.1 NM_005543.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19018135.hg.1 TC19001279.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR19018137.hg.1 ENST00000317306.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19018137.hg.1 ENST00000379695.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19018137.hg.1 NM_005543.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19018137.hg.1 TC19001279.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR19018134.hg.1 ENST00000317306.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19018134.hg.1 ENST00000379695.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19018134.hg.1 NM_005543.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19018134.hg.1 TC19001279.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1900009989.hg.1 ENST00000317306.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009989.hg.1 ENST00000317306.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009989.hg.1 ENST00000317306.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009989.hg.1 ENST00000317306.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009989.hg.1 ENST00000379695.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009989.hg.1 ENST00000379695.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009989.hg.1 ENST00000379695.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009989.hg.1 ENST00000379695.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009989.hg.1 ENST00000379695.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009989.hg.1 ENST00000379695.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009989.hg.1 ENST00000598577.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009989.hg.1 ENST00000598577.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009989.hg.1 ENST00000598577.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009989.hg.1 ENST00000598577.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009989.hg.1 NM_001265587.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900009989.hg.1 NM_001265587.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900009989.hg.1 NM_001265587.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900009989.hg.1 NM_001265587.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900009989.hg.1 NM_001265587.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900009989.hg.1 NM_001265587.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900009989.hg.1 NM_005543.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900009989.hg.1 NM_005543.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900009989.hg.1 NM_005543.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900009989.hg.1 NM_005543.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900009989.hg.1 NM_005543.3 Refseq
          Affymetrix Human Clariom D Assay PSR1900181975.hg.1 ENST00000317306.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1900181975.hg.1 ENST00000379695.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900181975.hg.1 ENST00000598577.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900181975.hg.1 NM_001265587.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900181975.hg.1 NM_005543.3 Refseq
          Affymetrix Human Clariom D Assay PSR1900181975.hg.1 TC1900009989.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900181975.hg.1 ENST00000317306.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1900181975.hg.1 ENST00000379695.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900181975.hg.1 ENST00000598577.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900181975.hg.1 NM_001265587.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900181975.hg.1 NM_005543.3 Refseq
          Affymetrix Human Clariom D Assay TC1900009989.hg.1 ENST00000317306.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900009989.hg.1 ENST00000317306.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900009989.hg.1 ENST00000317306.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900009989.hg.1 ENST00000317306.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900009989.hg.1 ENST00000379695.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900009989.hg.1 ENST00000379695.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900009989.hg.1 ENST00000379695.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900009989.hg.1 ENST00000379695.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900009989.hg.1 ENST00000379695.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900009989.hg.1 ENST00000379695.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900009989.hg.1 ENST00000598577.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009989.hg.1 ENST00000598577.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009989.hg.1 ENST00000598577.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009989.hg.1 ENST00000598577.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009989.hg.1 NM_001265587.1 Refseq
          Affymetrix Human Clariom D Assay TC1900009989.hg.1 NM_001265587.1 Refseq
          Affymetrix Human Clariom D Assay TC1900009989.hg.1 NM_001265587.1 Refseq
          Affymetrix Human Clariom D Assay TC1900009989.hg.1 NM_001265587.1 Refseq
          Affymetrix Human Clariom D Assay TC1900009989.hg.1 NM_001265587.1 Refseq
          Affymetrix Human Clariom D Assay TC1900009989.hg.1 NM_001265587.1 Refseq
          Affymetrix Human Clariom D Assay TC1900009989.hg.1 NM_005543.3 Refseq
          Affymetrix Human Clariom D Assay TC1900009989.hg.1 NM_005543.3 Refseq
          Affymetrix Human Clariom D Assay TC1900009989.hg.1 NM_005543.3 Refseq
          Affymetrix Human Clariom D Assay TC1900009989.hg.1 NM_005543.3 Refseq
          Affymetrix Human Clariom D Assay TC1900009989.hg.1 NM_005543.3 Refseq
          Affymetrix Human Clariom D Assay PSR1900181972.hg.1 ENST00000317306.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1900181972.hg.1 ENST00000379695.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900181972.hg.1 ENST00000598577.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900181972.hg.1 NM_001265587.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900181972.hg.1 NM_005543.3 Refseq
          Affymetrix Human Clariom D Assay PSR1900181972.hg.1 TC1900009989.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900181972.hg.1 ENST00000317306.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1900181972.hg.1 ENST00000379695.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900181972.hg.1 ENST00000598577.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900181972.hg.1 NM_001265587.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900181972.hg.1 NM_005543.3 Refseq
          Affymetrix Human Clariom D Assay PSR1900181980.hg.1 ENST00000379695.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900181980.hg.1 NM_001265587.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900181980.hg.1 NM_005543.3 Refseq
          Affymetrix Human Clariom D Assay PSR1900181980.hg.1 TC1900009989.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900181980.hg.1 ENST00000379695.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900181980.hg.1 NM_001265587.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900181980.hg.1 NM_005543.3 Refseq
          Affymetrix Human Clariom D Assay PSR1900181970.hg.1 ENST00000317306.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1900181970.hg.1 ENST00000379695.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900181970.hg.1 ENST00000598577.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900181970.hg.1 NM_001265587.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900181970.hg.1 NM_005543.3 Refseq
          Affymetrix Human Clariom D Assay PSR1900181970.hg.1 TC1900009989.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900181970.hg.1 ENST00000317306.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1900181970.hg.1 ENST00000379695.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900181970.hg.1 ENST00000598577.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900181970.hg.1 NM_001265587.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900181970.hg.1 NM_005543.3 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 635630 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline