| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04005406.hg.1 |
ENST00000309395.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04005406.hg.1 |
ENST00000396051.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04005406.hg.1 |
NM_001244766.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04005406.hg.1 |
NM_001244767.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04005406.hg.1 |
NM_173468.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04005406.hg.1 |
TC04002912.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04005404.hg.1 |
ENST00000396051.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04005404.hg.1 |
NM_001244766.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04005404.hg.1 |
TC04002912.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012810.hg.1 |
ENST00000309395.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012810.hg.1 |
ENST00000309395.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012810.hg.1 |
ENST00000309395.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012810.hg.1 |
ENST00000309395.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012810.hg.1 |
ENST00000309395.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012810.hg.1 |
ENST00000309395.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012810.hg.1 |
ENST00000309395.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012810.hg.1 |
ENST00000309395.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012810.hg.1 |
ENST00000309395.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012810.hg.1 |
ENST00000309395.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012810.hg.1 |
ENST00000396051.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012810.hg.1 |
ENST00000396051.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012810.hg.1 |
ENST00000396051.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012810.hg.1 |
ENST00000396051.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012810.hg.1 |
ENST00000396051.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012810.hg.1 |
ENST00000396051.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012810.hg.1 |
ENST00000396051.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012810.hg.1 |
ENST00000396051.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012810.hg.1 |
ENST00000396051.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012810.hg.1 |
ENST00000396051.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012810.hg.1 |
ENST00000502869.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012810.hg.1 |
ENST00000502869.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012810.hg.1 |
ENST00000502869.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012810.hg.1 |
ENST00000502869.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012810.hg.1 |
ENST00000502869.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012810.hg.1 |
ENST00000502869.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012810.hg.1 |
ENST00000502869.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012810.hg.1 |
ENST00000502869.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012810.hg.1 |
ENST00000509041.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012810.hg.1 |
ENST00000509041.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012810.hg.1 |
ENST00000509041.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012810.hg.1 |
ENST00000509041.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012810.hg.1 |
ENST00000509041.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012810.hg.1 |
ENST00000509041.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012810.hg.1 |
ENST00000509041.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012810.hg.1 |
NM_001244766.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012810.hg.1 |
NM_001244766.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012810.hg.1 |
NM_001244766.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012810.hg.1 |
NM_001244766.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012810.hg.1 |
NM_001244766.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012810.hg.1 |
NM_001244766.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012810.hg.1 |
NM_001244766.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012810.hg.1 |
NM_001244766.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012810.hg.1 |
NM_001244766.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012810.hg.1 |
NM_001244766.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012810.hg.1 |
NM_001244766.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012810.hg.1 |
NM_001244766.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012810.hg.1 |
NM_001244767.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012810.hg.1 |
NM_001244767.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012810.hg.1 |
NM_001244767.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012810.hg.1 |
NM_001244767.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012810.hg.1 |
NM_001244767.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012810.hg.1 |
NM_001244767.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012810.hg.1 |
NM_001244767.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012810.hg.1 |
NM_173468.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012810.hg.1 |
NM_173468.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012810.hg.1 |
NM_173468.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012810.hg.1 |
NM_173468.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012810.hg.1 |
NM_173468.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012810.hg.1 |
NM_173468.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012810.hg.1 |
NM_173468.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012810.hg.1 |
NM_173468.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012810.hg.1 |
NM_173468.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012810.hg.1 |
NM_173468.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012810.hg.1 |
ENST00000309395.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012810.hg.1 |
ENST00000309395.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012810.hg.1 |
ENST00000309395.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012810.hg.1 |
ENST00000309395.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012810.hg.1 |
ENST00000309395.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012810.hg.1 |
ENST00000309395.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012810.hg.1 |
ENST00000309395.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012810.hg.1 |
ENST00000309395.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012810.hg.1 |
ENST00000309395.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012810.hg.1 |
ENST00000309395.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012810.hg.1 |
ENST00000396051.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012810.hg.1 |
ENST00000396051.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012810.hg.1 |
ENST00000396051.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012810.hg.1 |
ENST00000396051.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012810.hg.1 |
ENST00000396051.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012810.hg.1 |
ENST00000396051.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012810.hg.1 |
ENST00000396051.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012810.hg.1 |
ENST00000396051.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012810.hg.1 |
ENST00000396051.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012810.hg.1 |
ENST00000396051.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012810.hg.1 |
ENST00000502869.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012810.hg.1 |
ENST00000502869.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012810.hg.1 |
ENST00000502869.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012810.hg.1 |
ENST00000502869.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012810.hg.1 |
ENST00000502869.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012810.hg.1 |
ENST00000502869.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012810.hg.1 |
ENST00000502869.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012810.hg.1 |
ENST00000502869.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012810.hg.1 |
ENST00000509041.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012810.hg.1 |
ENST00000509041.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012810.hg.1 |
ENST00000509041.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012810.hg.1 |
ENST00000509041.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012810.hg.1 |
ENST00000509041.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012810.hg.1 |
ENST00000509041.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012810.hg.1 |
ENST00000509041.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012810.hg.1 |
NM_001244766.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012810.hg.1 |
NM_001244766.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012810.hg.1 |
NM_001244766.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012810.hg.1 |
NM_001244766.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012810.hg.1 |
NM_001244766.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012810.hg.1 |
NM_001244766.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012810.hg.1 |
NM_001244766.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012810.hg.1 |
NM_001244766.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012810.hg.1 |
NM_001244766.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012810.hg.1 |
NM_001244766.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012810.hg.1 |
NM_001244766.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012810.hg.1 |
NM_001244766.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012810.hg.1 |
NM_001244767.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012810.hg.1 |
NM_001244767.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012810.hg.1 |
NM_001244767.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012810.hg.1 |
NM_001244767.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012810.hg.1 |
NM_001244767.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012810.hg.1 |
NM_001244767.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012810.hg.1 |
NM_001244767.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012810.hg.1 |
NM_173468.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012810.hg.1 |
NM_173468.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012810.hg.1 |
NM_173468.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012810.hg.1 |
NM_173468.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012810.hg.1 |
NM_173468.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012810.hg.1 |
NM_173468.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012810.hg.1 |
NM_173468.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012810.hg.1 |
NM_173468.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012810.hg.1 |
NM_173468.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012810.hg.1 |
NM_173468.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400188437.hg.1 |
ENST00000396051.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400188437.hg.1 |
ENST00000502869.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400188437.hg.1 |
NM_001244766.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400188437.hg.1 |
ENST00000396051.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400188437.hg.1 |
ENST00000502869.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400188437.hg.1 |
NM_001244766.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400188437.hg.1 |
TC0400012810.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0400188441.hg.1 |
ENST00000309395.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400188441.hg.1 |
ENST00000396051.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400188441.hg.1 |
ENST00000502869.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400188441.hg.1 |
ENST00000509041.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400188441.hg.1 |
NM_001244766.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400188441.hg.1 |
NM_001244767.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400188441.hg.1 |
NM_173468.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400188441.hg.1 |
ENST00000309395.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400188441.hg.1 |
ENST00000396051.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400188441.hg.1 |
ENST00000502869.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400188441.hg.1 |
ENST00000509041.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400188441.hg.1 |
NM_001244766.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400188441.hg.1 |
NM_001244767.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400188441.hg.1 |
NM_173468.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400188441.hg.1 |
TC0400012810.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0400188438.hg.1 |
ENST00000396051.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400188438.hg.1 |
ENST00000502869.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400188438.hg.1 |
NM_001244766.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400188438.hg.1 |
ENST00000396051.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400188438.hg.1 |
ENST00000502869.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400188438.hg.1 |
NM_001244766.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400188438.hg.1 |
TC0400012810.hg.1 |
Clariom D TC |